Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116528933-116529435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM26D
Entrez Gene ID
221301
Gene Name
family with sequence similarity 26 member D
Gene Aliases
C6orf78
UniGene
Hs.350750
Gene Chromosome Location
Chr.6: 116529013-116561123 on Build GRCh38
Transcripts
NM_001256887.2 NM_001256888.2 NM_001256889.2 NM_153036.4 XM_011535560.2 XM_011535561.2 XM_011535562.2 XM_011535563.2 XM_017010390.1 XM_017010391.1 XM_017010392.1 NM_001139444.2

Gene Symbol
TRAPPC3L
Entrez Gene ID
100128327
Gene Name
trafficking protein particle complex 3 like
Gene Aliases
BET3L, bA259P20.2
UniGene
Hs.134795
Gene Chromosome Location
Chr.6: 116496483-116545610 on Build GRCh38
Transcripts
NM_001256887.2 NM_001256888.2 NM_001256889.2 NM_153036.4 XM_011535560.2 XM_011535561.2 XM_011535562.2 XM_011535563.2 XM_017010390.1 XM_017010391.1 XM_017010392.1 NM_001139444.2

SNP Detail

SNP ID Chromosome Location
rs927535747 Chr.6: 116529011-116529011 on Build GRCh38
rs577772852 Chr.6: 116529061-116529061 on Build GRCh38
rs757873774 Chr.6: 116529187-116529187 on Build GRCh38
rs376256717 Chr.6: 116529378-116529386 on Build GRCh38
rs958933739 Chr.6: 116529044-116529044 on Build GRCh38
rs993109987 Chr.6: 116529311-116529311 on Build GRCh38
rs754176847 Chr.6: 116529112-116529112 on Build GRCh38
rs767990068 Chr.6: 116529070-116529070 on Build GRCh38
rs55857439 Chr.6: 116529071-116529071 on Build GRCh38
rs186610997 Chr.6: 116529129-116529129 on Build GRCh38
rs868106686 Chr.6: 116529056-116529056 on Build GRCh38
rs1009665045 Chr.6: 116529314-116529314 on Build GRCh38
rs758042532 Chr.6: 116529362-116529362 on Build GRCh38
rs933364797 Chr.6: 116529184-116529184 on Build GRCh38
rs139410278 Chr.6: 116529188-116529188 on Build GRCh38
rs762089622 Chr.6: 116528968-116528968 on Build GRCh38
rs1040310200 Chr.6: 116529387-116529387 on Build GRCh38
rs1005587799 Chr.6: 116529255-116529255 on Build GRCh38
rs78023416 Chr.6: 116529010-116529010 on Build GRCh38
rs993443323 Chr.6: 116529066-116529066 on Build GRCh38
rs555773355 Chr.6: 116529391-116529391 on Build GRCh38
rs34776007 Chr.6: 116529168-116529168 on Build GRCh38
rs1021082428 Chr.6: 116529014-116529014 on Build GRCh38
rs1033626629 Chr.6: 116529111-116529111 on Build GRCh38
rs149099354 Chr.6: 116529376-116529384 on Build GRCh38
rs993712583 Chr.6: 116529236-116529236 on Build GRCh38
rs530410075 Chr.6: 116529395-116529395 on Build GRCh38
rs11969533 Chr.6: 116529297-116529297 on Build GRCh38
rs1025728879 Chr.6: 116529241-116529241 on Build GRCh38
rs377303770 Chr.6: 116529113-116529113 on Build GRCh38
rs952738618 Chr.6: 116529247-116529247 on Build GRCh38
rs1033756284 Chr.6: 116529346-116529346 on Build GRCh38
rs958994443 Chr.6: 116529355-116529355 on Build GRCh38
rs917468682 Chr.6: 116529318-116529318 on Build GRCh38
rs985784256 Chr.6: 116529364-116529364 on Build GRCh38
rs757066491 Chr.6: 116529408-116529408 on Build GRCh38
rs748664351 Chr.6: 116529173-116529173 on Build GRCh38
rs188299013 Chr.6: 116529368-116529368 on Build GRCh38
rs1034346319 Chr.6: 116529281-116529281 on Build GRCh38
rs1003251686 Chr.6: 116529338-116529338 on Build GRCh38
rs191393259 Chr.6: 116529226-116529226 on Build GRCh38
rs943614599 Chr.6: 116529293-116529293 on Build GRCh38
rs1052179689 Chr.6: 116529240-116529240 on Build GRCh38
rs548545970 Chr.6: 116529149-116529149 on Build GRCh38
rs925739364 Chr.6: 116529356-116529356 on Build GRCh38
rs183980152 Chr.6: 116529319-116529319 on Build GRCh38
rs925657068 Chr.6: 116529119-116529119 on Build GRCh38
rs752176969 Chr.6: 116529185-116529185 on Build GRCh38
rs946179533 Chr.6: 116529073-116529073 on Build GRCh38
rs954955385 Chr.6: 116529399-116529399 on Build GRCh38
rs192810526 Chr.6: 116529378-116529378 on Build GRCh38
rs1041487811 Chr.6: 116529089-116529089 on Build GRCh38
rs970434689 Chr.6: 116529032-116529032 on Build GRCh38
rs929162990 Chr.6: 116529405-116529405 on Build GRCh38
rs544403890 Chr.6: 116529003-116529003 on Build GRCh38
rs552045851 Chr.6: 116529321-116529321 on Build GRCh38
rs56056252 Chr.6: 116529145-116529145 on Build GRCh38

View Full Product Details