Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118468979-118469521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP85L
Entrez Gene ID
387119
Gene Name
centrosomal protein 85 like
Gene Aliases
C6orf204, NY-BR-15, bA57K17.2
UniGene
Hs.656959
Gene Chromosome Location
Chr.6: 118460772-118710101 on Build GRCh38
Transcripts
NM_001042475.2 NM_001178035.1 XM_005266970.1 XM_005266971.1 XM_005266972.4 XM_006715475.3 XM_011535809.2 XM_011535810.2 XM_011535811.1 XM_017010846.1 XM_017010847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5671409 c.2220G>C p.K740N Chr.6: 118469106-118469106 on Build GRCh38
1595784 c.2147A>G p.D716G Chr.6: 118469188-118469188 on Build GRCh38
7121455 c.2111G>C p.R704P Chr.6: 118469215-118469215 on Build GRCh38
4879534 c.2232G>A p.E744E Chr.6: 118469103-118469103 on Build GRCh38
1132047 c.2046A>C p.T682T Chr.6: 118469280-118469280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046849683 Chr.6: 118469491-118469491 on Build GRCh38
rs370313035 Chr.6: 118469078-118469078 on Build GRCh38
rs202174000 Chr.6: 118469208-118469208 on Build GRCh38
rs748051039 Chr.6: 118469108-118469108 on Build GRCh38
rs770361555 Chr.6: 118469465-118469465 on Build GRCh38
rs1053480353 Chr.6: 118469313-118469313 on Build GRCh38
rs906225919 Chr.6: 118469461-118469461 on Build GRCh38
rs754444194 Chr.6: 118469100-118469100 on Build GRCh38
rs780805462 Chr.6: 118469229-118469229 on Build GRCh38
rs777753792 Chr.6: 118469122-118469122 on Build GRCh38
rs772766278 Chr.6: 118469442-118469442 on Build GRCh38
rs748223615 Chr.6: 118469321-118469321 on Build GRCh38
rs185732895 Chr.6: 118469103-118469103 on Build GRCh38
rs765952639 Chr.6: 118469222-118469222 on Build GRCh38
rs201806484 Chr.6: 118469216-118469216 on Build GRCh38
rs767819980 Chr.6: 118469036-118469036 on Build GRCh38
rs375443089 Chr.6: 118469307-118469307 on Build GRCh38
rs751184064 Chr.6: 118469224-118469224 on Build GRCh38
rs749094256 Chr.6: 118469129-118469129 on Build GRCh38
rs141587519 Chr.6: 118469060-118469060 on Build GRCh38
rs752231171 Chr.6: 118469238-118469238 on Build GRCh38
rs1025801080 Chr.6: 118469211-118469211 on Build GRCh38
rs766724962 Chr.6: 118469340-118469340 on Build GRCh38
rs190588573 Chr.6: 118469458-118469458 on Build GRCh38
rs777845684 Chr.6: 118469249-118469249 on Build GRCh38
rs774738796 Chr.6: 118469028-118469028 on Build GRCh38
rs1046304410 Chr.6: 118469272-118469272 on Build GRCh38
rs959034546 Chr.6: 118469472-118469472 on Build GRCh38
rs762184975 Chr.6: 118469056-118469056 on Build GRCh38
rs759888437 Chr.6: 118469032-118469032 on Build GRCh38
rs368068619 Chr.6: 118469140-118469140 on Build GRCh38
rs368501966 Chr.6: 118469221-118469221 on Build GRCh38
rs780174451 Chr.6: 118469288-118469288 on Build GRCh38
rs572576749 Chr.6: 118469380-118469381 on Build GRCh38
rs1042627122 Chr.6: 118469015-118469015 on Build GRCh38
rs754470866 Chr.6: 118469228-118469228 on Build GRCh38
rs115332958 Chr.6: 118469013-118469013 on Build GRCh38
rs776596138 Chr.6: 118469334-118469334 on Build GRCh38
rs1032068976 Chr.6: 118469017-118469017 on Build GRCh38
rs77465615 Chr.6: 118469217-118469217 on Build GRCh38
rs763330230 Chr.6: 118469329-118469329 on Build GRCh38
rs746370053 Chr.6: 118469120-118469120 on Build GRCh38
rs774546415 Chr.6: 118469350-118469350 on Build GRCh38
rs371610026 Chr.6: 118469119-118469119 on Build GRCh38
rs765726985 Chr.6: 118469215-118469215 on Build GRCh38
rs1025861878 Chr.6: 118469112-118469112 on Build GRCh38
rs569828141 Chr.6: 118469446-118469446 on Build GRCh38
rs747299548 Chr.6: 118469292-118469292 on Build GRCh38
rs754327334 Chr.6: 118469055-118469055 on Build GRCh38
rs751043572 Chr.6: 118469090-118469090 on Build GRCh38
rs770401587 Chr.6: 118469150-118469150 on Build GRCh38
rs770714469 Chr.6: 118469271-118469271 on Build GRCh38
rs371812819 Chr.6: 118469178-118469178 on Build GRCh38
rs75680069 Chr.6: 118469450-118469450 on Build GRCh38
rs137896316 Chr.6: 118469447-118469447 on Build GRCh38
rs977558330 Chr.6: 118469362-118469362 on Build GRCh38
rs772483577 Chr.6: 118469134-118469134 on Build GRCh38
rs775503333 Chr.6: 118469037-118469037 on Build GRCh38
rs115966151 Chr.6: 118469423-118469423 on Build GRCh38
rs71559830 Chr.6: 118469480-118469480 on Build GRCh38
rs562827924 Chr.6: 118469466-118469466 on Build GRCh38
rs146186036 Chr.6: 118469389-118469389 on Build GRCh38
rs770290333 Chr.6: 118469322-118469322 on Build GRCh38
rs991554091 Chr.6: 118469475-118469475 on Build GRCh38
rs200864764 Chr.6: 118469212-118469212 on Build GRCh38
rs760800208 Chr.6: 118469041-118469041 on Build GRCh38
rs993901506 Chr.6: 118469210-118469210 on Build GRCh38
rs567310018 Chr.6: 118469388-118469388 on Build GRCh38
rs749096201 Chr.6: 118469269-118469269 on Build GRCh38
rs760696954 Chr.6: 118469142-118469142 on Build GRCh38
rs758111625 Chr.6: 118469073-118469073 on Build GRCh38
rs371892462 Chr.6: 118469242-118469242 on Build GRCh38
rs1040269742 Chr.6: 118469382-118469382 on Build GRCh38
rs551300699 Chr.6: 118469051-118469051 on Build GRCh38
rs989242128 Chr.6: 118469393-118469393 on Build GRCh38
rs773651202 Chr.6: 118469328-118469328 on Build GRCh38
rs747798332 Chr.6: 118469019-118469019 on Build GRCh38

View Full Product Details