Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118893870-118894376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASF1A
Entrez Gene ID
25842
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
CGI-98, CIA, HSPC146
UniGene
Hs.292316
Gene Chromosome Location
Chr.6: 118894077-118909171 on Build GRCh38
Transcripts
NM_014034.2 NM_017696.2

Gene Symbol
MCM9
Entrez Gene ID
254394
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Gene Aliases
C6orf61, MCMDC1, ODG4, dJ329L24.1, dJ329L24.3
UniGene
Hs.279008
Gene Chromosome Location
Chr.6: 118813449-118935162 on Build GRCh38
Transcripts
NM_014034.2 NM_017696.2

SNP Detail

SNP ID Chromosome Location
rs1029007458 Chr.6: 118894259-118894259 on Build GRCh38
rs1042525363 Chr.6: 118894315-118894315 on Build GRCh38
rs758742292 Chr.6: 118894257-118894257 on Build GRCh38
rs988642142 Chr.6: 118894322-118894322 on Build GRCh38
rs565660986 Chr.6: 118894215-118894215 on Build GRCh38
rs931219561 Chr.6: 118893923-118893941 on Build GRCh38
rs963495611 Chr.6: 118894237-118894237 on Build GRCh38
rs867283445 Chr.6: 118893922-118893922 on Build GRCh38
rs568349177 Chr.6: 118893961-118893961 on Build GRCh38
rs903847191 Chr.6: 118894323-118894323 on Build GRCh38
rs891528762 Chr.6: 118893895-118893895 on Build GRCh38
rs1015039995 Chr.6: 118893962-118893962 on Build GRCh38
rs921925512 Chr.6: 118893980-118893980 on Build GRCh38
rs1021700269 Chr.6: 118893919-118893919 on Build GRCh38
rs1002152051 Chr.6: 118893929-118893929 on Build GRCh38
rs568505676 Chr.6: 118893931-118893931 on Build GRCh38
rs970920926 Chr.6: 118893935-118893935 on Build GRCh38
rs370372401 Chr.6: 118894274-118894274 on Build GRCh38
rs1018739356 Chr.6: 118894224-118894224 on Build GRCh38
rs912508743 Chr.6: 118894303-118894303 on Build GRCh38
rs574624424 Chr.6: 118894264-118894264 on Build GRCh38
rs1057001350 Chr.6: 118894301-118894301 on Build GRCh38
rs963444856 Chr.6: 118893965-118893965 on Build GRCh38
rs375387719 Chr.6: 118893905-118893905 on Build GRCh38
rs953233553 Chr.6: 118894208-118894208 on Build GRCh38
rs903879622 Chr.6: 118894156-118894156 on Build GRCh38
rs558559674 Chr.6: 118894193-118894193 on Build GRCh38
rs191321515 Chr.6: 118893891-118893891 on Build GRCh38
rs534746521 Chr.6: 118894218-118894218 on Build GRCh38
rs985053161 Chr.6: 118894278-118894278 on Build GRCh38
rs142622007 Chr.6: 118894325-118894325 on Build GRCh38
rs912413618 Chr.6: 118894020-118894021 on Build GRCh38
rs980904233 Chr.6: 118893940-118893940 on Build GRCh38
rs891579412 Chr.6: 118894081-118894081 on Build GRCh38
rs926999319 Chr.6: 118894352-118894352 on Build GRCh38
rs986328027 Chr.6: 118894210-118894210 on Build GRCh38
rs372985609 Chr.6: 118894001-118894001 on Build GRCh38
rs1033374855 Chr.6: 118894197-118894197 on Build GRCh38
rs1049804233 Chr.6: 118894071-118894071 on Build GRCh38
rs778414808 Chr.6: 118894296-118894296 on Build GRCh38
rs1052868431 Chr.6: 118894063-118894063 on Build GRCh38
rs932592045 Chr.6: 118893994-118893994 on Build GRCh38
rs938212928 Chr.6: 118894098-118894098 on Build GRCh38
rs1011268316 Chr.6: 118893906-118893906 on Build GRCh38
rs1009667577 Chr.6: 118894125-118894125 on Build GRCh38
rs945424646 Chr.6: 118894311-118894311 on Build GRCh38
rs796862501 Chr.6: 118893921-118893922 on Build GRCh38
rs926898423 Chr.6: 118893950-118893950 on Build GRCh38
rs11542663 Chr.6: 118894238-118894238 on Build GRCh38
rs937029927 Chr.6: 118894355-118894355 on Build GRCh38
rs1132733 Chr.6: 118894194-118894194 on Build GRCh38
rs973463131 Chr.6: 118893975-118893975 on Build GRCh38
rs888996296 Chr.6: 118894332-118894332 on Build GRCh38
rs947120888 Chr.6: 118894085-118894085 on Build GRCh38
rs1007486664 Chr.6: 118894349-118894349 on Build GRCh38
rs926907132 Chr.6: 118894293-118894293 on Build GRCh38
rs760464761 Chr.6: 118894077-118894078 on Build GRCh38
rs918107330 Chr.6: 118894305-118894305 on Build GRCh38
rs144606482 Chr.6: 118894053-118894053 on Build GRCh38
rs538645481 Chr.6: 118894127-118894127 on Build GRCh38
rs1001809130 Chr.6: 118894126-118894126 on Build GRCh38
rs182572792 Chr.6: 118894103-118894103 on Build GRCh38
rs896534092 Chr.6: 118894117-118894117 on Build GRCh38
rs918907078 Chr.6: 118894242-118894242 on Build GRCh38
rs905349686 Chr.6: 118894072-118894072 on Build GRCh38
rs971807401 Chr.6: 118894240-118894240 on Build GRCh38
rs960168868 Chr.6: 118893957-118893958 on Build GRCh38
rs972168438 Chr.6: 118893926-118893926 on Build GRCh38
rs1049751293 Chr.6: 118894329-118894329 on Build GRCh38
rs372743698 Chr.6: 118893937-118893937 on Build GRCh38
rs1000104029 Chr.6: 118894157-118894157 on Build GRCh38
rs557229643 Chr.6: 118894343-118894343 on Build GRCh38

View Full Product Details