Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118975917-118976334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184A
Entrez Gene ID
79632
Gene Name
family with sequence similarity 184 member A
Gene Aliases
C6orf60
UniGene
Hs.443789
Gene Chromosome Location
Chr.6: 118959829-119149193 on Build GRCh38
Transcripts
NM_001100411.2 NM_001288576.1 NM_024581.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4825159 c.2524C>T p.Q842* Chr.6: 118975976-118975976 on Build GRCh38
4421265 c.2498T>G p.I833S Chr.6: 118976002-118976002 on Build GRCh38
7367200 c.2522A>T p.H841L Chr.6: 118975978-118975978 on Build GRCh38
7237318 c.2518C>T p.H840Y Chr.6: 118975982-118975982 on Build GRCh38
3828932 c.2527G>T p.E843* Chr.6: 118975973-118975973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771427640 Chr.6: 118975956-118975956 on Build GRCh38
rs919092952 Chr.6: 118976165-118976165 on Build GRCh38
rs904018698 Chr.6: 118976088-118976088 on Build GRCh38
rs910304938 Chr.6: 118976310-118976310 on Build GRCh38
rs768169717 Chr.6: 118976026-118976026 on Build GRCh38
rs760254903 Chr.6: 118976020-118976020 on Build GRCh38
rs750310450 Chr.6: 118976036-118976036 on Build GRCh38
rs377472426 Chr.6: 118976032-118976032 on Build GRCh38
rs41292558 Chr.6: 118975978-118975978 on Build GRCh38
rs879071991 Chr.6: 118976148-118976148 on Build GRCh38
rs985892854 Chr.6: 118976291-118976291 on Build GRCh38
rs930425012 Chr.6: 118976178-118976178 on Build GRCh38
rs569965528 Chr.6: 118976265-118976265 on Build GRCh38
rs984557725 Chr.6: 118976215-118976215 on Build GRCh38
rs1033775218 Chr.6: 118976104-118976104 on Build GRCh38
rs1000905754 Chr.6: 118975940-118975940 on Build GRCh38
rs896521715 Chr.6: 118976124-118976124 on Build GRCh38
rs766207121 Chr.6: 118976053-118976053 on Build GRCh38
rs371155850 Chr.6: 118975972-118975972 on Build GRCh38
rs190868190 Chr.6: 118976078-118976078 on Build GRCh38
rs536572230 Chr.6: 118976060-118976060 on Build GRCh38
rs537413097 Chr.6: 118976276-118976276 on Build GRCh38
rs757260892 Chr.6: 118976086-118976086 on Build GRCh38
rs562961238 Chr.6: 118976108-118976108 on Build GRCh38
rs1043948375 Chr.6: 118976071-118976071 on Build GRCh38
rs758096895 Chr.6: 118976116-118976116 on Build GRCh38
rs937724906 Chr.6: 118976246-118976246 on Build GRCh38
rs867152595 Chr.6: 118976210-118976210 on Build GRCh38
rs1013605272 Chr.6: 118976201-118976204 on Build GRCh38
rs762867874 Chr.6: 118976044-118976044 on Build GRCh38
rs1028339616 Chr.6: 118975954-118975954 on Build GRCh38
rs772310108 Chr.6: 118975991-118975991 on Build GRCh38
rs761690996 Chr.6: 118976034-118976034 on Build GRCh38
rs553055093 Chr.6: 118976232-118976233 on Build GRCh38
rs749833051 Chr.6: 118975955-118975955 on Build GRCh38
rs751960947 Chr.6: 118976278-118976278 on Build GRCh38
rs754009816 Chr.6: 118976085-118976085 on Build GRCh38
rs756352859 Chr.6: 118976061-118976061 on Build GRCh38
rs573673457 Chr.6: 118975995-118975995 on Build GRCh38
rs752977495 Chr.6: 118976057-118976057 on Build GRCh38
rs370372665 Chr.6: 118976035-118976035 on Build GRCh38
rs758408611 Chr.6: 118975975-118975977 on Build GRCh38
rs951874502 Chr.6: 118976287-118976287 on Build GRCh38
rs764957196 Chr.6: 118976011-118976011 on Build GRCh38
rs1002322240 Chr.6: 118976092-118976092 on Build GRCh38
rs6915914 Chr.6: 118976220-118976220 on Build GRCh38
rs986793264 Chr.6: 118975990-118975990 on Build GRCh38
rs547638773 Chr.6: 118976279-118976279 on Build GRCh38

View Full Product Details