Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 123055697-123056172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLVS2
Entrez Gene ID
134829
Gene Name
clavesin 2
Gene Aliases
C6orf212, C6orf213, RLBP1L2, bA160A10.4
UniGene
Hs.486361
Gene Chromosome Location
Chr.6: 122993693-123081282 on Build GRCh38
Transcripts
NM_001010852.3 XM_017010266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1697866 c.871G>A p.E291K Chr.6: 123056001-123056001 on Build GRCh38
5543802 c.740C>T p.S247F Chr.6: 123055870-123055870 on Build GRCh38
5601257 c.776G>A p.G259E Chr.6: 123055906-123055906 on Build GRCh38
739363 c.677T>G p.I226R Chr.6: 123055807-123055807 on Build GRCh38
1697861 c.724C>T p.P242S Chr.6: 123055854-123055854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77109261 Chr.6: 123055746-123055746 on Build GRCh38
rs765806616 Chr.6: 123055770-123055770 on Build GRCh38
rs1044816761 Chr.6: 123055795-123055795 on Build GRCh38
rs773758742 Chr.6: 123055979-123055979 on Build GRCh38
rs764027486 Chr.6: 123056054-123056054 on Build GRCh38
rs754824939 Chr.6: 123055779-123055779 on Build GRCh38
rs770453643 Chr.6: 123055906-123055906 on Build GRCh38
rs140575101 Chr.6: 123055968-123055968 on Build GRCh38
rs547464007 Chr.6: 123055868-123055868 on Build GRCh38
rs12192029 Chr.6: 123056080-123056080 on Build GRCh38
rs912762384 Chr.6: 123055743-123055743 on Build GRCh38
rs773981838 Chr.6: 123056034-123056034 on Build GRCh38
rs778065897 Chr.6: 123056004-123056004 on Build GRCh38
rs538249510 Chr.6: 123055964-123055964 on Build GRCh38
rs372072412 Chr.6: 123056040-123056040 on Build GRCh38
rs769013044 Chr.6: 123055833-123055833 on Build GRCh38
rs780230722 Chr.6: 123055805-123055805 on Build GRCh38
rs751549576 Chr.6: 123055775-123055775 on Build GRCh38
rs200941440 Chr.6: 123055814-123055814 on Build GRCh38
rs773715447 Chr.6: 123055907-123055907 on Build GRCh38
rs780221084 Chr.6: 123055965-123055965 on Build GRCh38
rs755563448 Chr.6: 123055969-123055969 on Build GRCh38
rs563452621 Chr.6: 123056116-123056116 on Build GRCh38
rs777116578 Chr.6: 123055845-123055845 on Build GRCh38
rs912688809 Chr.6: 123056072-123056072 on Build GRCh38
rs771554229 Chr.6: 123056036-123056036 on Build GRCh38
rs954452562 Chr.6: 123056062-123056062 on Build GRCh38
rs934860525 Chr.6: 123056117-123056117 on Build GRCh38
rs752596301 Chr.6: 123055944-123055944 on Build GRCh38
rs146800301 Chr.6: 123055962-123055962 on Build GRCh38
rs760560905 Chr.6: 123055960-123055960 on Build GRCh38
rs183242803 Chr.6: 123055790-123055790 on Build GRCh38
rs746030546 Chr.6: 123055797-123055797 on Build GRCh38
rs199839432 Chr.6: 123055898-123055898 on Build GRCh38
rs748619435 Chr.6: 123055983-123055983 on Build GRCh38
rs148805106 Chr.6: 123055859-123055859 on Build GRCh38
rs772764145 Chr.6: 123055766-123055766 on Build GRCh38
rs770173535 Chr.6: 123055991-123055991 on Build GRCh38
rs1033944404 Chr.6: 123055722-123055722 on Build GRCh38
rs751775763 Chr.6: 123055851-123055851 on Build GRCh38
rs187275685 Chr.6: 123055798-123055798 on Build GRCh38
rs906171681 Chr.6: 123056083-123056083 on Build GRCh38
rs553690624 Chr.6: 123056010-123056010 on Build GRCh38
rs952604927 Chr.6: 123055737-123055737 on Build GRCh38
rs762460952 Chr.6: 123055767-123055767 on Build GRCh38
rs780541656 Chr.6: 123056062-123056070 on Build GRCh38
rs752407735 Chr.6: 123055788-123055788 on Build GRCh38
rs61732197 Chr.6: 123055961-123055961 on Build GRCh38
rs574635525 Chr.6: 123055974-123055974 on Build GRCh38
rs758956163 Chr.6: 123056035-123056035 on Build GRCh38
rs147426485 Chr.6: 123055943-123055943 on Build GRCh38
rs1028642540 Chr.6: 123056038-123056038 on Build GRCh38
rs760105345 Chr.6: 123056019-123056019 on Build GRCh38
rs540970659 Chr.6: 123055953-123055953 on Build GRCh38
rs9388175 Chr.6: 123056137-123056137 on Build GRCh38
rs759563119 Chr.6: 123055910-123055910 on Build GRCh38
rs368778974 Chr.6: 123055890-123055890 on Build GRCh38
rs768633966 Chr.6: 123055992-123055992 on Build GRCh38
rs200964655 Chr.6: 123055931-123055931 on Build GRCh38
rs753654320 Chr.6: 123056067-123056067 on Build GRCh38
rs112034617 Chr.6: 123055750-123055750 on Build GRCh38
rs936846825 Chr.6: 123055920-123055920 on Build GRCh38
rs376247051 Chr.6: 123055787-123055787 on Build GRCh38

View Full Product Details