Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154334015-154334515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B2
Entrez Gene ID
57198
Gene Name
ATPase phospholipid transporting 8B2
Gene Aliases
ATPID
UniGene
Hs.435700
Gene Chromosome Location
Chr.1: 154325541-154351307 on Build GRCh38
Transcripts
NM_001005855.1 NM_020452.3 XM_005245355.2 XM_005245356.3 XM_017001871.1 XM_017001872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6533892 c.721T>G p.F241V Chr.1: 154334181-154334181 on Build GRCh38
3965851 c.734G>A p.S245N Chr.1: 154334152-154334152 on Build GRCh38
6507810 c.722T>A p.F241Y Chr.1: 154334182-154334182 on Build GRCh38
6507808 c.764T>A p.F255Y Chr.1: 154334182-154334182 on Build GRCh38
2208987 c.709C>T p.R237* Chr.1: 154334226-154334226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142320412 Chr.1: 154334394-154334394 on Build GRCh38
rs757123109 Chr.1: 154334225-154334225 on Build GRCh38
rs752759696 Chr.1: 154334453-154334453 on Build GRCh38
rs751576352 Chr.1: 154334201-154334201 on Build GRCh38
rs751668081 Chr.1: 154334478-154334478 on Build GRCh38
rs142408071 Chr.1: 154334153-154334153 on Build GRCh38
rs773430189 Chr.1: 154334389-154334389 on Build GRCh38
rs766296809 Chr.1: 154334450-154334450 on Build GRCh38
rs201913757 Chr.1: 154334246-154334246 on Build GRCh38
rs777151131 Chr.1: 154334295-154334295 on Build GRCh38
rs763172393 Chr.1: 154334472-154334472 on Build GRCh38
rs765402135 Chr.1: 154334191-154334191 on Build GRCh38
rs777553967 Chr.1: 154334112-154334112 on Build GRCh38
rs745486480 Chr.1: 154334140-154334140 on Build GRCh38
rs748127527 Chr.1: 154334096-154334096 on Build GRCh38
rs1039959000 Chr.1: 154334391-154334391 on Build GRCh38
rs769320146 Chr.1: 154334286-154334286 on Build GRCh38
rs993112211 Chr.1: 154334364-154334364 on Build GRCh38
rs749058130 Chr.1: 154334224-154334224 on Build GRCh38
rs183136294 Chr.1: 154334144-154334144 on Build GRCh38
rs746454699 Chr.1: 154334284-154334284 on Build GRCh38
rs749042644 Chr.1: 154334120-154334120 on Build GRCh38
rs765434624 Chr.1: 154334069-154334069 on Build GRCh38
rs369520582 Chr.1: 154334491-154334491 on Build GRCh38
rs976804834 Chr.1: 154334047-154334047 on Build GRCh38
rs748568798 Chr.1: 154334276-154334276 on Build GRCh38
rs979959173 Chr.1: 154334154-154334154 on Build GRCh38
rs759998962 Chr.1: 154334128-154334128 on Build GRCh38
rs777644148 Chr.1: 154334211-154334211 on Build GRCh38
rs758591309 Chr.1: 154334195-154334195 on Build GRCh38
rs767471283 Chr.1: 154334488-154334488 on Build GRCh38
rs754242543 Chr.1: 154334064-154334064 on Build GRCh38
rs894309923 Chr.1: 154334424-154334424 on Build GRCh38
rs762207935 Chr.1: 154334296-154334296 on Build GRCh38
rs766500819 Chr.1: 154334198-154334198 on Build GRCh38
rs770381848 Chr.1: 154334226-154334226 on Build GRCh38
rs376365867 Chr.1: 154334244-154334244 on Build GRCh38
rs1056935749 Chr.1: 154334381-154334381 on Build GRCh38
rs773447851 Chr.1: 154334467-154334467 on Build GRCh38
rs35339531 Chr.1: 154334162-154334162 on Build GRCh38
rs756029051 Chr.1: 154334110-154334110 on Build GRCh38
rs762200945 Chr.1: 154334167-154334167 on Build GRCh38
rs756047511 Chr.1: 154334206-154334206 on Build GRCh38
rs766517471 Chr.1: 154334474-154334474 on Build GRCh38
rs902851606 Chr.1: 154334342-154334342 on Build GRCh38
rs760650211 Chr.1: 154334356-154334356 on Build GRCh38
rs368979389 Chr.1: 154334270-154334270 on Build GRCh38
rs758457129 Chr.1: 154334075-154334075 on Build GRCh38
rs764620252 Chr.1: 154334056-154334056 on Build GRCh38
rs750711058 Chr.1: 154334192-154334192 on Build GRCh38
rs748834147 Chr.1: 154334414-154334414 on Build GRCh38
rs1056838477 Chr.1: 154334171-154334171 on Build GRCh38
rs774004907 Chr.1: 154334132-154334132 on Build GRCh38
rs1031503722 Chr.1: 154334055-154334055 on Build GRCh38
rs771747082 Chr.1: 154334141-154334141 on Build GRCh38
rs770519651 Chr.1: 154334125-154334125 on Build GRCh38
rs201417005 Chr.1: 154334314-154334314 on Build GRCh38
rs745600555 Chr.1: 154334247-154334247 on Build GRCh38
rs780089601 Chr.1: 154334083-154334083 on Build GRCh38
rs372820914 Chr.1: 154334065-154334065 on Build GRCh38
rs548738394 Chr.1: 154334101-154334101 on Build GRCh38
rs750502356 Chr.1: 154334074-154334074 on Build GRCh38
rs779724744 Chr.1: 154334271-154334271 on Build GRCh38
rs998583080 Chr.1: 154334408-154334408 on Build GRCh38
rs1013187903 Chr.1: 154334429-154334429 on Build GRCh38
rs759562111 Chr.1: 154334484-154334484 on Build GRCh38

View Full Product Details