Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155018285-155018783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
UniGene
Hs.591491
Gene Chromosome Location
Chr.1: 155018513-155033781 on Build GRCh38
Transcripts
NM_144622.2 XM_011509188.2 XM_011509189.2 NM_001252406.2 NM_001256455.1 NR_045515.1 NR_046206.1 NR_049765.1 XM_006711349.2 XM_006711353.1 XM_006711354.1 XM_006711356.3 XM_006711357.1 XM_006711358.1 XM_006711359.2 XM_011509598.2 XM_011509599.2 XM_017001399.1

Gene Symbol
ZBTB7B
Entrez Gene ID
51043
Gene Name
zinc finger and BTB domain containing 7B
Gene Aliases
CKROX, THPOK, ZBTB15, ZFP-67, ZFP67, ZNF857B, c-KROX, hcKROX
UniGene
Hs.729279
Gene Chromosome Location
Chr.1: 155002628-155018525 on Build GRCh38
Transcripts
NM_144622.2 XM_011509188.2 XM_011509189.2 NM_001252406.2 NM_001256455.1 NR_045515.1 NR_046206.1 NR_049765.1 XM_006711349.2 XM_006711353.1 XM_006711354.1 XM_006711356.3 XM_006711357.1 XM_006711358.1 XM_006711359.2 XM_011509598.2 XM_011509599.2 XM_017001399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2209692 c.2300C>A p.P767H Chr.1: 155018566-155018566 on Build GRCh38
896724 c.2119_2121delGAG p.E707delE Chr.1: 155018745-155018747 on Build GRCh38
2209691 c.2307C>T p.H769H Chr.1: 155018559-155018559 on Build GRCh38
2209690 c.2311C>T p.P771S Chr.1: 155018555-155018555 on Build GRCh38
6279779 c.2229C>T p.S743S Chr.1: 155018637-155018637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200361669 Chr.1: 155018642-155018642 on Build GRCh38
rs771043586 Chr.1: 155018497-155018497 on Build GRCh38
rs538649930 Chr.1: 155018652-155018652 on Build GRCh38
rs930492978 Chr.1: 155018402-155018402 on Build GRCh38
rs370200297 Chr.1: 155018720-155018720 on Build GRCh38
rs376614202 Chr.1: 155018732-155018732 on Build GRCh38
rs771008307 Chr.1: 155018592-155018592 on Build GRCh38
rs764552167 Chr.1: 155018739-155018739 on Build GRCh38
rs776607137 Chr.1: 155018526-155018526 on Build GRCh38
rs749574086 Chr.1: 155018663-155018663 on Build GRCh38
rs779534622 Chr.1: 155018489-155018490 on Build GRCh38
rs765139185 Chr.1: 155018603-155018603 on Build GRCh38
rs750995971 Chr.1: 155018555-155018555 on Build GRCh38
rs953898991 Chr.1: 155018444-155018444 on Build GRCh38
rs756462435 Chr.1: 155018580-155018580 on Build GRCh38
rs761199509 Chr.1: 155018736-155018736 on Build GRCh38
rs901856615 Chr.1: 155018460-155018460 on Build GRCh38
rs769816556 Chr.1: 155018705-155018705 on Build GRCh38
rs765049478 Chr.1: 155018530-155018530 on Build GRCh38
rs774519160 Chr.1: 155018593-155018593 on Build GRCh38
rs548943345 Chr.1: 155018481-155018481 on Build GRCh38
rs754276669 Chr.1: 155018741-155018741 on Build GRCh38
rs1003889384 Chr.1: 155018438-155018438 on Build GRCh38
rs745904765 Chr.1: 155018594-155018594 on Build GRCh38
rs768762307 Chr.1: 155018595-155018595 on Build GRCh38
rs201399069 Chr.1: 155018590-155018590 on Build GRCh38
rs990745792 Chr.1: 155018358-155018358 on Build GRCh38
rs1048869272 Chr.1: 155018416-155018416 on Build GRCh38
rs773838723 Chr.1: 155018512-155018513 on Build GRCh38
rs781651330 Chr.1: 155018556-155018556 on Build GRCh38
rs762699906 Chr.1: 155018631-155018631 on Build GRCh38
rs774601504 Chr.1: 155018745-155018747 on Build GRCh38
rs766710546 Chr.1: 155018677-155018678 on Build GRCh38
rs1008062314 Chr.1: 155018464-155018464 on Build GRCh38
rs561115317 Chr.1: 155018482-155018482 on Build GRCh38
rs907985168 Chr.1: 155018439-155018439 on Build GRCh38
rs948868168 Chr.1: 155018585-155018585 on Build GRCh38
rs376562403 Chr.1: 155018679-155018679 on Build GRCh38
rs760682601 Chr.1: 155018503-155018503 on Build GRCh38
rs756552401 Chr.1: 155018640-155018640 on Build GRCh38
rs544976317 Chr.1: 155018443-155018443 on Build GRCh38
rs750107120 Chr.1: 155018539-155018539 on Build GRCh38
rs773102244 Chr.1: 155018712-155018712 on Build GRCh38
rs1046031462 Chr.1: 155018318-155018318 on Build GRCh38
rs373443674 Chr.1: 155018725-155018725 on Build GRCh38
rs765894846 Chr.1: 155018500-155018500 on Build GRCh38
rs376521847 Chr.1: 155018462-155018462 on Build GRCh38
rs200943269 Chr.1: 155018750-155018750 on Build GRCh38
rs770679392 Chr.1: 155018721-155018721 on Build GRCh38
rs372351512 Chr.1: 155018578-155018578 on Build GRCh38
rs898169705 Chr.1: 155018710-155018710 on Build GRCh38
rs761687579 Chr.1: 155018529-155018529 on Build GRCh38
rs188509639 Chr.1: 155018474-155018474 on Build GRCh38
rs778131489 Chr.1: 155018660-155018660 on Build GRCh38
rs773866616 Chr.1: 155018724-155018724 on Build GRCh38
rs778569738 Chr.1: 155018636-155018636 on Build GRCh38
rs575823382 Chr.1: 155018366-155018366 on Build GRCh38
rs761777453 Chr.1: 155018600-155018600 on Build GRCh38
rs927612408 Chr.1: 155018344-155018344 on Build GRCh38
rs763376207 Chr.1: 155018618-155018619 on Build GRCh38
rs865802476 Chr.1: 155018743-155018743 on Build GRCh38
rs192742085 Chr.1: 155018551-155018551 on Build GRCh38
rs746027446 Chr.1: 155018689-155018689 on Build GRCh38
rs199916941 Chr.1: 155018669-155018669 on Build GRCh38
rs906827955 Chr.1: 155018415-155018415 on Build GRCh38
rs768075327 Chr.1: 155018639-155018639 on Build GRCh38
rs772282695 Chr.1: 155018691-155018691 on Build GRCh38
rs139608616 Chr.1: 155018502-155018504 on Build GRCh38
rs71586045 Chr.1: 155018431-155018432 on Build GRCh38
rs893368681 Chr.1: 155018502-155018502 on Build GRCh38
rs748120484 Chr.1: 155018702-155018702 on Build GRCh38
rs776706699 Chr.1: 155018598-155018598 on Build GRCh38
rs373089065 Chr.1: 155018752-155018752 on Build GRCh38
rs916475209 Chr.1: 155018382-155018382 on Build GRCh38
rs757932434 Chr.1: 155018546-155018546 on Build GRCh38
rs960323300 Chr.1: 155018354-155018354 on Build GRCh38
rs763897194 Chr.1: 155018509-155018509 on Build GRCh38
rs532972227 Chr.1: 155018442-155018442 on Build GRCh38
rs772836589 Chr.1: 155018610-155018610 on Build GRCh38
rs371755027 Chr.1: 155018637-155018637 on Build GRCh38
rs376878658 Chr.1: 155018588-155018588 on Build GRCh38
rs1040395421 Chr.1: 155018454-155018454 on Build GRCh38
rs775655201 Chr.1: 155018694-155018694 on Build GRCh38

View Full Product Details