Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155029947-155030451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
UniGene
Hs.591491
Gene Chromosome Location
Chr.1: 155018513-155033781 on Build GRCh38
Transcripts
NM_144622.2 XM_011509188.2 XM_011509189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411981 c.1160G>T p.R387M Chr.1: 155030101-155030101 on Build GRCh38
6385794 c.1094G>A p.R365H Chr.1: 155030167-155030167 on Build GRCh38
3862894 c.1153G>A p.E385K Chr.1: 155030108-155030108 on Build GRCh38
3474813 c.1098G>A p.M366I Chr.1: 155030163-155030163 on Build GRCh38
6058350 c.1174C>T p.P392S Chr.1: 155030087-155030087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116757455 Chr.1: 155030029-155030029 on Build GRCh38
rs1013793499 Chr.1: 155029981-155029981 on Build GRCh38
rs1045625259 Chr.1: 155030020-155030020 on Build GRCh38
rs199541110 Chr.1: 155030404-155030404 on Build GRCh38
rs548914229 Chr.1: 155030332-155030332 on Build GRCh38
rs780136070 Chr.1: 155030117-155030117 on Build GRCh38
rs770447615 Chr.1: 155030191-155030191 on Build GRCh38
rs201083456 Chr.1: 155030152-155030152 on Build GRCh38
rs745374187 Chr.1: 155030411-155030411 on Build GRCh38
rs967730219 Chr.1: 155030249-155030249 on Build GRCh38
rs772525716 Chr.1: 155030412-155030412 on Build GRCh38
rs371970604 Chr.1: 155030099-155030099 on Build GRCh38
rs113708914 Chr.1: 155030260-155030260 on Build GRCh38
rs776331182 Chr.1: 155030129-155030130 on Build GRCh38
rs768601965 Chr.1: 155030405-155030405 on Build GRCh38
rs375468532 Chr.1: 155030228-155030228 on Build GRCh38
rs200847364 Chr.1: 155030167-155030167 on Build GRCh38
rs527291216 Chr.1: 155030385-155030385 on Build GRCh38
rs775479125 Chr.1: 155030053-155030053 on Build GRCh38
rs764744204 Chr.1: 155030426-155030427 on Build GRCh38
rs375134478 Chr.1: 155030419-155030419 on Build GRCh38
rs755457920 Chr.1: 155030023-155030023 on Build GRCh38
rs568598201 Chr.1: 155030048-155030048 on Build GRCh38
rs958064640 Chr.1: 155030402-155030402 on Build GRCh38
rs370660873 Chr.1: 155030418-155030418 on Build GRCh38
rs978091323 Chr.1: 155030269-155030269 on Build GRCh38
rs761097567 Chr.1: 155030235-155030235 on Build GRCh38
rs1046810397 Chr.1: 155030317-155030317 on Build GRCh38
rs759942497 Chr.1: 155030066-155030067 on Build GRCh38
rs35788499 Chr.1: 155030272-155030272 on Build GRCh38
rs769466175 Chr.1: 155030126-155030126 on Build GRCh38
rs775781123 Chr.1: 155030227-155030227 on Build GRCh38
rs921779260 Chr.1: 155030077-155030077 on Build GRCh38
rs886893400 Chr.1: 155030322-155030322 on Build GRCh38
rs925295597 Chr.1: 155030273-155030273 on Build GRCh38
rs919610753 Chr.1: 155030311-155030311 on Build GRCh38
rs1028797134 Chr.1: 155030223-155030223 on Build GRCh38
rs945917329 Chr.1: 155029974-155029974 on Build GRCh38
rs755338188 Chr.1: 155030095-155030095 on Build GRCh38
rs188808511 Chr.1: 155030153-155030153 on Build GRCh38
rs375685016 Chr.1: 155030030-155030030 on Build GRCh38
rs772766407 Chr.1: 155030129-155030129 on Build GRCh38
rs746879456 Chr.1: 155030120-155030120 on Build GRCh38
rs758888288 Chr.1: 155030051-155030051 on Build GRCh38
rs369912447 Chr.1: 155030394-155030394 on Build GRCh38
rs746973517 Chr.1: 155030160-155030160 on Build GRCh38
rs747961590 Chr.1: 155030168-155030168 on Build GRCh38
rs754927933 Chr.1: 155030390-155030390 on Build GRCh38
rs749207498 Chr.1: 155030376-155030376 on Build GRCh38
rs768049974 Chr.1: 155030224-155030224 on Build GRCh38
rs916700806 Chr.1: 155030351-155030351 on Build GRCh38
rs781386052 Chr.1: 155030113-155030113 on Build GRCh38
rs764683716 Chr.1: 155030085-155030085 on Build GRCh38
rs138215888 Chr.1: 155030310-155030310 on Build GRCh38
rs559776184 Chr.1: 155030377-155030377 on Build GRCh38
rs528875474 Chr.1: 155030308-155030308 on Build GRCh38
rs748879555 Chr.1: 155030187-155030187 on Build GRCh38
rs769774494 Chr.1: 155030040-155030040 on Build GRCh38
rs768282062 Chr.1: 155030128-155030129 on Build GRCh38
rs373266671 Chr.1: 155030395-155030395 on Build GRCh38
rs754128062 Chr.1: 155030275-155030275 on Build GRCh38
rs765445449 Chr.1: 155030388-155030388 on Build GRCh38
rs201803286 Chr.1: 155030109-155030109 on Build GRCh38
rs751983174 Chr.1: 155030090-155030090 on Build GRCh38
rs1008385617 Chr.1: 155030331-155030331 on Build GRCh38
rs147359914 Chr.1: 155030177-155030177 on Build GRCh38
rs200223587 Chr.1: 155030098-155030098 on Build GRCh38
rs749360215 Chr.1: 155030108-155030108 on Build GRCh38
rs768350916 Chr.1: 155030121-155030121 on Build GRCh38
rs535978424 Chr.1: 155030068-155030068 on Build GRCh38
rs561922204 Chr.1: 155030204-155030204 on Build GRCh38
rs749917529 Chr.1: 155030086-155030086 on Build GRCh38
rs150211037 Chr.1: 155030145-155030145 on Build GRCh38
rs138904334 Chr.1: 155030146-155030146 on Build GRCh38
rs887285247 Chr.1: 155030083-155030083 on Build GRCh38
rs746460381 Chr.1: 155030116-155030116 on Build GRCh38
rs368042911 Chr.1: 155030087-155030087 on Build GRCh38
rs770541743 Chr.1: 155030406-155030406 on Build GRCh38
rs767806728 Chr.1: 155030149-155030149 on Build GRCh38
rs753076814 Chr.1: 155030150-155030150 on Build GRCh38
rs777863655 Chr.1: 155030045-155030045 on Build GRCh38
rs937279551 Chr.1: 155030214-155030214 on Build GRCh38
rs777759074 Chr.1: 155030106-155030106 on Build GRCh38
rs778446334 Chr.1: 155030409-155030409 on Build GRCh38
rs936996662 Chr.1: 155030024-155030024 on Build GRCh38
rs201752543 Chr.1: 155030122-155030122 on Build GRCh38
rs761456472 Chr.1: 155030390-155030390 on Build GRCh38
rs760533951 Chr.1: 155030056-155030056 on Build GRCh38
rs554275177 Chr.1: 155030134-155030134 on Build GRCh38
rs769395455 Chr.1: 155030169-155030169 on Build GRCh38
rs936734334 Chr.1: 155030291-155030291 on Build GRCh38
rs769167047 Chr.1: 155030425-155030425 on Build GRCh38
rs756322401 Chr.1: 155030104-155030104 on Build GRCh38
rs931018846 Chr.1: 155030312-155030312 on Build GRCh38
rs775246402 Chr.1: 155030309-155030309 on Build GRCh38
rs898218656 Chr.1: 155030033-155030033 on Build GRCh38
rs779079585 Chr.1: 155030159-155030159 on Build GRCh38
rs142958975 Chr.1: 155030340-155030340 on Build GRCh38
rs762442640 Chr.1: 155030133-155030133 on Build GRCh38
rs138589439 Chr.1: 155030158-155030158 on Build GRCh38
rs760005021 Chr.1: 155030217-155030217 on Build GRCh38
rs761477035 Chr.1: 155030075-155030075 on Build GRCh38

View Full Product Details