Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155034559-155035105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCST1
Entrez Gene ID
149095
Gene Name
DC-STAMP domain containing 1
Gene Aliases
-
UniGene
Hs.567717
Gene Chromosome Location
Chr.1: 155033806-155050930 on Build GRCh38
Transcripts
NM_001143687.2 NM_152494.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6578353 c.211C>T p.P71S Chr.1: 155034676-155034676 on Build GRCh38
3862897 c.241A>G p.M81V Chr.1: 155034706-155034706 on Build GRCh38
5102926 c.225C>T p.Y75Y Chr.1: 155034690-155034690 on Build GRCh38
675811 c.231A>G p.E77E Chr.1: 155034696-155034696 on Build GRCh38
6578352 c.211C>T p.P71S Chr.1: 155034676-155034676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755485103 Chr.1: 155034727-155034727 on Build GRCh38
rs374793317 Chr.1: 155034587-155034587 on Build GRCh38
rs770940125 Chr.1: 155034690-155034690 on Build GRCh38
rs752052385 Chr.1: 155034666-155034666 on Build GRCh38
rs781585881 Chr.1: 155034675-155034675 on Build GRCh38
rs147557352 Chr.1: 155035051-155035052 on Build GRCh38
rs186517730 Chr.1: 155034689-155034689 on Build GRCh38
rs956200868 Chr.1: 155034867-155034867 on Build GRCh38
rs759229908 Chr.1: 155034627-155034627 on Build GRCh38
rs900217176 Chr.1: 155034823-155034823 on Build GRCh38
rs142128744 Chr.1: 155034778-155034778 on Build GRCh38
rs918709548 Chr.1: 155034971-155034971 on Build GRCh38
rs900164666 Chr.1: 155035035-155035035 on Build GRCh38
rs373266993 Chr.1: 155034752-155034752 on Build GRCh38
rs151175890 Chr.1: 155034950-155034950 on Build GRCh38
rs80017536 Chr.1: 155035031-155035031 on Build GRCh38
rs777895634 Chr.1: 155034687-155034687 on Build GRCh38
rs201911650 Chr.1: 155034670-155034670 on Build GRCh38
rs747009978 Chr.1: 155034775-155034775 on Build GRCh38
rs1005643687 Chr.1: 155034803-155034803 on Build GRCh38
rs772354702 Chr.1: 155034605-155034605 on Build GRCh38
rs911769199 Chr.1: 155034997-155034997 on Build GRCh38
rs528090904 Chr.1: 155034943-155034943 on Build GRCh38
rs4845405 Chr.1: 155034826-155034826 on Build GRCh38
rs201300636 Chr.1: 155034718-155034718 on Build GRCh38
rs899997147 Chr.1: 155034806-155034806 on Build GRCh38
rs377329888 Chr.1: 155034917-155034917 on Build GRCh38
rs142074614 Chr.1: 155035059-155035060 on Build GRCh38
rs775843572 Chr.1: 155034698-155034700 on Build GRCh38
rs776466703 Chr.1: 155034704-155034704 on Build GRCh38
rs749130221 Chr.1: 155034759-155034759 on Build GRCh38
rs371187838 Chr.1: 155034761-155034761 on Build GRCh38
rs77429755 Chr.1: 155034779-155034779 on Build GRCh38
rs780714741 Chr.1: 155034664-155034664 on Build GRCh38
rs963077080 Chr.1: 155034954-155034954 on Build GRCh38
rs560789988 Chr.1: 155035045-155035045 on Build GRCh38
rs397831109 Chr.1: 155035076-155035077 on Build GRCh38
rs955825434 Chr.1: 155034836-155034836 on Build GRCh38
rs369174751 Chr.1: 155034642-155034642 on Build GRCh38
rs376021147 Chr.1: 155034733-155034733 on Build GRCh38
rs764979994 Chr.1: 155034706-155034706 on Build GRCh38
rs1054122376 Chr.1: 155035056-155035056 on Build GRCh38
rs752043744 Chr.1: 155034723-155034723 on Build GRCh38
rs199824527 Chr.1: 155034651-155034651 on Build GRCh38
rs999853431 Chr.1: 155035055-155035055 on Build GRCh38
rs769445537 Chr.1: 155035052-155035055 on Build GRCh38
rs189404322 Chr.1: 155034805-155034805 on Build GRCh38
rs775428856 Chr.1: 155034691-155034691 on Build GRCh38
rs964513108 Chr.1: 155034655-155034655 on Build GRCh38
rs980479633 Chr.1: 155034944-155034944 on Build GRCh38
rs200759818 Chr.1: 155034826-155034829 on Build GRCh38
rs887169521 Chr.1: 155034797-155034797 on Build GRCh38
rs772937499 Chr.1: 155034711-155034711 on Build GRCh38
rs761403439 Chr.1: 155034749-155034749 on Build GRCh38
rs770041987 Chr.1: 155034683-155034683 on Build GRCh38
rs560535932 Chr.1: 155034615-155034615 on Build GRCh38
rs749943505 Chr.1: 155034662-155034662 on Build GRCh38
rs1008956183 Chr.1: 155034809-155034809 on Build GRCh38
rs759044612 Chr.1: 155034663-155034663 on Build GRCh38
rs4845404 Chr.1: 155034817-155034817 on Build GRCh38
rs371258809 Chr.1: 155035017-155035017 on Build GRCh38
rs200630580 Chr.1: 155034762-155034762 on Build GRCh38
rs982743389 Chr.1: 155035026-155035026 on Build GRCh38
rs780216706 Chr.1: 155034923-155034923 on Build GRCh38
rs749254026 Chr.1: 155034958-155034958 on Build GRCh38
rs753050338 Chr.1: 155034608-155034608 on Build GRCh38
rs944650010 Chr.1: 155035002-155035002 on Build GRCh38
rs61730947 Chr.1: 155034705-155034705 on Build GRCh38
rs763901932 Chr.1: 155034616-155034616 on Build GRCh38
rs1033148802 Chr.1: 155034680-155034680 on Build GRCh38
rs577260657 Chr.1: 155034821-155034821 on Build GRCh38
rs759525566 Chr.1: 155034584-155034584 on Build GRCh38
rs139857431 Chr.1: 155034753-155034753 on Build GRCh38
rs929234288 Chr.1: 155035025-155035025 on Build GRCh38
rs752400113 Chr.1: 155034739-155034739 on Build GRCh38
rs181727297 Chr.1: 155034866-155034866 on Build GRCh38
rs1047614040 Chr.1: 155034980-155034980 on Build GRCh38
rs1024496626 Chr.1: 155034881-155034881 on Build GRCh38
rs200613020 Chr.1: 155034583-155034583 on Build GRCh38
rs1037502777 Chr.1: 155034740-155034740 on Build GRCh38
rs376209113 Chr.1: 155034732-155034732 on Build GRCh38
rs974151287 Chr.1: 155034957-155034957 on Build GRCh38
rs765943561 Chr.1: 155034721-155034721 on Build GRCh38
rs891572923 Chr.1: 155035066-155035066 on Build GRCh38
rs34390501 Chr.1: 155035052-155035053 on Build GRCh38
rs768377572 Chr.1: 155034703-155034703 on Build GRCh38
rs929110986 Chr.1: 155034974-155034974 on Build GRCh38
rs750759329 Chr.1: 155035055-155035056 on Build GRCh38
rs979142932 Chr.1: 155034913-155034913 on Build GRCh38
rs751598061 Chr.1: 155034847-155034847 on Build GRCh38
rs560656206 Chr.1: 155034877-155034877 on Build GRCh38
rs778939736 Chr.1: 155034763-155034763 on Build GRCh38
rs913623598 Chr.1: 155034607-155034607 on Build GRCh38
rs746925384 Chr.1: 155034695-155034695 on Build GRCh38

View Full Product Details