Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155133414-155133916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA1
Entrez Gene ID
1942
Gene Name
ephrin A1
Gene Aliases
B61, ECKLG, EFL1, EPLG1, LERK-1, LERK1, TNFAIP4
UniGene
Hs.516664
Gene Chromosome Location
Chr.1: 155127873-155134910 on Build GRCh38
Transcripts
NM_004428.2 NM_182685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996964 c.454+10A>G p.? Chr.1: 155133578-155133578 on Build GRCh38
2209916 c.425G>A p.R142K Chr.1: 155133539-155133539 on Build GRCh38
7151987 c.492G>A p.K164K Chr.1: 155133767-155133767 on Build GRCh38
6652056 c.451A>T p.I151F Chr.1: 155133565-155133565 on Build GRCh38
3750404 c.476A>T p.D159V Chr.1: 155133751-155133751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920217009 Chr.1: 155133689-155133689 on Build GRCh38
rs768187342 Chr.1: 155133819-155133821 on Build GRCh38
rs757949624 Chr.1: 155133458-155133458 on Build GRCh38
rs777016129 Chr.1: 155133698-155133698 on Build GRCh38
rs11545256 Chr.1: 155133548-155133548 on Build GRCh38
rs780477634 Chr.1: 155133484-155133485 on Build GRCh38
rs533781385 Chr.1: 155133620-155133620 on Build GRCh38
rs751092470 Chr.1: 155133551-155133551 on Build GRCh38
rs1039585370 Chr.1: 155133843-155133843 on Build GRCh38
rs758620834 Chr.1: 155133470-155133472 on Build GRCh38
rs762230947 Chr.1: 155133719-155133719 on Build GRCh38
rs775507556 Chr.1: 155133524-155133524 on Build GRCh38
rs200568335 Chr.1: 155133495-155133495 on Build GRCh38
rs4390169 Chr.1: 155133578-155133578 on Build GRCh38
rs973321901 Chr.1: 155133683-155133683 on Build GRCh38
rs1024845969 Chr.1: 155133605-155133605 on Build GRCh38
rs773180386 Chr.1: 155133505-155133505 on Build GRCh38
rs4745 Chr.1: 155133751-155133751 on Build GRCh38
rs754675430 Chr.1: 155133468-155133468 on Build GRCh38
rs1020851073 Chr.1: 155133450-155133450 on Build GRCh38
rs748652115 Chr.1: 155133760-155133762 on Build GRCh38
rs996991159 Chr.1: 155133675-155133676 on Build GRCh38
rs373741097 Chr.1: 155133704-155133704 on Build GRCh38
rs1033536381 Chr.1: 155133643-155133643 on Build GRCh38
rs745882383 Chr.1: 155133521-155133521 on Build GRCh38
rs369380852 Chr.1: 155133837-155133837 on Build GRCh38
rs751052776 Chr.1: 155133826-155133826 on Build GRCh38
rs558318083 Chr.1: 155133660-155133660 on Build GRCh38
rs750972580 Chr.1: 155133541-155133541 on Build GRCh38
rs201040880 Chr.1: 155133733-155133733 on Build GRCh38
rs766866514 Chr.1: 155133791-155133791 on Build GRCh38
rs755922880 Chr.1: 155133609-155133609 on Build GRCh38
rs760965678 Chr.1: 155133530-155133530 on Build GRCh38
rs765930865 Chr.1: 155133546-155133546 on Build GRCh38
rs967337849 Chr.1: 155133466-155133466 on Build GRCh38
rs780167990 Chr.1: 155133686-155133686 on Build GRCh38
rs770247628 Chr.1: 155133772-155133772 on Build GRCh38
rs752463962 Chr.1: 155133583-155133583 on Build GRCh38
rs941608799 Chr.1: 155133639-155133639 on Build GRCh38
rs1020756271 Chr.1: 155133848-155133848 on Build GRCh38
rs978703155 Chr.1: 155133470-155133470 on Build GRCh38
rs367947497 Chr.1: 155133460-155133460 on Build GRCh38
rs922133562 Chr.1: 155133847-155133847 on Build GRCh38
rs773607656 Chr.1: 155133773-155133781 on Build GRCh38
rs911656021 Chr.1: 155133803-155133803 on Build GRCh38
rs113727487 Chr.1: 155133715-155133715 on Build GRCh38
rs576921501 Chr.1: 155133676-155133676 on Build GRCh38
rs768786736 Chr.1: 155133688-155133689 on Build GRCh38
rs769691599 Chr.1: 155133504-155133504 on Build GRCh38
rs6699434 Chr.1: 155133540-155133540 on Build GRCh38
rs781446328 Chr.1: 155133707-155133709 on Build GRCh38
rs777130223 Chr.1: 155133535-155133535 on Build GRCh38
rs559002400 Chr.1: 155133577-155133577 on Build GRCh38
rs772220878 Chr.1: 155133523-155133523 on Build GRCh38
rs1050901869 Chr.1: 155133695-155133695 on Build GRCh38
rs983189814 Chr.1: 155133794-155133794 on Build GRCh38
rs777643748 Chr.1: 155133613-155133613 on Build GRCh38
rs866318420 Chr.1: 155133750-155133750 on Build GRCh38
rs41264899 Chr.1: 155133619-155133619 on Build GRCh38
rs763506729 Chr.1: 155133788-155133788 on Build GRCh38
rs768966808 Chr.1: 155133697-155133697 on Build GRCh38
rs936218194 Chr.1: 155133857-155133857 on Build GRCh38
rs755349435 Chr.1: 155133732-155133732 on Build GRCh38
rs143462709 Chr.1: 155133539-155133539 on Build GRCh38
rs773535406 Chr.1: 155133766-155133766 on Build GRCh38
rs1054674144 Chr.1: 155133874-155133874 on Build GRCh38
rs982376837 Chr.1: 155133626-155133626 on Build GRCh38
rs991598289 Chr.1: 155133554-155133554 on Build GRCh38
rs748005147 Chr.1: 155133499-155133499 on Build GRCh38
rs375381523 Chr.1: 155133756-155133756 on Build GRCh38
rs925972885 Chr.1: 155133494-155133494 on Build GRCh38
rs747078020 Chr.1: 155133573-155133574 on Build GRCh38

View Full Product Details