Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155133640-155134147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA1
Entrez Gene ID
1942
Gene Name
ephrin A1
Gene Aliases
B61, ECKLG, EFL1, EPLG1, LERK-1, LERK1, TNFAIP4
UniGene
Hs.516664
Gene Chromosome Location
Chr.1: 155127873-155134910 on Build GRCh38
Transcripts
NM_004428.2 NM_182685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6445313 c.539G>C p.G180A Chr.1: 155133988-155133988 on Build GRCh38
3385227 c.538G>A p.G180S Chr.1: 155133987-155133987 on Build GRCh38
3750404 c.476A>T p.D159V Chr.1: 155133751-155133751 on Build GRCh38
675804 c.538G>T p.G180C Chr.1: 155133987-155133987 on Build GRCh38
7151987 c.492G>A p.K164K Chr.1: 155133767-155133767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920217009 Chr.1: 155133689-155133689 on Build GRCh38
rs748396189 Chr.1: 155133968-155133968 on Build GRCh38
rs759098424 Chr.1: 155133906-155133906 on Build GRCh38
rs768187342 Chr.1: 155133819-155133821 on Build GRCh38
rs746384002 Chr.1: 155134063-155134063 on Build GRCh38
rs967914151 Chr.1: 155133903-155133903 on Build GRCh38
rs748308594 Chr.1: 155134027-155134027 on Build GRCh38
rs374822353 Chr.1: 155134087-155134087 on Build GRCh38
rs891790671 Chr.1: 155133908-155133908 on Build GRCh38
rs777016129 Chr.1: 155133698-155133698 on Build GRCh38
rs778755153 Chr.1: 155133945-155133945 on Build GRCh38
rs1039585370 Chr.1: 155133843-155133843 on Build GRCh38
rs755051226 Chr.1: 155134018-155134018 on Build GRCh38
rs771228758 Chr.1: 155134050-155134050 on Build GRCh38
rs760306344 Chr.1: 155134005-155134005 on Build GRCh38
rs762230947 Chr.1: 155133719-155133719 on Build GRCh38
rs114708813 Chr.1: 155133963-155133963 on Build GRCh38
rs757050149 Chr.1: 155134007-155134007 on Build GRCh38
rs973321901 Chr.1: 155133683-155133683 on Build GRCh38
rs756377342 Chr.1: 155134034-155134034 on Build GRCh38
rs917364523 Chr.1: 155133979-155133979 on Build GRCh38
rs4745 Chr.1: 155133751-155133751 on Build GRCh38
rs146781581 Chr.1: 155134061-155134061 on Build GRCh38
rs774976417 Chr.1: 155134072-155134072 on Build GRCh38
rs748652115 Chr.1: 155133760-155133762 on Build GRCh38
rs996991159 Chr.1: 155133675-155133676 on Build GRCh38
rs373741097 Chr.1: 155133704-155133704 on Build GRCh38
rs775058359 Chr.1: 155134109-155134114 on Build GRCh38
rs751052776 Chr.1: 155133826-155133826 on Build GRCh38
rs369380852 Chr.1: 155133837-155133837 on Build GRCh38
rs750303308 Chr.1: 155134106-155134106 on Build GRCh38
rs763269161 Chr.1: 155133971-155133971 on Build GRCh38
rs752373896 Chr.1: 155133914-155133914 on Build GRCh38
rs201040880 Chr.1: 155133733-155133733 on Build GRCh38
rs564674783 Chr.1: 155134075-155134075 on Build GRCh38
rs766866514 Chr.1: 155133791-155133791 on Build GRCh38
rs372418829 Chr.1: 155133941-155133941 on Build GRCh38
rs11545255 Chr.1: 155133994-155133994 on Build GRCh38
rs1032932621 Chr.1: 155134093-155134109 on Build GRCh38
rs780167990 Chr.1: 155133686-155133686 on Build GRCh38
rs770247628 Chr.1: 155133772-155133772 on Build GRCh38
rs11545254 Chr.1: 155133986-155133986 on Build GRCh38
rs1020756271 Chr.1: 155133848-155133848 on Build GRCh38
rs17855563 Chr.1: 155133988-155133988 on Build GRCh38
rs771708713 Chr.1: 155133937-155133938 on Build GRCh38
rs922133562 Chr.1: 155133847-155133847 on Build GRCh38
rs773607656 Chr.1: 155133773-155133781 on Build GRCh38
rs911656021 Chr.1: 155133803-155133803 on Build GRCh38
rs777908694 Chr.1: 155134044-155134044 on Build GRCh38
rs377221471 Chr.1: 155133947-155133947 on Build GRCh38
rs770201963 Chr.1: 155133969-155133969 on Build GRCh38
rs765183519 Chr.1: 155134011-155134011 on Build GRCh38
rs113727487 Chr.1: 155133715-155133715 on Build GRCh38
rs767104304 Chr.1: 155133913-155133913 on Build GRCh38
rs760336648 Chr.1: 155134111-155134112 on Build GRCh38
rs1027606030 Chr.1: 155134054-155134054 on Build GRCh38
rs576921501 Chr.1: 155133676-155133676 on Build GRCh38
rs768786736 Chr.1: 155133688-155133689 on Build GRCh38
rs777557894 Chr.1: 155133926-155133926 on Build GRCh38
rs201402574 Chr.1: 155134006-155134006 on Build GRCh38
rs781446328 Chr.1: 155133707-155133709 on Build GRCh38
rs767014352 Chr.1: 155133987-155133987 on Build GRCh38
rs9297 Chr.1: 155134074-155134074 on Build GRCh38
rs959193434 Chr.1: 155134030-155134030 on Build GRCh38
rs201450895 Chr.1: 155134084-155134084 on Build GRCh38
rs139969988 Chr.1: 155133970-155133970 on Build GRCh38
rs1050901869 Chr.1: 155133695-155133695 on Build GRCh38
rs983189814 Chr.1: 155133794-155133794 on Build GRCh38
rs574811781 Chr.1: 155133920-155133920 on Build GRCh38
rs866318420 Chr.1: 155133750-155133750 on Build GRCh38
rs763506729 Chr.1: 155133788-155133788 on Build GRCh38
rs768966808 Chr.1: 155133697-155133697 on Build GRCh38
rs774740181 Chr.1: 155133977-155133977 on Build GRCh38
rs753634426 Chr.1: 155133928-155133928 on Build GRCh38
rs750393153 Chr.1: 155134017-155134017 on Build GRCh38
rs771450050 Chr.1: 155133973-155133973 on Build GRCh38
rs936218194 Chr.1: 155133857-155133857 on Build GRCh38
rs768890807 Chr.1: 155133949-155133949 on Build GRCh38
rs755349435 Chr.1: 155133732-155133732 on Build GRCh38
rs772625154 Chr.1: 155134064-155134064 on Build GRCh38
rs773535406 Chr.1: 155133766-155133766 on Build GRCh38
rs1054674144 Chr.1: 155133874-155133874 on Build GRCh38
rs375381523 Chr.1: 155133756-155133756 on Build GRCh38
rs781409409 Chr.1: 155134020-155134020 on Build GRCh38
rs775062336 Chr.1: 155133992-155133992 on Build GRCh38

View Full Product Details