Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155236843-155237382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1334592 c.971G>A p.R324H Chr.1: 155237369-155237369 on Build GRCh38
79892 c.969A>G p.Q323Q Chr.1: 155237371-155237371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1002676919 Chr.1: 155237254-155237254 on Build GRCh38
rs905269905 Chr.1: 155237024-155237024 on Build GRCh38
rs756025144 Chr.1: 155237038-155237046 on Build GRCh38
rs572653386 Chr.1: 155236882-155236882 on Build GRCh38
rs923055532 Chr.1: 155237142-155237142 on Build GRCh38
rs531912953 Chr.1: 155236933-155236933 on Build GRCh38
rs1002406253 Chr.1: 155237025-155237025 on Build GRCh38
rs762819647 Chr.1: 155237307-155237307 on Build GRCh38
rs1026590373 Chr.1: 155237335-155237335 on Build GRCh38
rs547873878 Chr.1: 155237162-155237162 on Build GRCh38
rs946743963 Chr.1: 155237222-155237222 on Build GRCh38
rs984436889 Chr.1: 155237078-155237078 on Build GRCh38
rs975957205 Chr.1: 155237139-155237139 on Build GRCh38
rs943208356 Chr.1: 155236903-155236903 on Build GRCh38
rs1035570488 Chr.1: 155237029-155237029 on Build GRCh38
rs189774125 Chr.1: 155237271-155237271 on Build GRCh38
rs575660064 Chr.1: 155237058-155237058 on Build GRCh38
rs556277010 Chr.1: 155237265-155237265 on Build GRCh38
rs1043831718 Chr.1: 155237015-155237015 on Build GRCh38
rs994116445 Chr.1: 155237060-155237060 on Build GRCh38
rs760636654 Chr.1: 155237327-155237327 on Build GRCh38
rs1040599364 Chr.1: 155236905-155236905 on Build GRCh38
rs934601293 Chr.1: 155237152-155237152 on Build GRCh38
rs984573729 Chr.1: 155236883-155236883 on Build GRCh38
rs892429962 Chr.1: 155236945-155236945 on Build GRCh38
rs562556726 Chr.1: 155236957-155236957 on Build GRCh38
rs121908298 Chr.1: 155237357-155237357 on Build GRCh38
rs186893248 Chr.1: 155237177-155237177 on Build GRCh38
rs72704130 Chr.1: 155237239-155237239 on Build GRCh38
rs550723806 Chr.1: 155236942-155236942 on Build GRCh38
rs964786476 Chr.1: 155237134-155237134 on Build GRCh38
rs749925127 Chr.1: 155237259-155237259 on Build GRCh38
rs544673228 Chr.1: 155237185-155237185 on Build GRCh38
rs537631939 Chr.1: 155237256-155237256 on Build GRCh38
rs529756206 Chr.1: 155237073-155237073 on Build GRCh38
rs923224925 Chr.1: 155236907-155236907 on Build GRCh38
rs1043696149 Chr.1: 155237234-155237234 on Build GRCh38
rs12743554 Chr.1: 155236922-155236922 on Build GRCh38
rs398123535 Chr.1: 155237352-155237352 on Build GRCh38
rs546436642 Chr.1: 155236895-155236895 on Build GRCh38
rs766249489 Chr.1: 155237321-155237321 on Build GRCh38
rs775596648 Chr.1: 155237322-155237322 on Build GRCh38
rs570337924 Chr.1: 155237249-155237249 on Build GRCh38
rs142662866 Chr.1: 155237353-155237353 on Build GRCh38
rs1026559493 Chr.1: 155237072-155237072 on Build GRCh38
rs910331416 Chr.1: 155237101-155237101 on Build GRCh38
rs981698269 Chr.1: 155237122-155237122 on Build GRCh38
rs1010857275 Chr.1: 155236951-155236951 on Build GRCh38
rs764281170 Chr.1: 155237334-155237334 on Build GRCh38
rs564538639 Chr.1: 155236908-155236908 on Build GRCh38
rs533856111 Chr.1: 155237203-155237203 on Build GRCh38
rs35033592 Chr.1: 155237032-155237032 on Build GRCh38
rs778074573 Chr.1: 155237140-155237140 on Build GRCh38
rs370125418 Chr.1: 155237226-155237226 on Build GRCh38
rs545478869 Chr.1: 155237281-155237281 on Build GRCh38

View Full Product Details