Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155247965-155248472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189B
Entrez Gene ID
10712
Gene Name
family with sequence similarity 189 member B
Gene Aliases
C1orf2, COTE1
UniGene
Hs.348308
Gene Chromosome Location
Chr.1: 155247205-155255483 on Build GRCh38
Transcripts
NM_001267608.1 NM_006589.2 NM_198264.1 XM_005244845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367242 c.1600C>T p.L534F Chr.1: 155248283-155248283 on Build GRCh38
4978393 c.1543G>C p.E515Q Chr.1: 155248441-155248441 on Build GRCh38
1295166 c.1656G>C p.E552D Chr.1: 155248227-155248227 on Build GRCh38
3474899 c.1722C>T p.S574S Chr.1: 155248161-155248161 on Build GRCh38
3474898 c.1774C>T p.L592F Chr.1: 155248109-155248109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781670745 Chr.1: 155248011-155248011 on Build GRCh38
rs767941124 Chr.1: 155248108-155248108 on Build GRCh38
rs776540163 Chr.1: 155248162-155248162 on Build GRCh38
rs749980945 Chr.1: 155248233-155248233 on Build GRCh38
rs61732801 Chr.1: 155248283-155248283 on Build GRCh38
rs972648931 Chr.1: 155248141-155248141 on Build GRCh38
rs187706644 Chr.1: 155248431-155248431 on Build GRCh38
rs183179123 Chr.1: 155248268-155248268 on Build GRCh38
rs756717030 Chr.1: 155248129-155248129 on Build GRCh38
rs779785778 Chr.1: 155248148-155248148 on Build GRCh38
rs149473178 Chr.1: 155248230-155248230 on Build GRCh38
rs767172443 Chr.1: 155248422-155248422 on Build GRCh38
rs191547082 Chr.1: 155248267-155248267 on Build GRCh38
rs141156411 Chr.1: 155248183-155248183 on Build GRCh38
rs750943182 Chr.1: 155248311-155248311 on Build GRCh38
rs775722802 Chr.1: 155248387-155248387 on Build GRCh38
rs780804868 Chr.1: 155248351-155248351 on Build GRCh38
rs377201789 Chr.1: 155248419-155248419 on Build GRCh38
rs141883731 Chr.1: 155248154-155248154 on Build GRCh38
rs751095461 Chr.1: 155248409-155248409 on Build GRCh38
rs999885319 Chr.1: 155248121-155248121 on Build GRCh38
rs139386085 Chr.1: 155248277-155248277 on Build GRCh38
rs865832259 Chr.1: 155248382-155248382 on Build GRCh38
rs753768913 Chr.1: 155248437-155248437 on Build GRCh38
rs753012399 Chr.1: 155248211-155248211 on Build GRCh38
rs549321270 Chr.1: 155248160-155248160 on Build GRCh38
rs748772408 Chr.1: 155248040-155248040 on Build GRCh38
rs751728574 Chr.1: 155248083-155248083 on Build GRCh38
rs769030050 Chr.1: 155248390-155248390 on Build GRCh38
rs777582490 Chr.1: 155248323-155248323 on Build GRCh38
rs751148555 Chr.1: 155248337-155248337 on Build GRCh38
rs778321488 Chr.1: 155248333-155248333 on Build GRCh38
rs200607523 Chr.1: 155248039-155248039 on Build GRCh38
rs776759300 Chr.1: 155248392-155248392 on Build GRCh38
rs771779422 Chr.1: 155248145-155248145 on Build GRCh38
rs754642432 Chr.1: 155248339-155248339 on Build GRCh38
rs772210794 Chr.1: 155248386-155248386 on Build GRCh38
rs1056761847 Chr.1: 155248256-155248256 on Build GRCh38
rs368606056 Chr.1: 155248255-155248255 on Build GRCh38
rs767814005 Chr.1: 155248206-155248206 on Build GRCh38
rs759812062 Chr.1: 155248100-155248100 on Build GRCh38
rs754453209 Chr.1: 155248125-155248125 on Build GRCh38
rs777433089 Chr.1: 155248435-155248435 on Build GRCh38
rs531880757 Chr.1: 155248379-155248379 on Build GRCh38
rs745452883 Chr.1: 155248133-155248133 on Build GRCh38
rs896478611 Chr.1: 155248262-155248262 on Build GRCh38
rs991297444 Chr.1: 155248074-155248074 on Build GRCh38
rs970088462 Chr.1: 155248194-155248194 on Build GRCh38
rs150296362 Chr.1: 155248220-155248220 on Build GRCh38
rs761880750 Chr.1: 155248182-155248182 on Build GRCh38
rs41264929 Chr.1: 155248276-155248276 on Build GRCh38
rs754229836 Chr.1: 155248223-155248223 on Build GRCh38
rs148662301 Chr.1: 155248303-155248303 on Build GRCh38
rs201266020 Chr.1: 155248243-155248243 on Build GRCh38
rs1049941940 Chr.1: 155248186-155248186 on Build GRCh38
rs922260220 Chr.1: 155248164-155248164 on Build GRCh38
rs201013021 Chr.1: 155248114-155248114 on Build GRCh38
rs972618057 Chr.1: 155248324-155248324 on Build GRCh38
rs545648518 Chr.1: 155247991-155247991 on Build GRCh38
rs770520121 Chr.1: 155248047-155248047 on Build GRCh38
rs778416052 Chr.1: 155248050-155248050 on Build GRCh38
rs113547427 Chr.1: 155248377-155248377 on Build GRCh38
rs747940850 Chr.1: 155248355-155248355 on Build GRCh38
rs374790288 Chr.1: 155248326-155248326 on Build GRCh38
rs762921325 Chr.1: 155248078-155248078 on Build GRCh38
rs528673291 Chr.1: 155248159-155248159 on Build GRCh38
rs766542897 Chr.1: 155248081-155248081 on Build GRCh38
rs765244100 Chr.1: 155247990-155247990 on Build GRCh38
rs751860951 Chr.1: 155247997-155247997 on Build GRCh38
rs755929645 Chr.1: 155248432-155248432 on Build GRCh38
rs776591173 Chr.1: 155248072-155248072 on Build GRCh38
rs768490162 Chr.1: 155248161-155248161 on Build GRCh38
rs774506720 Chr.1: 155248185-155248185 on Build GRCh38
rs780240370 Chr.1: 155248443-155248443 on Build GRCh38
rs779660735 Chr.1: 155248242-155248242 on Build GRCh38
rs760557877 Chr.1: 155248067-155248067 on Build GRCh38
rs374893297 Chr.1: 155248222-155248222 on Build GRCh38
rs762215488 Chr.1: 155248394-155248394 on Build GRCh38
rs752348283 Chr.1: 155248423-155248423 on Build GRCh38
rs745600657 Chr.1: 155248060-155248060 on Build GRCh38
rs893702196 Chr.1: 155248331-155248331 on Build GRCh38
rs762187714 Chr.1: 155248302-155248302 on Build GRCh38
rs562632194 Chr.1: 155248008-155248008 on Build GRCh38
rs202173544 Chr.1: 155248115-155248115 on Build GRCh38
rs757796054 Chr.1: 155248227-155248227 on Build GRCh38
rs775275636 Chr.1: 155248061-155248061 on Build GRCh38
rs555295594 Chr.1: 155248372-155248372 on Build GRCh38
rs367769810 Chr.1: 155248070-155248070 on Build GRCh38
rs758579088 Chr.1: 155247992-155247992 on Build GRCh38
rs759635026 Chr.1: 155248200-155248200 on Build GRCh38
rs777075680 Chr.1: 155248297-155248297 on Build GRCh38
rs372840456 Chr.1: 155248244-155248244 on Build GRCh38
rs142191946 Chr.1: 155248329-155248329 on Build GRCh38
rs765580657 Chr.1: 155248396-155248396 on Build GRCh38
rs367558749 Chr.1: 155248238-155248238 on Build GRCh38
rs748602727 Chr.1: 155248023-155248023 on Build GRCh38
rs747179964 Chr.1: 155248388-155248388 on Build GRCh38
rs910895637 Chr.1: 155248361-155248361 on Build GRCh38
rs201906741 Chr.1: 155248010-155248010 on Build GRCh38
rs570944384 Chr.1: 155248399-155248399 on Build GRCh38
rs2075568 Chr.1: 155248377-155248377 on Build GRCh38
rs140120577 Chr.1: 155248439-155248439 on Build GRCh38
rs867740325 Chr.1: 155248153-155248153 on Build GRCh38
rs778488147 Chr.1: 155248130-155248130 on Build GRCh38

View Full Product Details