Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155250193-155250697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189B
Entrez Gene ID
10712
Gene Name
family with sequence similarity 189 member B
Gene Aliases
C1orf2, COTE1
UniGene
Hs.348308
Gene Chromosome Location
Chr.1: 155247205-155255483 on Build GRCh38
Transcripts
NM_001267608.1 NM_006589.2 NM_198264.1 XM_005244845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402755 c.1320G>T p.R440S Chr.1: 155250466-155250466 on Build GRCh38
132687 c.1228C>G p.P410A Chr.1: 155250558-155250558 on Build GRCh38
6397098 c.1385G>A p.R462H Chr.1: 155250401-155250401 on Build GRCh38
4847299 c.1491C>T p.I497I Chr.1: 155250295-155250295 on Build GRCh38
3689033 c.1324C>A p.R442R Chr.1: 155250462-155250462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016916386 Chr.1: 155250331-155250331 on Build GRCh38
rs373097775 Chr.1: 155250286-155250286 on Build GRCh38
rs911612235 Chr.1: 155250608-155250608 on Build GRCh38
rs540528085 Chr.1: 155250416-155250416 on Build GRCh38
rs761687447 Chr.1: 155250523-155250523 on Build GRCh38
rs979714163 Chr.1: 155250635-155250635 on Build GRCh38
rs746599359 Chr.1: 155250519-155250528 on Build GRCh38
rs868276096 Chr.1: 155250345-155250345 on Build GRCh38
rs959260709 Chr.1: 155250461-155250461 on Build GRCh38
rs768391432 Chr.1: 155250598-155250599 on Build GRCh38
rs150919039 Chr.1: 155250325-155250325 on Build GRCh38
rs896183593 Chr.1: 155250440-155250440 on Build GRCh38
rs1000466806 Chr.1: 155250371-155250371 on Build GRCh38
rs765067964 Chr.1: 155250636-155250636 on Build GRCh38
rs757157468 Chr.1: 155250319-155250319 on Build GRCh38
rs752324498 Chr.1: 155250280-155250280 on Build GRCh38
rs1006135553 Chr.1: 155250552-155250552 on Build GRCh38
rs538670178 Chr.1: 155250247-155250247 on Build GRCh38
rs765192462 Chr.1: 155250528-155250528 on Build GRCh38
rs201977546 Chr.1: 155250346-155250346 on Build GRCh38
rs758284321 Chr.1: 155250547-155250547 on Build GRCh38
rs201687408 Chr.1: 155250405-155250405 on Build GRCh38
rs937665274 Chr.1: 155250419-155250419 on Build GRCh38
rs964087093 Chr.1: 155250576-155250576 on Build GRCh38
rs751567836 Chr.1: 155250664-155250664 on Build GRCh38
rs777955078 Chr.1: 155250569-155250569 on Build GRCh38
rs763898256 Chr.1: 155250314-155250314 on Build GRCh38
rs774833185 Chr.1: 155250438-155250438 on Build GRCh38
rs766198748 Chr.1: 155250648-155250648 on Build GRCh38
rs774883352 Chr.1: 155250465-155250465 on Build GRCh38
rs956052904 Chr.1: 155250376-155250376 on Build GRCh38
rs776419224 Chr.1: 155250620-155250620 on Build GRCh38
rs946980779 Chr.1: 155250394-155250394 on Build GRCh38
rs74953707 Chr.1: 155250599-155250599 on Build GRCh38
rs900723865 Chr.1: 155250347-155250347 on Build GRCh38
rs753717767 Chr.1: 155250318-155250318 on Build GRCh38
rs142630415 Chr.1: 155250323-155250323 on Build GRCh38
rs1047498623 Chr.1: 155250307-155250307 on Build GRCh38
rs926523258 Chr.1: 155250216-155250216 on Build GRCh38
rs761562894 Chr.1: 155250623-155250623 on Build GRCh38
rs937940399 Chr.1: 155250236-155250236 on Build GRCh38
rs565282523 Chr.1: 155250446-155250446 on Build GRCh38
rs971893299 Chr.1: 155250434-155250434 on Build GRCh38
rs964390254 Chr.1: 155250360-155250360 on Build GRCh38
rs1056222684 Chr.1: 155250422-155250422 on Build GRCh38
rs748347744 Chr.1: 155250404-155250404 on Build GRCh38
rs374832056 Chr.1: 155250406-155250406 on Build GRCh38
rs896351946 Chr.1: 155250244-155250244 on Build GRCh38
rs768339669 Chr.1: 155250610-155250610 on Build GRCh38
rs749859862 Chr.1: 155250495-155250496 on Build GRCh38
rs926463771 Chr.1: 155250459-155250459 on Build GRCh38
rs750319934 Chr.1: 155250540-155250540 on Build GRCh38
rs544625953 Chr.1: 155250558-155250558 on Build GRCh38
rs914170380 Chr.1: 155250383-155250383 on Build GRCh38
rs1026189635 Chr.1: 155250259-155250259 on Build GRCh38
rs753594378 Chr.1: 155250518-155250518 on Build GRCh38
rs200219862 Chr.1: 155250502-155250502 on Build GRCh38
rs1037302482 Chr.1: 155250260-155250260 on Build GRCh38
rs929008504 Chr.1: 155250596-155250596 on Build GRCh38
rs749575079 Chr.1: 155250574-155250574 on Build GRCh38
rs145532508 Chr.1: 155250285-155250285 on Build GRCh38
rs760335191 Chr.1: 155250493-155250493 on Build GRCh38
rs779260143 Chr.1: 155250588-155250588 on Build GRCh38
rs377616047 Chr.1: 155250373-155250373 on Build GRCh38
rs993701245 Chr.1: 155250248-155250248 on Build GRCh38
rs767587475 Chr.1: 155250555-155250555 on Build GRCh38
rs374123477 Chr.1: 155250352-155250352 on Build GRCh38
rs922645232 Chr.1: 155250368-155250368 on Build GRCh38
rs756140855 Chr.1: 155250669-155250669 on Build GRCh38
rs771338708 Chr.1: 155250432-155250432 on Build GRCh38
rs749626426 Chr.1: 155250417-155250417 on Build GRCh38
rs776103856 Chr.1: 155250603-155250603 on Build GRCh38
rs756261217 Chr.1: 155250567-155250567 on Build GRCh38
rs760038265 Chr.1: 155250445-155250445 on Build GRCh38
rs944467161 Chr.1: 155250616-155250616 on Build GRCh38
rs375107408 Chr.1: 155250647-155250647 on Build GRCh38
rs929040983 Chr.1: 155250302-155250302 on Build GRCh38
rs746478595 Chr.1: 155250609-155250609 on Build GRCh38
rs773166356 Chr.1: 155250639-155250639 on Build GRCh38
rs568409290 Chr.1: 155250348-155250348 on Build GRCh38
rs757549144 Chr.1: 155250580-155250580 on Build GRCh38

View Full Product Details