Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155251007-155251519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189B
Entrez Gene ID
10712
Gene Name
family with sequence similarity 189 member B
Gene Aliases
C1orf2, COTE1
UniGene
Hs.348308
Gene Chromosome Location
Chr.1: 155247205-155255483 on Build GRCh38
Transcripts
NM_001267608.1 NM_006589.2 NM_198264.1 XM_005244845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2210290 c.925G>A p.G309S Chr.1: 155251134-155251134 on Build GRCh38
4398843 c.951C>T p.A317A Chr.1: 155251108-155251108 on Build GRCh38
5649068 c.903C>T p.L301L Chr.1: 155251156-155251156 on Build GRCh38
4682825 c.943C>T p.R315* Chr.1: 155251116-155251116 on Build GRCh38
7138785 c.913C>G p.Q305E Chr.1: 155251146-155251146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756664090 Chr.1: 155251130-155251130 on Build GRCh38
rs542308432 Chr.1: 155251425-155251425 on Build GRCh38
rs755437309 Chr.1: 155251054-155251054 on Build GRCh38
rs757292283 Chr.1: 155251121-155251121 on Build GRCh38
rs140698922 Chr.1: 155251304-155251304 on Build GRCh38
rs200662522 Chr.1: 155251487-155251487 on Build GRCh38
rs764614621 Chr.1: 155251465-155251465 on Build GRCh38
rs758598226 Chr.1: 155251492-155251492 on Build GRCh38
rs754315860 Chr.1: 155251469-155251469 on Build GRCh38
rs769706484 Chr.1: 155251093-155251093 on Build GRCh38
rs768065778 Chr.1: 155251116-155251116 on Build GRCh38
rs768671471 Chr.1: 155251176-155251176 on Build GRCh38
rs983796366 Chr.1: 155251213-155251213 on Build GRCh38
rs758094983 Chr.1: 155251486-155251486 on Build GRCh38
rs201258573 Chr.1: 155251136-155251136 on Build GRCh38
rs763813056 Chr.1: 155251106-155251106 on Build GRCh38
rs143355494 Chr.1: 155251088-155251088 on Build GRCh38
rs777345764 Chr.1: 155251062-155251062 on Build GRCh38
rs748841184 Chr.1: 155251064-155251064 on Build GRCh38
rs966132976 Chr.1: 155251257-155251257 on Build GRCh38
rs756721419 Chr.1: 155251474-155251474 on Build GRCh38
rs749369109 Chr.1: 155251199-155251199 on Build GRCh38
rs759655945 Chr.1: 155251058-155251058 on Build GRCh38
rs753356393 Chr.1: 155251137-155251137 on Build GRCh38
rs767841313 Chr.1: 155251078-155251089 on Build GRCh38
rs760979630 Chr.1: 155251464-155251464 on Build GRCh38
rs533360424 Chr.1: 155251296-155251296 on Build GRCh38
rs755614526 Chr.1: 155251061-155251061 on Build GRCh38
rs112300182 Chr.1: 155251303-155251303 on Build GRCh38
rs775402105 Chr.1: 155251085-155251085 on Build GRCh38
rs536355967 Chr.1: 155251074-155251074 on Build GRCh38
rs946304066 Chr.1: 155251437-155251437 on Build GRCh38
rs774714417 Chr.1: 155251211-155251211 on Build GRCh38
rs750366503 Chr.1: 155251308-155251308 on Build GRCh38
rs373214687 Chr.1: 155251115-155251115 on Build GRCh38
rs763048566 Chr.1: 155251099-155251099 on Build GRCh38
rs372755199 Chr.1: 155251055-155251055 on Build GRCh38
rs374391371 Chr.1: 155251477-155251477 on Build GRCh38
rs892021484 Chr.1: 155251049-155251049 on Build GRCh38
rs771911280 Chr.1: 155251084-155251084 on Build GRCh38
rs778982894 Chr.1: 155251463-155251463 on Build GRCh38
rs139005146 Chr.1: 155251232-155251232 on Build GRCh38
rs751323051 Chr.1: 155251107-155251107 on Build GRCh38
rs764795248 Chr.1: 155251476-155251476 on Build GRCh38
rs774243806 Chr.1: 155251049-155251084 on Build GRCh38
rs568471287 Chr.1: 155251138-155251138 on Build GRCh38
rs566720731 Chr.1: 155251095-155251095 on Build GRCh38
rs577492637 Chr.1: 155251202-155251202 on Build GRCh38
rs1051684500 Chr.1: 155251281-155251281 on Build GRCh38
rs755167737 Chr.1: 155251326-155251326 on Build GRCh38
rs368261049 Chr.1: 155251193-155251193 on Build GRCh38
rs758123518 Chr.1: 155251160-155251160 on Build GRCh38
rs759847327 Chr.1: 155251459-155251459 on Build GRCh38
rs968098738 Chr.1: 155251377-155251377 on Build GRCh38
rs776475829 Chr.1: 155251187-155251187 on Build GRCh38
rs779838603 Chr.1: 155251164-155251164 on Build GRCh38
rs990290981 Chr.1: 155251318-155251318 on Build GRCh38
rs745640277 Chr.1: 155251081-155251081 on Build GRCh38
rs767941030 Chr.1: 155251460-155251460 on Build GRCh38
rs781319458 Chr.1: 155251141-155251141 on Build GRCh38
rs1001045584 Chr.1: 155251381-155251381 on Build GRCh38

View Full Product Details