Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155263137-155263653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK2
Entrez Gene ID
1196
Gene Name
CDC like kinase 2
Gene Aliases
-
UniGene
Hs.73986
Gene Chromosome Location
Chr.1: 155262868-155273529 on Build GRCh38
Transcripts
NM_001294338.1 NM_001294339.1 NM_003993.3 XM_005244878.1 XM_011509143.1 XM_011509147.2 XM_017000241.1 XM_017000242.1 XM_017000243.1 XM_017000244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2210337 c.1395G>T p.K465N Chr.1: 155263320-155263320 on Build GRCh38
7116528 c.1366G>A p.E456K Chr.1: 155263349-155263349 on Build GRCh38
896848 c.1316G>A p.R439Q Chr.1: 155263399-155263399 on Build GRCh38
3474908 c.1356C>T p.F452F Chr.1: 155263359-155263359 on Build GRCh38
6630490 c.1423C>T p.H475Y Chr.1: 155263292-155263292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs36053924 Chr.1: 155263275-155263275 on Build GRCh38
rs201056646 Chr.1: 155263414-155263414 on Build GRCh38
rs909544886 Chr.1: 155263174-155263174 on Build GRCh38
rs201343633 Chr.1: 155263438-155263438 on Build GRCh38
rs776265874 Chr.1: 155263609-155263609 on Build GRCh38
rs993038124 Chr.1: 155263405-155263405 on Build GRCh38
rs751860229 Chr.1: 155263256-155263256 on Build GRCh38
rs892293018 Chr.1: 155263616-155263616 on Build GRCh38
rs139671051 Chr.1: 155263396-155263396 on Build GRCh38
rs368845441 Chr.1: 155263167-155263167 on Build GRCh38
rs1008499760 Chr.1: 155263380-155263380 on Build GRCh38
rs771979441 Chr.1: 155263212-155263212 on Build GRCh38
rs542635243 Chr.1: 155263554-155263554 on Build GRCh38
rs373658244 Chr.1: 155263409-155263409 on Build GRCh38
rs745674031 Chr.1: 155263276-155263276 on Build GRCh38
rs778696939 Chr.1: 155263359-155263359 on Build GRCh38
rs987892179 Chr.1: 155263224-155263224 on Build GRCh38
rs147637184 Chr.1: 155263251-155263251 on Build GRCh38
rs933717290 Chr.1: 155263603-155263603 on Build GRCh38
rs1019418225 Chr.1: 155263388-155263388 on Build GRCh38
rs938246633 Chr.1: 155263608-155263608 on Build GRCh38
rs996682954 Chr.1: 155263478-155263478 on Build GRCh38
rs537075843 Chr.1: 155263193-155263193 on Build GRCh38
rs375003420 Chr.1: 155263614-155263617 on Build GRCh38
rs1046542228 Chr.1: 155263258-155263258 on Build GRCh38
rs930196230 Chr.1: 155263163-155263163 on Build GRCh38
rs750093925 Chr.1: 155263368-155263368 on Build GRCh38
rs372227526 Chr.1: 155263211-155263211 on Build GRCh38
rs746962823 Chr.1: 155263280-155263280 on Build GRCh38
rs747197029 Chr.1: 155263406-155263408 on Build GRCh38
rs771821636 Chr.1: 155263277-155263277 on Build GRCh38
rs760232768 Chr.1: 155263557-155263557 on Build GRCh38
rs773683868 Chr.1: 155263547-155263547 on Build GRCh38
rs951647215 Chr.1: 155263410-155263410 on Build GRCh38
rs942304354 Chr.1: 155263443-155263443 on Build GRCh38
rs111367678 Chr.1: 155263610-155263613 on Build GRCh38
rs982702178 Chr.1: 155263574-155263574 on Build GRCh38
rs779277228 Chr.1: 155263203-155263203 on Build GRCh38
rs765285447 Chr.1: 155263227-155263227 on Build GRCh38
rs759928699 Chr.1: 155263313-155263313 on Build GRCh38
rs773231332 Chr.1: 155263288-155263288 on Build GRCh38
rs762252894 Chr.1: 155263508-155263508 on Build GRCh38
rs753215592 Chr.1: 155263335-155263335 on Build GRCh38
rs777465511 Chr.1: 155263269-155263269 on Build GRCh38
rs202231278 Chr.1: 155263311-155263311 on Build GRCh38
rs756894509 Chr.1: 155263358-155263358 on Build GRCh38
rs545679414 Chr.1: 155263425-155263425 on Build GRCh38
rs534870900 Chr.1: 155263307-155263307 on Build GRCh38
rs745672736 Chr.1: 155263207-155263207 on Build GRCh38
rs775358392 Chr.1: 155263213-155263213 on Build GRCh38
rs878927862 Chr.1: 155263441-155263441 on Build GRCh38
rs913666414 Chr.1: 155263561-155263561 on Build GRCh38
rs758184333 Chr.1: 155263389-155263389 on Build GRCh38
rs369716792 Chr.1: 155263355-155263355 on Build GRCh38
rs1049090 Chr.1: 155263324-155263324 on Build GRCh38
rs773395300 Chr.1: 155263238-155263238 on Build GRCh38
rs763125375 Chr.1: 155263243-155263243 on Build GRCh38
rs2990237 Chr.1: 155263579-155263579 on Build GRCh38
rs748615667 Chr.1: 155263400-155263400 on Build GRCh38
rs759716885 Chr.1: 155263448-155263448 on Build GRCh38
rs761045355 Chr.1: 155263338-155263338 on Build GRCh38
rs753200761 Chr.1: 155263263-155263263 on Build GRCh38
rs890531904 Chr.1: 155263376-155263376 on Build GRCh38
rs555477843 Chr.1: 155263201-155263201 on Build GRCh38
rs148908482 Chr.1: 155263216-155263216 on Build GRCh38
rs774619061 Chr.1: 155263312-155263312 on Build GRCh38
rs920838931 Chr.1: 155263612-155263612 on Build GRCh38
rs765866255 Chr.1: 155263594-155263594 on Build GRCh38
rs768060382 Chr.1: 155263261-155263261 on Build GRCh38
rs373524272 Chr.1: 155263260-155263260 on Build GRCh38
rs150456292 Chr.1: 155263342-155263342 on Build GRCh38
rs772594143 Chr.1: 155263518-155263518 on Build GRCh38
rs767818963 Chr.1: 155263326-155263326 on Build GRCh38
rs970480708 Chr.1: 155263568-155263568 on Build GRCh38
rs774246504 Chr.1: 155263202-155263202 on Build GRCh38
rs909554887 Chr.1: 155263437-155263437 on Build GRCh38
rs768615621 Chr.1: 155263281-155263281 on Build GRCh38
rs781108604 Chr.1: 155263413-155263413 on Build GRCh38
rs527508796 Chr.1: 155263187-155263188 on Build GRCh38
rs753428819 Chr.1: 155263270-155263270 on Build GRCh38
rs780021739 Chr.1: 155263279-155263279 on Build GRCh38
rs775175824 Chr.1: 155263252-155263254 on Build GRCh38
rs575212862 Chr.1: 155263487-155263487 on Build GRCh38
rs200444017 Chr.1: 155263271-155263271 on Build GRCh38
rs913647980 Chr.1: 155263236-155263236 on Build GRCh38
rs755626349 Chr.1: 155263266-155263266 on Build GRCh38
rs1039420776 Chr.1: 155263197-155263197 on Build GRCh38

View Full Product Details