Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155312173-155312672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDPS
Entrez Gene ID
2224
Gene Name
farnesyl diphosphate synthase
Gene Aliases
FPPS, FPS, POROK9
UniGene
Hs.335918
Gene Chromosome Location
Chr.1: 155308748-155320666 on Build GRCh38
Transcripts
NM_001135821.1 NM_001135822.1 NM_001242824.1 NM_001242825.1 NM_002004.3 XM_005244962.1 XM_005244963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817291 c.448C>A p.R150R Chr.1: 155312363-155312363 on Build GRCh38
5774788 c.340-5T>A p.? Chr.1: 155312250-155312250 on Build GRCh38
346543 c.456G>T p.W152C Chr.1: 155312371-155312371 on Build GRCh38
424048 c.458C>A p.T153N Chr.1: 155312373-155312373 on Build GRCh38
4023005 c.449G>A p.R150Q Chr.1: 155312364-155312364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2148136 Chr.1: 155312273-155312273 on Build GRCh38
rs763852459 Chr.1: 155312310-155312310 on Build GRCh38
rs1032408070 Chr.1: 155312548-155312549 on Build GRCh38
rs959758430 Chr.1: 155312607-155312607 on Build GRCh38
rs907532468 Chr.1: 155312268-155312268 on Build GRCh38
rs752341142 Chr.1: 155312301-155312301 on Build GRCh38
rs548425134 Chr.1: 155312632-155312632 on Build GRCh38
rs760944514 Chr.1: 155312472-155312472 on Build GRCh38
rs973066799 Chr.1: 155312637-155312637 on Build GRCh38
rs933183034 Chr.1: 155312533-155312533 on Build GRCh38
rs781491033 Chr.1: 155312374-155312374 on Build GRCh38
rs35806025 Chr.1: 155312222-155312223 on Build GRCh38
rs758912651 Chr.1: 155312286-155312286 on Build GRCh38
rs773490730 Chr.1: 155312253-155312253 on Build GRCh38
rs1031395572 Chr.1: 155312604-155312604 on Build GRCh38
rs993376440 Chr.1: 155312335-155312335 on Build GRCh38
rs1052011868 Chr.1: 155312550-155312550 on Build GRCh38
rs111791396 Chr.1: 155312396-155312396 on Build GRCh38
rs192217770 Chr.1: 155312518-155312518 on Build GRCh38
rs940330939 Chr.1: 155312633-155312633 on Build GRCh38
rs766748706 Chr.1: 155312271-155312271 on Build GRCh38
rs753523692 Chr.1: 155312318-155312318 on Build GRCh38
rs376502714 Chr.1: 155312264-155312264 on Build GRCh38
rs868642909 Chr.1: 155312372-155312372 on Build GRCh38
rs1037142178 Chr.1: 155312487-155312487 on Build GRCh38
rs141763891 Chr.1: 155312424-155312424 on Build GRCh38
rs990958989 Chr.1: 155312587-155312587 on Build GRCh38
rs770189684 Chr.1: 155312394-155312394 on Build GRCh38
rs139247941 Chr.1: 155312640-155312640 on Build GRCh38
rs778927797 Chr.1: 155312222-155312222 on Build GRCh38
rs777350213 Chr.1: 155312598-155312598 on Build GRCh38
rs748441551 Chr.1: 155312242-155312242 on Build GRCh38
rs774705644 Chr.1: 155312410-155312410 on Build GRCh38
rs902500622 Chr.1: 155312477-155312478 on Build GRCh38
rs916182747 Chr.1: 155312354-155312354 on Build GRCh38
rs771339846 Chr.1: 155312405-155312405 on Build GRCh38
rs200386627 Chr.1: 155312386-155312386 on Build GRCh38
rs370107498 Chr.1: 155312361-155312361 on Build GRCh38
rs569302174 Chr.1: 155312204-155312204 on Build GRCh38
rs778653025 Chr.1: 155312338-155312338 on Build GRCh38
rs577190794 Chr.1: 155312600-155312600 on Build GRCh38
rs200530077 Chr.1: 155312383-155312383 on Build GRCh38
rs982765621 Chr.1: 155312620-155312620 on Build GRCh38
rs757597083 Chr.1: 155312447-155312448 on Build GRCh38
rs770279503 Chr.1: 155312245-155312245 on Build GRCh38
rs11556432 Chr.1: 155312319-155312319 on Build GRCh38
rs531725009 Chr.1: 155312216-155312216 on Build GRCh38
rs767069544 Chr.1: 155312292-155312292 on Build GRCh38
rs150590717 Chr.1: 155312589-155312589 on Build GRCh38
rs771536978 Chr.1: 155312428-155312428 on Build GRCh38
rs999640276 Chr.1: 155312521-155312521 on Build GRCh38
rs748351433 Chr.1: 155312385-155312385 on Build GRCh38
rs749216760 Chr.1: 155312219-155312219 on Build GRCh38
rs534532668 Chr.1: 155312581-155312581 on Build GRCh38
rs373639609 Chr.1: 155312364-155312364 on Build GRCh38
rs763975527 Chr.1: 155312401-155312402 on Build GRCh38
rs773507199 Chr.1: 155312397-155312397 on Build GRCh38
rs755801542 Chr.1: 155312302-155312302 on Build GRCh38
rs747024248 Chr.1: 155312235-155312235 on Build GRCh38
rs757187037 Chr.1: 155312221-155312221 on Build GRCh38
rs768866332 Chr.1: 155312238-155312238 on Build GRCh38
rs749547725 Chr.1: 155312398-155312398 on Build GRCh38
rs899020702 Chr.1: 155312330-155312330 on Build GRCh38
rs1007922025 Chr.1: 155312579-155312579 on Build GRCh38
rs61729685 Chr.1: 155312274-155312274 on Build GRCh38
rs901248142 Chr.1: 155312502-155312502 on Build GRCh38
rs755888309 Chr.1: 155312214-155312214 on Build GRCh38
rs1037842685 Chr.1: 155312329-155312329 on Build GRCh38
rs367778160 Chr.1: 155312441-155312441 on Build GRCh38
rs1048398740 Chr.1: 155312379-155312379 on Build GRCh38
rs11556437 Chr.1: 155312333-155312333 on Build GRCh38
rs889044978 Chr.1: 155312567-155312567 on Build GRCh38
rs890385049 Chr.1: 155312362-155312362 on Build GRCh38
rs753971608 Chr.1: 155312417-155312419 on Build GRCh38
rs779109918 Chr.1: 155312447-155312447 on Build GRCh38

View Full Product Details