Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155688376-155688873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
XM_017002291.1 XM_017002292.1 XM_017002294.1 XM_017002295.1 NM_001198900.1 NM_001198901.1 NM_001198902.1 NM_001198903.1 NM_001198904.1 NM_001198905.1 NM_001198906.1 NM_018253.3 NM_139118.2 NM_139119.2 NM_139121.2

Gene Symbol
YY1AP1
Entrez Gene ID
55249
Gene Name
YY1 associated protein 1
Gene Aliases
HCCA1, HCCA2, YAP, YY1AP
UniGene
Hs.584927
Gene Chromosome Location
Chr.1: 155659442-155689032 on Build GRCh38
Transcripts
XM_017002291.1 XM_017002292.1 XM_017002294.1 XM_017002295.1 NM_001198900.1 NM_001198901.1 NM_001198902.1 NM_001198903.1 NM_001198904.1 NM_001198905.1 NM_001198906.1 NM_018253.3 NM_139118.2 NM_139119.2 NM_139121.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4603881 c.38_40delGAA p.R13delR Chr.1: 155688425-155688427 on Build GRCh38
1578473 c.71_73TGT>GGG p.V24_S25>GA Chr.1: 155688392-155688394 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs955953653 Chr.1: 155688408-155688408 on Build GRCh38
rs111741196 Chr.1: 155688436-155688436 on Build GRCh38
rs181621149 Chr.1: 155688809-155688809 on Build GRCh38
rs756380288 Chr.1: 155688645-155688645 on Build GRCh38
rs28362635 Chr.1: 155688648-155688648 on Build GRCh38
rs546473729 Chr.1: 155688827-155688827 on Build GRCh38
rs960260176 Chr.1: 155688706-155688706 on Build GRCh38
rs749575750 Chr.1: 155688801-155688801 on Build GRCh38
rs572593060 Chr.1: 155688819-155688819 on Build GRCh38
rs774098986 Chr.1: 155688433-155688433 on Build GRCh38
rs570422110 Chr.1: 155688647-155688647 on Build GRCh38
rs1039306078 Chr.1: 155688556-155688556 on Build GRCh38
rs1973600 Chr.1: 155688438-155688438 on Build GRCh38
rs199955075 Chr.1: 155688464-155688464 on Build GRCh38
rs983551686 Chr.1: 155688547-155688547 on Build GRCh38
rs778798759 Chr.1: 155688654-155688654 on Build GRCh38
rs921914458 Chr.1: 155688746-155688746 on Build GRCh38
rs904200988 Chr.1: 155688679-155688679 on Build GRCh38
rs201373497 Chr.1: 155688460-155688460 on Build GRCh38
rs200747892 Chr.1: 155688477-155688477 on Build GRCh38
rs199861612 Chr.1: 155688447-155688447 on Build GRCh38
rs1030099653 Chr.1: 155688399-155688399 on Build GRCh38
rs566359631 Chr.1: 155688576-155688576 on Build GRCh38
rs929403684 Chr.1: 155688506-155688506 on Build GRCh38
rs1034191921 Chr.1: 155688472-155688472 on Build GRCh38
rs529365618 Chr.1: 155688566-155688566 on Build GRCh38
rs189141963 Chr.1: 155688567-155688567 on Build GRCh38
rs41264947 Chr.1: 155688479-155688479 on Build GRCh38
rs556204470 Chr.1: 155688713-155688713 on Build GRCh38
rs867985922 Chr.1: 155688734-155688734 on Build GRCh38
rs201567646 Chr.1: 155688463-155688463 on Build GRCh38
rs550152287 Chr.1: 155688493-155688493 on Build GRCh38
rs984822275 Chr.1: 155688830-155688830 on Build GRCh38
rs1001229156 Chr.1: 155688844-155688844 on Build GRCh38
rs780567044 Chr.1: 155688726-155688726 on Build GRCh38
rs200247944 Chr.1: 155688710-155688710 on Build GRCh38
rs979997894 Chr.1: 155688459-155688459 on Build GRCh38
rs535731792 Chr.1: 155688762-155688763 on Build GRCh38
rs560678874 Chr.1: 155688748-155688748 on Build GRCh38
rs993043963 Chr.1: 155688489-155688489 on Build GRCh38
rs564498010 Chr.1: 155688434-155688434 on Build GRCh38
rs678157 Chr.1: 155688550-155688550 on Build GRCh38
rs951236395 Chr.1: 155688728-155688728 on Build GRCh38
rs1021296153 Chr.1: 155688669-155688669 on Build GRCh38
rs983948468 Chr.1: 155688740-155688740 on Build GRCh38
rs891961492 Chr.1: 155688594-155688594 on Build GRCh38
rs115908216 Chr.1: 155688743-155688743 on Build GRCh38
rs368170583 Chr.1: 155688703-155688703 on Build GRCh38
rs942027025 Chr.1: 155688741-155688741 on Build GRCh38
rs968348515 Chr.1: 155688446-155688446 on Build GRCh38
rs558617497 Chr.1: 155688591-155688591 on Build GRCh38
rs770337647 Chr.1: 155688398-155688398 on Build GRCh38
rs184902288 Chr.1: 155688810-155688810 on Build GRCh38
rs941974670 Chr.1: 155688554-155688554 on Build GRCh38
rs992737707 Chr.1: 155688709-155688709 on Build GRCh38
rs933607591 Chr.1: 155688755-155688755 on Build GRCh38
rs1025935379 Chr.1: 155688727-155688731 on Build GRCh38
rs1021286154 Chr.1: 155688439-155688439 on Build GRCh38
rs1001134398 Chr.1: 155688680-155688680 on Build GRCh38
rs534077108 Chr.1: 155688580-155688580 on Build GRCh38

View Full Product Details