Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155688850-155689353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1 NM_001198901.1 NM_001198906.1 NM_139118.2 NM_139121.2

Gene Symbol
YY1AP1
Entrez Gene ID
55249
Gene Name
YY1 associated protein 1
Gene Aliases
HCCA1, HCCA2, YAP, YY1AP
UniGene
Hs.584927
Gene Chromosome Location
Chr.1: 155659442-155689032 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1 NM_001198901.1 NM_001198906.1 NM_139118.2 NM_139121.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6533916 c.43G>C p.G15R Chr.1: 155688934-155688934 on Build GRCh38
5772208 c.14C>T p.A5V Chr.1: 155688963-155688963 on Build GRCh38
3770246 c.29C>G p.A10G Chr.1: 155688948-155688948 on Build GRCh38
3474994 c.36G>A p.T12T Chr.1: 155688941-155688941 on Build GRCh38
6241535 c.47+7T>G p.? Chr.1: 155688923-155688923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs559794230 Chr.1: 155689165-155689165 on Build GRCh38
rs996270587 Chr.1: 155689102-155689102 on Build GRCh38
rs967629512 Chr.1: 155689126-155689126 on Build GRCh38
rs1009376220 Chr.1: 155689312-155689312 on Build GRCh38
rs745553832 Chr.1: 155688925-155688925 on Build GRCh38
rs553958470 Chr.1: 155689271-155689271 on Build GRCh38
rs771702169 Chr.1: 155688987-155688987 on Build GRCh38
rs890571520 Chr.1: 155689307-155689307 on Build GRCh38
rs770603355 Chr.1: 155689250-155689250 on Build GRCh38
rs746636677 Chr.1: 155689003-155689003 on Build GRCh38
rs886755965 Chr.1: 155689244-155689244 on Build GRCh38
rs776241568 Chr.1: 155689013-155689013 on Build GRCh38
rs767796919 Chr.1: 155688964-155688964 on Build GRCh38
rs781551482 Chr.1: 155688898-155688898 on Build GRCh38
rs1029503385 Chr.1: 155689291-155689291 on Build GRCh38
rs751838515 Chr.1: 155688890-155688890 on Build GRCh38
rs768604709 Chr.1: 155689010-155689010 on Build GRCh38
rs745307798 Chr.1: 155688978-155688978 on Build GRCh38
rs369751610 Chr.1: 155688991-155688991 on Build GRCh38
rs773200259 Chr.1: 155689023-155689023 on Build GRCh38
rs4824 Chr.1: 155689149-155689149 on Build GRCh38
rs189413971 Chr.1: 155689119-155689119 on Build GRCh38
rs759672644 Chr.1: 155689025-155689025 on Build GRCh38
rs1016801674 Chr.1: 155689078-155689078 on Build GRCh38
rs887000836 Chr.1: 155689030-155689030 on Build GRCh38
rs756523686 Chr.1: 155688966-155688966 on Build GRCh38
rs369012638 Chr.1: 155689284-155689284 on Build GRCh38
rs1038260687 Chr.1: 155689266-155689266 on Build GRCh38
rs512551 Chr.1: 155688881-155688881 on Build GRCh38
rs949765672 Chr.1: 155689203-155689203 on Build GRCh38
rs760613801 Chr.1: 155688934-155688934 on Build GRCh38
rs757652844 Chr.1: 155688972-155688972 on Build GRCh38
rs1020320666 Chr.1: 155689313-155689313 on Build GRCh38
rs758562980 Chr.1: 155688886-155688886 on Build GRCh38
rs375716262 Chr.1: 155688938-155688938 on Build GRCh38
rs1005458751 Chr.1: 155689071-155689071 on Build GRCh38
rs529680175 Chr.1: 155689090-155689090 on Build GRCh38
rs41264953 Chr.1: 155689253-155689253 on Build GRCh38
rs1047440327 Chr.1: 155689027-155689027 on Build GRCh38
rs372502183 Chr.1: 155689021-155689021 on Build GRCh38
rs774577192 Chr.1: 155689283-155689283 on Build GRCh38
rs147058489 Chr.1: 155688975-155688975 on Build GRCh38
rs662116 Chr.1: 155689328-155689328 on Build GRCh38
rs996658130 Chr.1: 155689288-155689288 on Build GRCh38
rs1055470894 Chr.1: 155689195-155689195 on Build GRCh38
rs755201199 Chr.1: 155688963-155688963 on Build GRCh38
rs963866056 Chr.1: 155689080-155689080 on Build GRCh38
rs747549507 Chr.1: 155689092-155689092 on Build GRCh38
rs41264951 Chr.1: 155689137-155689137 on Build GRCh38
rs543380974 Chr.1: 155688907-155688907 on Build GRCh38
rs946221999 Chr.1: 155688957-155688957 on Build GRCh38
rs41264949 Chr.1: 155689073-155689073 on Build GRCh38
rs771789810 Chr.1: 155688930-155688930 on Build GRCh38
rs376878473 Chr.1: 155688910-155688910 on Build GRCh38
rs780180211 Chr.1: 155689022-155689022 on Build GRCh38
rs749828251 Chr.1: 155688971-155688971 on Build GRCh38
rs765019580 Chr.1: 155689022-155689022 on Build GRCh38
rs181055129 Chr.1: 155689123-155689123 on Build GRCh38
rs371563232 Chr.1: 155688913-155688913 on Build GRCh38
rs951291827 Chr.1: 155688896-155688896 on Build GRCh38
rs779967031 Chr.1: 155688887-155688887 on Build GRCh38
rs764054066 Chr.1: 155688880-155688880 on Build GRCh38
rs374163667 Chr.1: 155688875-155688875 on Build GRCh38
rs761825516 Chr.1: 155689020-155689020 on Build GRCh38
rs1025524201 Chr.1: 155688892-155688892 on Build GRCh38
rs532041441 Chr.1: 155688902-155688902 on Build GRCh38
rs752907312 Chr.1: 155688965-155688965 on Build GRCh38
rs778076319 Chr.1: 155688967-155688967 on Build GRCh38
rs916981289 Chr.1: 155689201-155689201 on Build GRCh38
rs755107941 Chr.1: 155688895-155688895 on Build GRCh38
rs759531821 Chr.1: 155689265-155689265 on Build GRCh38
rs913462326 Chr.1: 155688945-155688945 on Build GRCh38
rs775428208 Chr.1: 155688932-155688932 on Build GRCh38
rs373858221 Chr.1: 155688903-155688903 on Build GRCh38
rs1046756003 Chr.1: 155689221-155689221 on Build GRCh38
rs939190212 Chr.1: 155689164-155689164 on Build GRCh38
rs117182857 Chr.1: 155689104-155689104 on Build GRCh38
rs555991092 Chr.1: 155689146-155689147 on Build GRCh38
rs766504302 Chr.1: 155688953-155688953 on Build GRCh38
rs34413968 Chr.1: 155688931-155688932 on Build GRCh38
rs368291594 Chr.1: 155688922-155688922 on Build GRCh38
rs150181705 Chr.1: 155688939-155688939 on Build GRCh38
rs751537839 Chr.1: 155688958-155688958 on Build GRCh38

View Full Product Details