Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155716629-155717175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002293.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640002 c.58C>A p.R20S Chr.1: 155717018-155717018 on Build GRCh38
3802125 c.103C>T p.H35Y Chr.1: 155717063-155717063 on Build GRCh38
6640001 c.135G>A p.P45P Chr.1: 155717095-155717095 on Build GRCh38
896970 c.117G>T p.Q39H Chr.1: 155717077-155717077 on Build GRCh38
1293219 c.157G>A p.E53K Chr.1: 155717117-155717117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200523835 Chr.1: 155717054-155717054 on Build GRCh38
rs775041378 Chr.1: 155717138-155717138 on Build GRCh38
rs370828134 Chr.1: 155717143-155717143 on Build GRCh38
rs769520204 Chr.1: 155716979-155716979 on Build GRCh38
rs752102492 Chr.1: 155717074-155717074 on Build GRCh38
rs974570033 Chr.1: 155716798-155716798 on Build GRCh38
rs370236932 Chr.1: 155717093-155717093 on Build GRCh38
rs1034711182 Chr.1: 155716719-155716719 on Build GRCh38
rs775549205 Chr.1: 155717031-155717031 on Build GRCh38
rs375876872 Chr.1: 155717113-155717113 on Build GRCh38
rs369774038 Chr.1: 155717046-155717046 on Build GRCh38
rs148684040 Chr.1: 155717128-155717128 on Build GRCh38
rs145929335 Chr.1: 155717045-155717045 on Build GRCh38
rs139804160 Chr.1: 155717068-155717068 on Build GRCh38
rs373473811 Chr.1: 155716908-155716908 on Build GRCh38
rs534139079 Chr.1: 155716787-155716787 on Build GRCh38
rs776731938 Chr.1: 155717107-155717107 on Build GRCh38
rs373966776 Chr.1: 155717094-155717094 on Build GRCh38
rs962569039 Chr.1: 155716736-155716736 on Build GRCh38
rs986524820 Chr.1: 155716815-155716815 on Build GRCh38
rs760054393 Chr.1: 155716966-155716970 on Build GRCh38
rs569201309 Chr.1: 155716840-155716840 on Build GRCh38
rs768765455 Chr.1: 155717038-155717038 on Build GRCh38
rs755738906 Chr.1: 155717085-155717085 on Build GRCh38
rs145834196 Chr.1: 155717035-155717035 on Build GRCh38
rs1024952406 Chr.1: 155716683-155716683 on Build GRCh38
rs747817733 Chr.1: 155716978-155716978 on Build GRCh38
rs544570828 Chr.1: 155716837-155716837 on Build GRCh38
rs752189525 Chr.1: 155717151-155717151 on Build GRCh38
rs936844895 Chr.1: 155716870-155716870 on Build GRCh38
rs770793096 Chr.1: 155717018-155717018 on Build GRCh38
rs1013730444 Chr.1: 155716882-155716882 on Build GRCh38
rs963240267 Chr.1: 155716763-155716763 on Build GRCh38
rs541967448 Chr.1: 155716910-155716910 on Build GRCh38
rs773518709 Chr.1: 155716930-155716931 on Build GRCh38
rs969814286 Chr.1: 155716709-155716709 on Build GRCh38
rs574584490 Chr.1: 155716879-155716879 on Build GRCh38
rs921350699 Chr.1: 155716804-155716804 on Build GRCh38
rs749012692 Chr.1: 155717087-155717087 on Build GRCh38
rs373262328 Chr.1: 155716816-155716816 on Build GRCh38
rs1042652765 Chr.1: 155716848-155716848 on Build GRCh38
rs951098964 Chr.1: 155716808-155716808 on Build GRCh38
rs925547082 Chr.1: 155717078-155717078 on Build GRCh38
rs1055187714 Chr.1: 155716873-155716873 on Build GRCh38
rs936896915 Chr.1: 155717097-155717097 on Build GRCh38
rs751049730 Chr.1: 155716972-155716972 on Build GRCh38
rs766962516 Chr.1: 155717147-155717147 on Build GRCh38
rs763561074 Chr.1: 155717055-155717055 on Build GRCh38
rs201385663 Chr.1: 155717127-155717127 on Build GRCh38
rs773565559 Chr.1: 155717111-155717111 on Build GRCh38
rs753321958 Chr.1: 155717140-155717140 on Build GRCh38
rs532334569 Chr.1: 155717152-155717152 on Build GRCh38
rs373060703 Chr.1: 155716976-155716976 on Build GRCh38
rs1019811247 Chr.1: 155717061-155717061 on Build GRCh38
rs1052571858 Chr.1: 155716850-155716850 on Build GRCh38
rs1039943352 Chr.1: 155716820-155716820 on Build GRCh38
rs973914187 Chr.1: 155716741-155716741 on Build GRCh38
rs942383493 Chr.1: 155716819-155716819 on Build GRCh38
rs112796215 Chr.1: 155717083-155717083 on Build GRCh38
rs763690503 Chr.1: 155717045-155717052 on Build GRCh38
rs377463084 Chr.1: 155717011-155717011 on Build GRCh38
rs898364585 Chr.1: 155717008-155717008 on Build GRCh38
rs750906763 Chr.1: 155716833-155716833 on Build GRCh38
rs755677116 Chr.1: 155717075-155717075 on Build GRCh38
rs768657534 Chr.1: 155717095-155717095 on Build GRCh38
rs752327968 Chr.1: 155717077-155717077 on Build GRCh38
rs370992701 Chr.1: 155717051-155717051 on Build GRCh38
rs779724693 Chr.1: 155716965-155716965 on Build GRCh38
rs149919712 Chr.1: 155717109-155717109 on Build GRCh38
rs757038621 Chr.1: 155717092-155717092 on Build GRCh38
rs377474444 Chr.1: 155717108-155717108 on Build GRCh38
rs142107923 Chr.1: 155716878-155716878 on Build GRCh38
rs892652497 Chr.1: 155716867-155716867 on Build GRCh38
rs1001442390 Chr.1: 155716716-155716716 on Build GRCh38
rs777508861 Chr.1: 155717004-155717004 on Build GRCh38
rs377724521 Chr.1: 155716949-155716949 on Build GRCh38
rs560216710 Chr.1: 155717028-155717028 on Build GRCh38
rs112343005 Chr.1: 155716822-155716822 on Build GRCh38
rs146211768 Chr.1: 155717076-155717076 on Build GRCh38

View Full Product Details