Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127314579-127315125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC1
Entrez Gene ID
55862
Gene Name
ethylmalonyl-CoA decarboxylase 1
Gene Aliases
HEL-S-76, MMCD, dJ351K20.2
UniGene
Hs.486410
Gene Chromosome Location
Chr.6: 127288712-127343609 on Build GRCh38
Transcripts
NM_001002030.1 NM_001105544.1 NM_001105545.1 NM_001139510.1 NM_018479.3 XM_005267047.2 XM_005267048.1 XM_005267049.2 XM_005267050.2 XM_011535942.2 XM_011535943.2 XM_017011056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373247 c.513C>G p.F171L Chr.6: 127314818-127314818 on Build GRCh38
1072979 c.456G>A p.A152A Chr.6: 127314875-127314875 on Build GRCh38
1072980 c.385G>A p.A129T Chr.6: 127314875-127314875 on Build GRCh38
5486516 c.452T>C p.V151A Chr.6: 127314879-127314879 on Build GRCh38
5486517 c.381T>C p.C127C Chr.6: 127314879-127314879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940362527 Chr.6: 127315027-127315027 on Build GRCh38
rs547307421 Chr.6: 127314961-127314961 on Build GRCh38
rs776540701 Chr.6: 127314963-127314963 on Build GRCh38
rs767323362 Chr.6: 127314966-127314968 on Build GRCh38
rs747770040 Chr.6: 127314795-127314795 on Build GRCh38
rs367829930 Chr.6: 127315004-127315004 on Build GRCh38
rs777898254 Chr.6: 127314891-127314891 on Build GRCh38
rs749270818 Chr.6: 127314984-127314984 on Build GRCh38
rs761934091 Chr.6: 127314862-127314862 on Build GRCh38
rs780327409 Chr.6: 127314882-127314882 on Build GRCh38
rs776336868 Chr.6: 127314745-127314745 on Build GRCh38
rs776827775 Chr.6: 127314848-127314848 on Build GRCh38
rs746287294 Chr.6: 127314815-127314815 on Build GRCh38
rs777251619 Chr.6: 127314773-127314776 on Build GRCh38
rs1056028826 Chr.6: 127314732-127314732 on Build GRCh38
rs112248003 Chr.6: 127314813-127314813 on Build GRCh38
rs780925208 Chr.6: 127314768-127314768 on Build GRCh38
rs948785961 Chr.6: 127314679-127314679 on Build GRCh38
rs914594403 Chr.6: 127314653-127314653 on Build GRCh38
rs143031566 Chr.6: 127314831-127314831 on Build GRCh38
rs777534740 Chr.6: 127314925-127314925 on Build GRCh38
rs754963257 Chr.6: 127315052-127315052 on Build GRCh38
rs776003051 Chr.6: 127314827-127314827 on Build GRCh38
rs186749074 Chr.6: 127314928-127314928 on Build GRCh38
rs878889407 Chr.6: 127315090-127315090 on Build GRCh38
rs866592396 Chr.6: 127314909-127314909 on Build GRCh38
rs563645893 Chr.6: 127314766-127314766 on Build GRCh38
rs369602393 Chr.6: 127314875-127314875 on Build GRCh38
rs376600385 Chr.6: 127314899-127314899 on Build GRCh38
rs111828900 Chr.6: 127315002-127315002 on Build GRCh38
rs367812403 Chr.6: 127314818-127314818 on Build GRCh38
rs955159970 Chr.6: 127314924-127314924 on Build GRCh38
rs970086207 Chr.6: 127315037-127315037 on Build GRCh38
rs146146402 Chr.6: 127314876-127314876 on Build GRCh38
rs138841072 Chr.6: 127315075-127315075 on Build GRCh38
rs764167963 Chr.6: 127314846-127314846 on Build GRCh38
rs755694377 Chr.6: 127314904-127314904 on Build GRCh38
rs762001020 Chr.6: 127314965-127314965 on Build GRCh38
rs535637553 Chr.6: 127314931-127314931 on Build GRCh38
rs751460672 Chr.6: 127315013-127315013 on Build GRCh38
rs769549965 Chr.6: 127314796-127314796 on Build GRCh38
rs371154778 Chr.6: 127314721-127314721 on Build GRCh38
rs772427543 Chr.6: 127314933-127314933 on Build GRCh38
rs1044493300 Chr.6: 127314691-127314691 on Build GRCh38
rs762770431 Chr.6: 127314977-127314977 on Build GRCh38
rs763131683 Chr.6: 127314805-127314805 on Build GRCh38
rs776027372 Chr.6: 127314909-127314910 on Build GRCh38
rs747498203 Chr.6: 127314959-127314959 on Build GRCh38
rs368649472 Chr.6: 127314803-127314803 on Build GRCh38
rs765328292 Chr.6: 127314966-127314966 on Build GRCh38
rs938827400 Chr.6: 127314738-127314738 on Build GRCh38
rs763358235 Chr.6: 127314910-127314910 on Build GRCh38
rs755132805 Chr.6: 127315025-127315025 on Build GRCh38
rs775913420 Chr.6: 127314947-127314947 on Build GRCh38
rs906909032 Chr.6: 127314717-127314717 on Build GRCh38
rs983005936 Chr.6: 127315005-127315005 on Build GRCh38
rs773513620 Chr.6: 127314972-127314972 on Build GRCh38
rs765535043 Chr.6: 127314863-127314863 on Build GRCh38
rs759972555 Chr.6: 127314793-127314796 on Build GRCh38
rs1028071902 Chr.6: 127314906-127314906 on Build GRCh38
rs964466333 Chr.6: 127314960-127314960 on Build GRCh38
rs780555626 Chr.6: 127314886-127314886 on Build GRCh38

View Full Product Details