Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127589994-127590540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf58
Entrez Gene ID
352999
Gene Name
chromosome 6 open reading frame 58
Gene Aliases
LEG1
UniGene
Hs.226268
Gene Chromosome Location
Chr.6: 127577174-127591818 on Build GRCh38
Transcripts
NM_001010905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3858100 c.715A>C p.S239R Chr.6: 127590127-127590127 on Build GRCh38
6220471 c.830delC p.P278fs*12 Chr.6: 127590242-127590242 on Build GRCh38
5639830 c.701C>A p.A234E Chr.6: 127590113-127590113 on Build GRCh38
1697885 c.748G>A p.V250I Chr.6: 127590160-127590160 on Build GRCh38
3172051 c.797T>G p.M266R Chr.6: 127590209-127590209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201775588 Chr.6: 127590206-127590206 on Build GRCh38
rs746148834 Chr.6: 127590364-127590364 on Build GRCh38
rs776628630 Chr.6: 127590378-127590378 on Build GRCh38
rs780579826 Chr.6: 127590113-127590113 on Build GRCh38
rs527828175 Chr.6: 127590217-127590217 on Build GRCh38
rs777664922 Chr.6: 127590318-127590318 on Build GRCh38
rs567496932 Chr.6: 127590365-127590365 on Build GRCh38
rs766216541 Chr.6: 127590072-127590072 on Build GRCh38
rs1013369811 Chr.6: 127590296-127590296 on Build GRCh38
rs866353236 Chr.6: 127590205-127590205 on Build GRCh38
rs988055471 Chr.6: 127590442-127590442 on Build GRCh38
rs756672403 Chr.6: 127590317-127590317 on Build GRCh38
rs774161660 Chr.6: 127590158-127590158 on Build GRCh38
rs146910440 Chr.6: 127590308-127590308 on Build GRCh38
rs750143731 Chr.6: 127590283-127590283 on Build GRCh38
rs187501399 Chr.6: 127590112-127590112 on Build GRCh38
rs753374472 Chr.6: 127590186-127590186 on Build GRCh38
rs143015566 Chr.6: 127590114-127590114 on Build GRCh38
rs933136989 Chr.6: 127590489-127590489 on Build GRCh38
rs761511783 Chr.6: 127590277-127590277 on Build GRCh38
rs759671633 Chr.6: 127590500-127590500 on Build GRCh38
rs749348166 Chr.6: 127590321-127590321 on Build GRCh38
rs760676764 Chr.6: 127590264-127590264 on Build GRCh38
rs749468767 Chr.6: 127590218-127590218 on Build GRCh38
rs746881318 Chr.6: 127590369-127590369 on Build GRCh38
rs762649107 Chr.6: 127590058-127590058 on Build GRCh38
rs755378167 Chr.6: 127590290-127590290 on Build GRCh38
rs771163070 Chr.6: 127590232-127590232 on Build GRCh38
rs945448233 Chr.6: 127590313-127590313 on Build GRCh38
rs561575563 Chr.6: 127590046-127590046 on Build GRCh38
rs145162146 Chr.6: 127590263-127590263 on Build GRCh38
rs150023916 Chr.6: 127590163-127590163 on Build GRCh38
rs1025129588 Chr.6: 127590305-127590305 on Build GRCh38
rs771967415 Chr.6: 127590254-127590254 on Build GRCh38
rs200268132 Chr.6: 127590242-127590242 on Build GRCh38
rs774604569 Chr.6: 127590233-127590233 on Build GRCh38
rs553308278 Chr.6: 127590503-127590503 on Build GRCh38
rs770643503 Chr.6: 127590154-127590154 on Build GRCh38
rs987596970 Chr.6: 127590494-127590494 on Build GRCh38
rs779102164 Chr.6: 127590359-127590359 on Build GRCh38
rs751436936 Chr.6: 127590073-127590073 on Build GRCh38
rs532390887 Chr.6: 127590198-127590198 on Build GRCh38
rs149209214 Chr.6: 127590307-127590307 on Build GRCh38
rs1024023175 Chr.6: 127590191-127590191 on Build GRCh38
rs764701299 Chr.6: 127590287-127590288 on Build GRCh38
rs773290440 Chr.6: 127590144-127590144 on Build GRCh38
rs957903478 Chr.6: 127590419-127590419 on Build GRCh38
rs534479433 Chr.6: 127590484-127590484 on Build GRCh38
rs763172302 Chr.6: 127590145-127590145 on Build GRCh38
rs904028463 Chr.6: 127590346-127590346 on Build GRCh38
rs374323952 Chr.6: 127590273-127590273 on Build GRCh38
rs1014313891 Chr.6: 127590417-127590417 on Build GRCh38
rs762919219 Chr.6: 127590252-127590252 on Build GRCh38
rs760101103 Chr.6: 127590173-127590173 on Build GRCh38
rs775359964 Chr.6: 127590366-127590366 on Build GRCh38
rs777258595 Chr.6: 127590097-127590097 on Build GRCh38
rs753594986 Chr.6: 127590103-127590103 on Build GRCh38
rs781523347 Chr.6: 127590195-127590195 on Build GRCh38
rs1052235234 Chr.6: 127590064-127590064 on Build GRCh38
rs767418778 Chr.6: 127590168-127590168 on Build GRCh38
rs764965462 Chr.6: 127590281-127590281 on Build GRCh38
rs752170519 Chr.6: 127590090-127590090 on Build GRCh38
rs370609859 Chr.6: 127590177-127590177 on Build GRCh38
rs756975963 Chr.6: 127590187-127590187 on Build GRCh38
rs753420094 Chr.6: 127590276-127590276 on Build GRCh38
rs781287818 Chr.6: 127590118-127590118 on Build GRCh38
rs748744284 Chr.6: 127590204-127590204 on Build GRCh38
rs374948798 Chr.6: 127590074-127590074 on Build GRCh38
rs78854930 Chr.6: 127590143-127590143 on Build GRCh38
rs755445575 Chr.6: 127590193-127590193 on Build GRCh38
rs202206885 Chr.6: 127590192-127590192 on Build GRCh38
rs913771863 Chr.6: 127590463-127590463 on Build GRCh38
rs139119636 Chr.6: 127590133-127590133 on Build GRCh38
rs1034059647 Chr.6: 127590142-127590142 on Build GRCh38
rs755697977 Chr.6: 127590092-127590092 on Build GRCh38
rs752718950 Chr.6: 127590169-127590169 on Build GRCh38
rs372856580 Chr.6: 127590333-127590333 on Build GRCh38
rs944663519 Chr.6: 127590289-127590289 on Build GRCh38
rs768664987 Chr.6: 127590376-127590376 on Build GRCh38
rs894144182 Chr.6: 127590403-127590403 on Build GRCh38
rs748158523 Chr.6: 127590128-127590128 on Build GRCh38
rs759294285 Chr.6: 127590241-127590241 on Build GRCh38
rs753845400 Chr.6: 127590194-127590194 on Build GRCh38
rs780530426 Chr.6: 127590189-127590189 on Build GRCh38

View Full Product Details