Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 129599077-129599621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP18
Entrez Gene ID
93663
Gene Name
Rho GTPase activating protein 18
Gene Aliases
MacGAP, SENEX, bA307O14.2
UniGene
Hs.486458
Gene Chromosome Location
Chr.6: 129576145-129710225 on Build GRCh38
Transcripts
NM_033515.2 XM_005267212.2 XM_005267213.1 XM_017011494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357934 c.1713+1G>C p.? Chr.6: 129599215-129599215 on Build GRCh38
362985 c.1693C>T p.Q565* Chr.6: 129599236-129599236 on Build GRCh38
3374042 c.1577C>T p.P526L Chr.6: 129599352-129599352 on Build GRCh38
5898291 c.1676G>A p.R559Q Chr.6: 129599253-129599253 on Build GRCh38
5749717 c.1584T>G p.F528L Chr.6: 129599345-129599345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973339175 Chr.6: 129599561-129599561 on Build GRCh38
rs373864720 Chr.6: 129599416-129599416 on Build GRCh38
rs568607023 Chr.6: 129599183-129599183 on Build GRCh38
rs565399225 Chr.6: 129599254-129599254 on Build GRCh38
rs767450745 Chr.6: 129599207-129599207 on Build GRCh38
rs771211647 Chr.6: 129599292-129599292 on Build GRCh38
rs778349052 Chr.6: 129599392-129599392 on Build GRCh38
rs201000981 Chr.6: 129599221-129599221 on Build GRCh38
rs928373661 Chr.6: 129599587-129599587 on Build GRCh38
rs759358228 Chr.6: 129599310-129599310 on Build GRCh38
rs569544030 Chr.6: 129599376-129599376 on Build GRCh38
rs748645425 Chr.6: 129599272-129599272 on Build GRCh38
rs755626500 Chr.6: 129599215-129599215 on Build GRCh38
rs756960625 Chr.6: 129599231-129599231 on Build GRCh38
rs755034128 Chr.6: 129599562-129599562 on Build GRCh38
rs755014075 Chr.6: 129599166-129599166 on Build GRCh38
rs974514044 Chr.6: 129599136-129599136 on Build GRCh38
rs541194328 Chr.6: 129599488-129599488 on Build GRCh38
rs748679047 Chr.6: 129599381-129599381 on Build GRCh38
rs747303231 Chr.6: 129599246-129599246 on Build GRCh38
rs1018993074 Chr.6: 129599415-129599415 on Build GRCh38
rs750094615 Chr.6: 129599369-129599369 on Build GRCh38
rs774745215 Chr.6: 129599293-129599293 on Build GRCh38
rs553214142 Chr.6: 129599584-129599584 on Build GRCh38
rs746131257 Chr.6: 129599400-129599400 on Build GRCh38
rs368639406 Chr.6: 129599360-129599360 on Build GRCh38
rs1032793581 Chr.6: 129599494-129599494 on Build GRCh38
rs756523513 Chr.6: 129599382-129599382 on Build GRCh38
rs186649882 Chr.6: 129599396-129599396 on Build GRCh38
rs538371710 Chr.6: 129599199-129599199 on Build GRCh38
rs769915140 Chr.6: 129599279-129599279 on Build GRCh38
rs769817264 Chr.6: 129599192-129599192 on Build GRCh38
rs781686114 Chr.6: 129599265-129599265 on Build GRCh38
rs753283561 Chr.6: 129599338-129599338 on Build GRCh38
rs781289568 Chr.6: 129599568-129599568 on Build GRCh38
rs1043972016 Chr.6: 129599404-129599404 on Build GRCh38
rs1028912595 Chr.6: 129599142-129599142 on Build GRCh38
rs777559929 Chr.6: 129599153-129599154 on Build GRCh38
rs965884850 Chr.6: 129599134-129599134 on Build GRCh38
rs918151398 Chr.6: 129599524-129599524 on Build GRCh38
rs550118312 Chr.6: 129599157-129599157 on Build GRCh38
rs949492655 Chr.6: 129599146-129599146 on Build GRCh38
rs767671391 Chr.6: 129599228-129599228 on Build GRCh38
rs377167149 Chr.6: 129599364-129599364 on Build GRCh38
rs182167388 Chr.6: 129599200-129599200 on Build GRCh38
rs931064034 Chr.6: 129599214-129599214 on Build GRCh38
rs74353230 Chr.6: 129599306-129599306 on Build GRCh38
rs145578474 Chr.6: 129599154-129599176 on Build GRCh38
rs759409586 Chr.6: 129599202-129599202 on Build GRCh38
rs6934315 Chr.6: 129599399-129599399 on Build GRCh38
rs771895448 Chr.6: 129599403-129599403 on Build GRCh38
rs769089837 Chr.6: 129599172-129599172 on Build GRCh38
rs902536521 Chr.6: 129599414-129599414 on Build GRCh38
rs763645667 Chr.6: 129599319-129599319 on Build GRCh38
rs889456708 Chr.6: 129599313-129599313 on Build GRCh38
rs918098053 Chr.6: 129599103-129599103 on Build GRCh38
rs752599904 Chr.6: 129599213-129599213 on Build GRCh38
rs931094464 Chr.6: 129599565-129599565 on Build GRCh38
rs79303022 Chr.6: 129599424-129599424 on Build GRCh38
rs75939530 Chr.6: 129599426-129599426 on Build GRCh38
rs940348303 Chr.6: 129599253-129599253 on Build GRCh38
rs773454493 Chr.6: 129599195-129599195 on Build GRCh38
rs148650117 Chr.6: 129599315-129599315 on Build GRCh38
rs753484384 Chr.6: 129599222-129599222 on Build GRCh38
rs775464255 Chr.6: 129599316-129599316 on Build GRCh38
rs748385995 Chr.6: 129599582-129599582 on Build GRCh38
rs991072918 Chr.6: 129599523-129599523 on Build GRCh38
rs1029214070 Chr.6: 129599471-129599471 on Build GRCh38
rs755341797 Chr.6: 129599372-129599372 on Build GRCh38
rs1020104630 Chr.6: 129599421-129599421 on Build GRCh38
rs949545219 Chr.6: 129599542-129599542 on Build GRCh38
rs773226477 Chr.6: 129599283-129599283 on Build GRCh38
rs959327010 Chr.6: 129599518-129599518 on Build GRCh38
rs1048134634 Chr.6: 129599578-129599578 on Build GRCh38
rs749396735 Chr.6: 129599288-129599288 on Build GRCh38
rs777007053 Chr.6: 129599177-129599177 on Build GRCh38
rs144231632 Chr.6: 129599431-129599431 on Build GRCh38
rs778546389 Chr.6: 129599243-129599243 on Build GRCh38
rs1048557931 Chr.6: 129599287-129599287 on Build GRCh38
rs143946112 Chr.6: 129599255-129599255 on Build GRCh38
rs764302231 Chr.6: 129599415-129599416 on Build GRCh38
rs952031603 Chr.6: 129599144-129599144 on Build GRCh38

View Full Product Details