Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 130835553-130836055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L2
Entrez Gene ID
2037
Gene Name
erythrocyte membrane protein band 4.1 like 2
Gene Aliases
4.1-G, 4.1G
UniGene
Hs.486470
Gene Chromosome Location
Chr.6: 130829111-131063322 on Build GRCh38
Transcripts
XM_017010353.1 NM_001195597.1

Gene Symbol
SMLR1
Entrez Gene ID
100507203
Gene Name
small leucine rich protein 1
Gene Aliases
-
UniGene
Hs.388715
Gene Chromosome Location
Chr.6: 130827405-130837135 on Build GRCh38
Transcripts
XM_017010353.1 NM_001195597.1

SNP Detail

SNP ID Chromosome Location
rs150397654 Chr.6: 130835860-130835860 on Build GRCh38
rs549904093 Chr.6: 130835691-130835691 on Build GRCh38
rs1001850987 Chr.6: 130835740-130835740 on Build GRCh38
rs543969352 Chr.6: 130835817-130835817 on Build GRCh38
rs1041249665 Chr.6: 130835609-130835609 on Build GRCh38
rs1053649370 Chr.6: 130835773-130835773 on Build GRCh38
rs1003551820 Chr.6: 130835835-130835835 on Build GRCh38
rs78432948 Chr.6: 130835701-130835701 on Build GRCh38
rs964293959 Chr.6: 130835713-130835713 on Build GRCh38
rs138215708 Chr.6: 130835741-130835741 on Build GRCh38
rs562484648 Chr.6: 130835819-130835819 on Build GRCh38
rs114120155 Chr.6: 130835761-130835761 on Build GRCh38
rs753322158 Chr.6: 130835969-130835969 on Build GRCh38
rs975649586 Chr.6: 130835717-130835717 on Build GRCh38
rs905507444 Chr.6: 130836002-130836003 on Build GRCh38
rs533207915 Chr.6: 130835963-130835963 on Build GRCh38
rs1001487008 Chr.6: 130836020-130836021 on Build GRCh38
rs1005760033 Chr.6: 130835659-130835659 on Build GRCh38
rs1045118953 Chr.6: 130835602-130835602 on Build GRCh38
rs111628117 Chr.6: 130835879-130835879 on Build GRCh38
rs1047238117 Chr.6: 130835959-130835959 on Build GRCh38
rs1003519332 Chr.6: 130835614-130835614 on Build GRCh38
rs576354171 Chr.6: 130835782-130835782 on Build GRCh38
rs146997363 Chr.6: 130835729-130835729 on Build GRCh38
rs559396924 Chr.6: 130835924-130835924 on Build GRCh38
rs911212730 Chr.6: 130835834-130835834 on Build GRCh38
rs922834164 Chr.6: 130835728-130835728 on Build GRCh38
rs893762370 Chr.6: 130835586-130835586 on Build GRCh38
rs548657222 Chr.6: 130835625-130835625 on Build GRCh38
rs1029878334 Chr.6: 130835990-130835990 on Build GRCh38
rs113424909 Chr.6: 130835596-130835596 on Build GRCh38
rs750635345 Chr.6: 130835743-130835743 on Build GRCh38
rs1041676977 Chr.6: 130835895-130835895 on Build GRCh38
rs558083970 Chr.6: 130835762-130835762 on Build GRCh38
rs942910759 Chr.6: 130835606-130835606 on Build GRCh38
rs951539778 Chr.6: 130835658-130835658 on Build GRCh38
rs1012151922 Chr.6: 130835600-130835600 on Build GRCh38
rs1005941334 Chr.6: 130835986-130835986 on Build GRCh38
rs913338154 Chr.6: 130835788-130835788 on Build GRCh38
rs887224618 Chr.6: 130835937-130835937 on Build GRCh38
rs770654788 Chr.6: 130835962-130835962 on Build GRCh38
rs761567493 Chr.6: 130835670-130835670 on Build GRCh38
rs1015898696 Chr.6: 130835710-130835710 on Build GRCh38
rs997301417 Chr.6: 130835648-130835648 on Build GRCh38
rs978737602 Chr.6: 130835832-130835832 on Build GRCh38
rs955614789 Chr.6: 130836011-130836011 on Build GRCh38
rs900029471 Chr.6: 130835965-130835965 on Build GRCh38
rs930151237 Chr.6: 130835949-130835949 on Build GRCh38
rs779072386 Chr.6: 130835932-130835942 on Build GRCh38
rs141400986 Chr.6: 130835828-130835828 on Build GRCh38
rs190664462 Chr.6: 130835603-130835603 on Build GRCh38
rs551384274 Chr.6: 130835989-130835989 on Build GRCh38
rs922826241 Chr.6: 130835939-130835939 on Build GRCh38
rs766536559 Chr.6: 130835827-130835827 on Build GRCh38
rs751605660 Chr.6: 130835943-130835943 on Build GRCh38
rs764045236 Chr.6: 130835583-130835583 on Build GRCh38
rs117891622 Chr.6: 130835730-130835730 on Build GRCh38

View Full Product Details