Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131854804-131855316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP1
Entrez Gene ID
5167
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 1
Gene Aliases
ARHR2, COLED, M6S1, NPP1, NPPS, PC-1, PCA1, PDNP1
UniGene
Hs.527295
Gene Chromosome Location
Chr.6: 131808016-131895155 on Build GRCh38
Transcripts
NM_006208.2

SNP Detail

SNP ID Chromosome Location
rs562521329 Chr.6: 131854873-131854873 on Build GRCh38
rs912963404 Chr.6: 131855265-131855265 on Build GRCh38
rs571853912 Chr.6: 131855150-131855150 on Build GRCh38
rs771842086 Chr.6: 131854896-131854896 on Build GRCh38
rs760402307 Chr.6: 131855104-131855104 on Build GRCh38
rs550495051 Chr.6: 131855142-131855142 on Build GRCh38
rs778559374 Chr.6: 131854888-131854888 on Build GRCh38
rs761774560 Chr.6: 131855223-131855223 on Build GRCh38
rs945720765 Chr.6: 131855106-131855106 on Build GRCh38
rs760744593 Chr.6: 131854900-131854901 on Build GRCh38
rs976612094 Chr.6: 131855252-131855252 on Build GRCh38
rs1036378173 Chr.6: 131855179-131855179 on Build GRCh38
rs1014261171 Chr.6: 131854843-131854843 on Build GRCh38
rs1018206109 Chr.6: 131855236-131855236 on Build GRCh38
rs753181538 Chr.6: 131854944-131854944 on Build GRCh38
rs187764662 Chr.6: 131854934-131854934 on Build GRCh38
rs73539675 Chr.6: 131855169-131855169 on Build GRCh38
rs561587786 Chr.6: 131854910-131854910 on Build GRCh38
rs895950310 Chr.6: 131855151-131855151 on Build GRCh38
rs746466658 Chr.6: 131854900-131854900 on Build GRCh38
rs750147184 Chr.6: 131855049-131855049 on Build GRCh38
rs758385512 Chr.6: 131855052-131855052 on Build GRCh38
rs766216956 Chr.6: 131854931-131854931 on Build GRCh38
rs771145573 Chr.6: 131854930-131854930 on Build GRCh38
rs773321237 Chr.6: 131854891-131854891 on Build GRCh38
rs147798392 Chr.6: 131854993-131854993 on Build GRCh38
rs1055981613 Chr.6: 131855147-131855147 on Build GRCh38
rs111465897 Chr.6: 131855055-131855055 on Build GRCh38
rs918390857 Chr.6: 131855166-131855166 on Build GRCh38
rs772818357 Chr.6: 131854901-131854901 on Build GRCh38
rs1044333603 Chr.6: 131855199-131855199 on Build GRCh38
rs112683816 Chr.6: 131854838-131854838 on Build GRCh38
rs200074342 Chr.6: 131854894-131854894 on Build GRCh38
rs926224112 Chr.6: 131855114-131855114 on Build GRCh38
rs761550283 Chr.6: 131854911-131854911 on Build GRCh38
rs779649521 Chr.6: 131854897-131854897 on Build GRCh38
rs994827234 Chr.6: 131855210-131855210 on Build GRCh38
rs1048774676 Chr.6: 131855180-131855180 on Build GRCh38
rs1036795045 Chr.6: 131855162-131855162 on Build GRCh38
rs1028106776 Chr.6: 131855221-131855221 on Build GRCh38
rs778778246 Chr.6: 131855041-131855041 on Build GRCh38
rs1014966696 Chr.6: 131854855-131854855 on Build GRCh38
rs948718718 Chr.6: 131855185-131855185 on Build GRCh38
rs770252632 Chr.6: 131854901-131854903 on Build GRCh38
rs962049279 Chr.6: 131854862-131854862 on Build GRCh38
rs764155541 Chr.6: 131854972-131854972 on Build GRCh38
rs143637835 Chr.6: 131854906-131854906 on Build GRCh38
rs1032224789 Chr.6: 131855013-131855013 on Build GRCh38
rs763862810 Chr.6: 131855110-131855110 on Build GRCh38
rs774521656 Chr.6: 131854923-131854923 on Build GRCh38
rs965183358 Chr.6: 131855237-131855237 on Build GRCh38
rs760786509 Chr.6: 131854955-131854955 on Build GRCh38
rs895096212 Chr.6: 131855183-131855183 on Build GRCh38
rs897931051 Chr.6: 131855205-131855205 on Build GRCh38
rs990785510 Chr.6: 131855103-131855103 on Build GRCh38
rs1002914068 Chr.6: 131854938-131854938 on Build GRCh38
rs374026835 Chr.6: 131855072-131855072 on Build GRCh38
rs768256071 Chr.6: 131854901-131854901 on Build GRCh38
rs551058857 Chr.6: 131855225-131855225 on Build GRCh38
rs985285692 Chr.6: 131854913-131854913 on Build GRCh38
rs774561788 Chr.6: 131854935-131854935 on Build GRCh38
rs771384565 Chr.6: 131854922-131854922 on Build GRCh38

View Full Product Details