Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131950679-131951174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTGF
Entrez Gene ID
1490
Gene Name
connective tissue growth factor
Gene Aliases
CCN2, HCS24, IGFBP8, NOV2
UniGene
Hs.410037
Gene Chromosome Location
Chr.6: 131948176-131951378 on Build GRCh38
Transcripts
NM_001901.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296317 c.226C>A p.P76T Chr.6: 131950833-131950833 on Build GRCh38
4631815 c.168C>T p.C56C Chr.6: 131950891-131950891 on Build GRCh38
1620920 c.30C>T p.R10R Chr.6: 131951143-131951143 on Build GRCh38
3749947 c.240A>C p.L80L Chr.6: 131950819-131950819 on Build GRCh38
3749946 c.247C>G p.H83D Chr.6: 131950812-131950812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952920158 Chr.6: 131951016-131951016 on Build GRCh38
rs985683514 Chr.6: 131951034-131951034 on Build GRCh38
rs778832399 Chr.6: 131950832-131950832 on Build GRCh38
rs753677977 Chr.6: 131950827-131950827 on Build GRCh38
rs1038500057 Chr.6: 131951098-131951098 on Build GRCh38
rs771548344 Chr.6: 131950807-131950807 on Build GRCh38
rs750290405 Chr.6: 131950833-131950833 on Build GRCh38
rs771666259 Chr.6: 131950747-131950747 on Build GRCh38
rs773619523 Chr.6: 131950767-131950767 on Build GRCh38
rs936577744 Chr.6: 131951115-131951115 on Build GRCh38
rs964622495 Chr.6: 131951043-131951043 on Build GRCh38
rs563027002 Chr.6: 131950958-131950958 on Build GRCh38
rs1054975818 Chr.6: 131951120-131951120 on Build GRCh38
rs777585673 Chr.6: 131950804-131950804 on Build GRCh38
rs577763812 Chr.6: 131950903-131950903 on Build GRCh38
rs764203876 Chr.6: 131950826-131950826 on Build GRCh38
rs978719302 Chr.6: 131951071-131951071 on Build GRCh38
rs752906843 Chr.6: 131950802-131950802 on Build GRCh38
rs775691003 Chr.6: 131950909-131950909 on Build GRCh38
rs747737409 Chr.6: 131951130-131951130 on Build GRCh38
rs757046328 Chr.6: 131950831-131950831 on Build GRCh38
rs529572329 Chr.6: 131951054-131951054 on Build GRCh38
rs777587233 Chr.6: 131950733-131950733 on Build GRCh38
rs745364919 Chr.6: 131950745-131950745 on Build GRCh38
rs781006644 Chr.6: 131951128-131951128 on Build GRCh38
rs537427255 Chr.6: 131950730-131950730 on Build GRCh38
rs940226832 Chr.6: 131951087-131951087 on Build GRCh38
rs768461734 Chr.6: 131950770-131950771 on Build GRCh38
rs925175156 Chr.6: 131950715-131950715 on Build GRCh38
rs767569105 Chr.6: 131950821-131950821 on Build GRCh38
rs934031221 Chr.6: 131950823-131950823 on Build GRCh38
rs1038619364 Chr.6: 131950949-131950949 on Build GRCh38
rs762093685 Chr.6: 131950763-131950763 on Build GRCh38
rs746630581 Chr.6: 131950810-131950810 on Build GRCh38
rs749165106 Chr.6: 131950748-131950749 on Build GRCh38
rs113144034 Chr.6: 131951065-131951065 on Build GRCh38
rs977202318 Chr.6: 131951048-131951048 on Build GRCh38
rs961009845 Chr.6: 131950952-131950952 on Build GRCh38
rs758408030 Chr.6: 131951010-131951010 on Build GRCh38
rs1018849685 Chr.6: 131951045-131951045 on Build GRCh38
rs530485396 Chr.6: 131950979-131950979 on Build GRCh38
rs13212409 Chr.6: 131950935-131950935 on Build GRCh38
rs771364910 Chr.6: 131950860-131950860 on Build GRCh38
rs775004398 Chr.6: 131950873-131950873 on Build GRCh38
rs966966564 Chr.6: 131951018-131951018 on Build GRCh38
rs749248801 Chr.6: 131950805-131950805 on Build GRCh38
rs7451102 Chr.6: 131950812-131950812 on Build GRCh38
rs767059535 Chr.6: 131950817-131950817 on Build GRCh38
rs763378768 Chr.6: 131950816-131950816 on Build GRCh38
rs375793270 Chr.6: 131950969-131950969 on Build GRCh38
rs1020802955 Chr.6: 131951017-131951017 on Build GRCh38
rs965497939 Chr.6: 131951046-131951046 on Build GRCh38
rs548061360 Chr.6: 131951062-131951062 on Build GRCh38
rs6934749 Chr.6: 131950840-131950840 on Build GRCh38
rs751147206 Chr.6: 131951102-131951102 on Build GRCh38
rs868791465 Chr.6: 131950708-131950708 on Build GRCh38
rs979249373 Chr.6: 131951029-131951029 on Build GRCh38
rs12206231 Chr.6: 131950819-131950819 on Build GRCh38
rs749727188 Chr.6: 131950850-131950850 on Build GRCh38
rs560896250 Chr.6: 131950996-131950996 on Build GRCh38
rs766969428 Chr.6: 131950792-131950792 on Build GRCh38
rs952830020 Chr.6: 131950712-131950712 on Build GRCh38
rs369981825 Chr.6: 131950760-131950760 on Build GRCh38
rs372597924 Chr.6: 131950770-131950770 on Build GRCh38
rs887245529 Chr.6: 131950892-131950892 on Build GRCh38
rs894929214 Chr.6: 131950929-131950929 on Build GRCh38
rs376929267 Chr.6: 131950765-131950765 on Build GRCh38
rs866471933 Chr.6: 131951111-131951111 on Build GRCh38
rs767690353 Chr.6: 131950799-131950799 on Build GRCh38
rs781444667 Chr.6: 131950719-131950719 on Build GRCh38
rs775008083 Chr.6: 131950809-131950809 on Build GRCh38
rs778795621 Chr.6: 131950806-131950806 on Build GRCh38
rs867106833 Chr.6: 131950790-131950790 on Build GRCh38
rs763503956 Chr.6: 131950786-131950786 on Build GRCh38
rs1013718997 Chr.6: 131950947-131950947 on Build GRCh38
rs757875277 Chr.6: 131950836-131950836 on Build GRCh38
rs934105560 Chr.6: 131951060-131951060 on Build GRCh38
rs752896023 Chr.6: 131950824-131950824 on Build GRCh38
rs944116755 Chr.6: 131950791-131950791 on Build GRCh38
rs569804142 Chr.6: 131950728-131950728 on Build GRCh38
rs894933526 Chr.6: 131951124-131951124 on Build GRCh38
rs755868110 Chr.6: 131950803-131950803 on Build GRCh38
rs779565110 Chr.6: 131950837-131950837 on Build GRCh38
rs760642979 Chr.6: 131950927-131950927 on Build GRCh38
rs773759696 Chr.6: 131950814-131950814 on Build GRCh38
rs773804335 Chr.6: 131950738-131950738 on Build GRCh38
rs771454878 Chr.6: 131951148-131951148 on Build GRCh38
rs780986630 Chr.6: 131950849-131950849 on Build GRCh38
rs111779788 Chr.6: 131950887-131950887 on Build GRCh38
rs779822040 Chr.6: 131951076-131951076 on Build GRCh38
rs749158712 Chr.6: 131950737-131950737 on Build GRCh38
rs751729070 Chr.6: 131950796-131950796 on Build GRCh38
rs548588117 Chr.6: 131950990-131950990 on Build GRCh38
rs1000734182 Chr.6: 131950891-131950891 on Build GRCh38
rs754049250 Chr.6: 131950948-131950948 on Build GRCh38
rs925227422 Chr.6: 131951033-131951033 on Build GRCh38
rs768393319 Chr.6: 131950848-131950848 on Build GRCh38
rs755104088 Chr.6: 131950797-131950797 on Build GRCh38
rs532271775 Chr.6: 131950953-131950953 on Build GRCh38
rs754432225 Chr.6: 131951107-131951107 on Build GRCh38
rs747960681 Chr.6: 131950722-131950722 on Build GRCh38
rs112030855 Chr.6: 131950957-131950957 on Build GRCh38
rs759549246 Chr.6: 131950820-131950820 on Build GRCh38
rs936018221 Chr.6: 131950743-131950743 on Build GRCh38

View Full Product Details