Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136259331-136259860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

SNP Detail

SNP ID Chromosome Location
rs754337337 Chr.6: 136259568-136259568 on Build GRCh38
rs368739484 Chr.6: 136259633-136259633 on Build GRCh38
rs764642141 Chr.6: 136259460-136259460 on Build GRCh38
rs907238726 Chr.6: 136259722-136259722 on Build GRCh38
rs371841764 Chr.6: 136259684-136259684 on Build GRCh38
rs367778992 Chr.6: 136259788-136259789 on Build GRCh38
rs150244054 Chr.6: 136259529-136259530 on Build GRCh38
rs953744225 Chr.6: 136259404-136259404 on Build GRCh38
rs532058609 Chr.6: 136259472-136259472 on Build GRCh38
rs566681480 Chr.6: 136259562-136259563 on Build GRCh38
rs1031583022 Chr.6: 136259558-136259558 on Build GRCh38
rs1027000598 Chr.6: 136259822-136259822 on Build GRCh38
rs889542979 Chr.6: 136259788-136259791 on Build GRCh38
rs765306276 Chr.6: 136259693-136259693 on Build GRCh38
rs1028505821 Chr.6: 136259573-136259573 on Build GRCh38
rs374278886 Chr.6: 136259521-136259521 on Build GRCh38
rs568950269 Chr.6: 136259532-136259532 on Build GRCh38
rs568320753 Chr.6: 136259695-136259695 on Build GRCh38
rs1019207333 Chr.6: 136259688-136259688 on Build GRCh38
rs957217123 Chr.6: 136259377-136259377 on Build GRCh38
rs1007967330 Chr.6: 136259794-136259794 on Build GRCh38
rs893026514 Chr.6: 136259764-136259764 on Build GRCh38
rs944922461 Chr.6: 136259430-136259430 on Build GRCh38
rs972414286 Chr.6: 136259820-136259820 on Build GRCh38
rs907127117 Chr.6: 136259498-136259498 on Build GRCh38
rs934628886 Chr.6: 136259738-136259738 on Build GRCh38
rs575743558 Chr.6: 136259834-136259834 on Build GRCh38
rs1028120800 Chr.6: 136259787-136259788 on Build GRCh38
rs761948381 Chr.6: 136259629-136259629 on Build GRCh38
rs953774935 Chr.6: 136259620-136259620 on Build GRCh38
rs553373253 Chr.6: 136259730-136259730 on Build GRCh38
rs957454717 Chr.6: 136259569-136259569 on Build GRCh38
rs990343746 Chr.6: 136259390-136259390 on Build GRCh38
rs972766013 Chr.6: 136259694-136259694 on Build GRCh38
rs113624754 Chr.6: 136259830-136259830 on Build GRCh38
rs986402232 Chr.6: 136259417-136259418 on Build GRCh38
rs749896964 Chr.6: 136259708-136259708 on Build GRCh38
rs764664195 Chr.6: 136259630-136259630 on Build GRCh38
rs139977295 Chr.6: 136259774-136259774 on Build GRCh38
rs115229429 Chr.6: 136259692-136259692 on Build GRCh38
rs866551131 Chr.6: 136259707-136259707 on Build GRCh38
rs776830532 Chr.6: 136259451-136259451 on Build GRCh38
rs1036497231 Chr.6: 136259437-136259437 on Build GRCh38
rs574510374 Chr.6: 136259420-136259420 on Build GRCh38
rs1019320731 Chr.6: 136259817-136259817 on Build GRCh38
rs538186306 Chr.6: 136259570-136259570 on Build GRCh38
rs540843600 Chr.6: 136259475-136259475 on Build GRCh38
rs867458503 Chr.6: 136259450-136259450 on Build GRCh38
rs1031885501 Chr.6: 136259357-136259357 on Build GRCh38
rs1045723740 Chr.6: 136259479-136259479 on Build GRCh38
rs986434871 Chr.6: 136259685-136259686 on Build GRCh38
rs552383502 Chr.6: 136259493-136259493 on Build GRCh38
rs185633546 Chr.6: 136259741-136259741 on Build GRCh38
rs980773881 Chr.6: 136259394-136259394 on Build GRCh38
rs375793599 Chr.6: 136259709-136259709 on Build GRCh38
rs1049636096 Chr.6: 136259455-136259455 on Build GRCh38
rs961053245 Chr.6: 136259818-136259818 on Build GRCh38
rs998804979 Chr.6: 136259555-136259555 on Build GRCh38

View Full Product Details