Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136259986-136260536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

SNP Detail

SNP ID Chromosome Location
rs775676022 Chr.6: 136260337-136260337 on Build GRCh38
rs1035828810 Chr.6: 136260385-136260385 on Build GRCh38
rs745921650 Chr.6: 136260140-136260140 on Build GRCh38
rs1023202358 Chr.6: 136260061-136260061 on Build GRCh38
rs751916669 Chr.6: 136260395-136260396 on Build GRCh38
rs781348169 Chr.6: 136260038-136260038 on Build GRCh38
rs539167611 Chr.6: 136260349-136260349 on Build GRCh38
rs752727860 Chr.6: 136260065-136260065 on Build GRCh38
rs553436714 Chr.6: 136260282-136260282 on Build GRCh38
rs1007998378 Chr.6: 136260021-136260021 on Build GRCh38
rs533529080 Chr.6: 136260086-136260086 on Build GRCh38
rs918466706 Chr.6: 136260451-136260451 on Build GRCh38
rs530859476 Chr.6: 136260370-136260370 on Build GRCh38
rs985399584 Chr.6: 136260425-136260425 on Build GRCh38
rs1003046030 Chr.6: 136260362-136260362 on Build GRCh38
rs145877926 Chr.6: 136260034-136260034 on Build GRCh38
rs532450043 Chr.6: 136260092-136260092 on Build GRCh38
rs746853267 Chr.6: 136260015-136260015 on Build GRCh38
rs1040932723 Chr.6: 136260444-136260444 on Build GRCh38
rs1012311029 Chr.6: 136260339-136260339 on Build GRCh38
rs554723798 Chr.6: 136260432-136260432 on Build GRCh38
rs889655962 Chr.6: 136260019-136260019 on Build GRCh38
rs866714936 Chr.6: 136260048-136260048 on Build GRCh38
rs774318337 Chr.6: 136260093-136260093 on Build GRCh38
rs1040815207 Chr.6: 136260260-136260260 on Build GRCh38
rs568383755 Chr.6: 136260268-136260268 on Build GRCh38
rs914622734 Chr.6: 136260410-136260410 on Build GRCh38
rs970273121 Chr.6: 136260359-136260359 on Build GRCh38
rs148972267 Chr.6: 136260404-136260404 on Build GRCh38
rs769715497 Chr.6: 136260074-136260074 on Build GRCh38
rs533347950 Chr.6: 136260254-136260255 on Build GRCh38
rs562897961 Chr.6: 136260245-136260245 on Build GRCh38
rs544371813 Chr.6: 136260055-136260055 on Build GRCh38
rs369456697 Chr.6: 136260501-136260502 on Build GRCh38
rs755957082 Chr.6: 136260411-136260411 on Build GRCh38
rs545725303 Chr.6: 136260317-136260317 on Build GRCh38
rs182295681 Chr.6: 136260091-136260091 on Build GRCh38
rs540515983 Chr.6: 136260504-136260504 on Build GRCh38
rs914615611 Chr.6: 136260183-136260183 on Build GRCh38
rs538080705 Chr.6: 136260407-136260407 on Build GRCh38
rs929778464 Chr.6: 136260499-136260499 on Build GRCh38
rs558784509 Chr.6: 136260401-136260401 on Build GRCh38
rs759481553 Chr.6: 136260018-136260018 on Build GRCh38
rs766701834 Chr.6: 136260504-136260505 on Build GRCh38
rs776920842 Chr.6: 136260494-136260494 on Build GRCh38
rs777163723 Chr.6: 136260506-136260507 on Build GRCh38
rs534057513 Chr.6: 136260280-136260280 on Build GRCh38
rs568889560 Chr.6: 136260076-136260076 on Build GRCh38
rs772224072 Chr.6: 136260253-136260253 on Build GRCh38
rs767527519 Chr.6: 136260039-136260048 on Build GRCh38
rs878993513 Chr.6: 136260203-136260203 on Build GRCh38
rs764445743 Chr.6: 136260464-136260464 on Build GRCh38
rs761093287 Chr.6: 136260435-136260435 on Build GRCh38
rs559764045 Chr.6: 136260505-136260505 on Build GRCh38
rs574831293 Chr.6: 136260450-136260450 on Build GRCh38
rs911157202 Chr.6: 136260427-136260427 on Build GRCh38
rs576767256 Chr.6: 136260506-136260506 on Build GRCh38
rs947292431 Chr.6: 136260012-136260012 on Build GRCh38
rs748103896 Chr.6: 136260047-136260047 on Build GRCh38
rs956112232 Chr.6: 136260103-136260103 on Build GRCh38
rs548330874 Chr.6: 136260178-136260178 on Build GRCh38
rs994240357 Chr.6: 136260037-136260037 on Build GRCh38
rs1019352117 Chr.6: 136260027-136260027 on Build GRCh38
rs947372199 Chr.6: 136260202-136260202 on Build GRCh38
rs943888239 Chr.6: 136260255-136260255 on Build GRCh38
rs564913809 Chr.6: 136260331-136260331 on Build GRCh38
rs1048615901 Chr.6: 136260330-136260330 on Build GRCh38
rs369430729 Chr.6: 136260343-136260343 on Build GRCh38
rs993965280 Chr.6: 136260288-136260288 on Build GRCh38

View Full Product Details