Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136260719-136261229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420769 c.2757+2_2758del149 p.? Chr.6: 136261115-136261263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111800140 Chr.6: 136261148-136261148 on Build GRCh38
rs1022172931 Chr.6: 136260754-136260754 on Build GRCh38
rs376948280 Chr.6: 136261156-136261156 on Build GRCh38
rs370457195 Chr.6: 136261065-136261066 on Build GRCh38
rs764587274 Chr.6: 136261069-136261069 on Build GRCh38
rs1016842914 Chr.6: 136261031-136261031 on Build GRCh38
rs892051912 Chr.6: 136260928-136260928 on Build GRCh38
rs745705042 Chr.6: 136261060-136261061 on Build GRCh38
rs761958652 Chr.6: 136261071-136261071 on Build GRCh38
rs761076060 Chr.6: 136261067-136261067 on Build GRCh38
rs1043310605 Chr.6: 136260953-136260953 on Build GRCh38
rs189589087 Chr.6: 136260899-136260899 on Build GRCh38
rs1010423161 Chr.6: 136260750-136260750 on Build GRCh38
rs946383743 Chr.6: 136261088-136261088 on Build GRCh38
rs777506614 Chr.6: 136261121-136261122 on Build GRCh38
rs111572965 Chr.6: 136261033-136261033 on Build GRCh38
rs752457268 Chr.6: 136261117-136261118 on Build GRCh38
rs777128127 Chr.6: 136261155-136261155 on Build GRCh38
rs1052509152 Chr.6: 136260903-136260903 on Build GRCh38
rs372939218 Chr.6: 136260844-136260844 on Build GRCh38
rs554785112 Chr.6: 136260900-136260900 on Build GRCh38
rs562481221 Chr.6: 136261137-136261137 on Build GRCh38
rs188600091 Chr.6: 136261005-136261005 on Build GRCh38
rs184151570 Chr.6: 136260925-136260925 on Build GRCh38
rs951333293 Chr.6: 136260861-136260861 on Build GRCh38
rs779916409 Chr.6: 136261086-136261086 on Build GRCh38
rs193167212 Chr.6: 136261193-136261193 on Build GRCh38
rs570210520 Chr.6: 136261157-136261158 on Build GRCh38
rs751526906 Chr.6: 136261087-136261087 on Build GRCh38
rs946324153 Chr.6: 136260945-136260947 on Build GRCh38
rs538662536 Chr.6: 136260764-136260764 on Build GRCh38
rs769327751 Chr.6: 136261113-136261113 on Build GRCh38
rs998199203 Chr.6: 136260989-136260989 on Build GRCh38
rs761272577 Chr.6: 136261147-136261147 on Build GRCh38
rs62431280 Chr.6: 136260954-136260954 on Build GRCh38
rs1018295252 Chr.6: 136260801-136260801 on Build GRCh38
rs534080602 Chr.6: 136260927-136260927 on Build GRCh38
rs901241422 Chr.6: 136260988-136260988 on Build GRCh38
rs780977812 Chr.6: 136261092-136261092 on Build GRCh38
rs74299323 Chr.6: 136261065-136261065 on Build GRCh38
rs983980370 Chr.6: 136260864-136260865 on Build GRCh38
rs765557987 Chr.6: 136261073-136261073 on Build GRCh38
rs909743107 Chr.6: 136260881-136260881 on Build GRCh38
rs965376141 Chr.6: 136260812-136260812 on Build GRCh38
rs754614460 Chr.6: 136261180-136261180 on Build GRCh38
rs754701506 Chr.6: 136261090-136261090 on Build GRCh38
rs763071484 Chr.6: 136261115-136261116 on Build GRCh38
rs754050691 Chr.6: 136261070-136261070 on Build GRCh38
rs766196853 Chr.6: 136261082-136261082 on Build GRCh38
rs62431281 Chr.6: 136260975-136260975 on Build GRCh38
rs545502239 Chr.6: 136261017-136261017 on Build GRCh38
rs987833730 Chr.6: 136261072-136261072 on Build GRCh38
rs141067744 Chr.6: 136261056-136261057 on Build GRCh38
rs1039776896 Chr.6: 136260983-136260983 on Build GRCh38
rs980574896 Chr.6: 136261040-136261040 on Build GRCh38
rs554055107 Chr.6: 136260964-136260964 on Build GRCh38
rs370252607 Chr.6: 136261161-136261161 on Build GRCh38
rs774419180 Chr.6: 136261116-136261117 on Build GRCh38
rs76890371 Chr.6: 136261103-136261103 on Build GRCh38
rs1025576634 Chr.6: 136260829-136260829 on Build GRCh38
rs552518235 Chr.6: 136260825-136260825 on Build GRCh38
rs1026030273 Chr.6: 136260996-136260996 on Build GRCh38
rs748726912 Chr.6: 136261130-136261130 on Build GRCh38
rs1006997317 Chr.6: 136260974-136260978 on Build GRCh38
rs951407076 Chr.6: 136260997-136260997 on Build GRCh38
rs976690083 Chr.6: 136260815-136260815 on Build GRCh38
rs770590143 Chr.6: 136261135-136261135 on Build GRCh38
rs112744301 Chr.6: 136261174-136261174 on Build GRCh38
rs569283372 Chr.6: 136260827-136260827 on Build GRCh38
rs550650804 Chr.6: 136260865-136260865 on Build GRCh38
rs756785936 Chr.6: 136261141-136261142 on Build GRCh38
rs777524158 Chr.6: 136261120-136261120 on Build GRCh38
rs751133979 Chr.6: 136261128-136261129 on Build GRCh38
rs904721825 Chr.6: 136260960-136260960 on Build GRCh38
rs371791937 Chr.6: 136260896-136260896 on Build GRCh38
rs758124399 Chr.6: 136261119-136261122 on Build GRCh38
rs1047141822 Chr.6: 136261158-136261158 on Build GRCh38
rs1006967937 Chr.6: 136260791-136260791 on Build GRCh38
rs769442560 Chr.6: 136261112-136261113 on Build GRCh38
rs775830032 Chr.6: 136261113-136261114 on Build GRCh38
rs750678338 Chr.6: 136261157-136261157 on Build GRCh38

View Full Product Details