Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136266908-136267420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3928104 c.2470A>C p.T824P Chr.6: 136267103-136267103 on Build GRCh38
1672793 c.2467A>C p.N823H Chr.6: 136267106-136267106 on Build GRCh38
3174928 c.2426G>A p.R809K Chr.6: 136267147-136267147 on Build GRCh38
7196168 c.2419A>C p.R807R Chr.6: 136267154-136267154 on Build GRCh38
1733212 c.2401C>T p.R801* Chr.6: 136267172-136267172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772243141 Chr.6: 136267378-136267378 on Build GRCh38
rs956468943 Chr.6: 136266958-136266958 on Build GRCh38
rs763353757 Chr.6: 136267071-136267071 on Build GRCh38
rs774005725 Chr.6: 136266989-136266989 on Build GRCh38
rs760037324 Chr.6: 136267104-136267104 on Build GRCh38
rs368408779 Chr.6: 136266978-136266978 on Build GRCh38
rs974042601 Chr.6: 136267271-136267271 on Build GRCh38
rs767791179 Chr.6: 136267122-136267122 on Build GRCh38
rs1035172844 Chr.6: 136267341-136267341 on Build GRCh38
rs1044996085 Chr.6: 136266933-136266933 on Build GRCh38
rs773025129 Chr.6: 136267151-136267151 on Build GRCh38
rs773707737 Chr.6: 136267059-136267059 on Build GRCh38
rs917751400 Chr.6: 136267349-136267349 on Build GRCh38
rs745911999 Chr.6: 136267234-136267234 on Build GRCh38
rs771685980 Chr.6: 136267001-136267001 on Build GRCh38
rs745646406 Chr.6: 136267204-136267204 on Build GRCh38
rs905127545 Chr.6: 136266942-136266942 on Build GRCh38
rs779645066 Chr.6: 136267225-136267225 on Build GRCh38
rs775176708 Chr.6: 136267018-136267018 on Build GRCh38
rs955760413 Chr.6: 136267181-136267181 on Build GRCh38
rs911436322 Chr.6: 136267239-136267239 on Build GRCh38
rs919955263 Chr.6: 136267277-136267277 on Build GRCh38
rs964309786 Chr.6: 136267261-136267261 on Build GRCh38
rs771863884 Chr.6: 136267206-136267206 on Build GRCh38
rs983311306 Chr.6: 136267348-136267348 on Build GRCh38
rs1000450027 Chr.6: 136266948-136266948 on Build GRCh38
rs370472276 Chr.6: 136267219-136267220 on Build GRCh38
rs1047016213 Chr.6: 136267263-136267263 on Build GRCh38
rs760134383 Chr.6: 136267038-136267038 on Build GRCh38
rs770703920 Chr.6: 136266985-136266985 on Build GRCh38
rs762626590 Chr.6: 136267360-136267360 on Build GRCh38
rs930061375 Chr.6: 136267303-136267303 on Build GRCh38
rs763595083 Chr.6: 136267050-136267050 on Build GRCh38
rs778972627 Chr.6: 136267197-136267197 on Build GRCh38
rs745745742 Chr.6: 136266999-136266999 on Build GRCh38
rs1044880785 Chr.6: 136267375-136267375 on Build GRCh38
rs905005640 Chr.6: 136267332-136267332 on Build GRCh38
rs986971958 Chr.6: 136267198-136267198 on Build GRCh38
rs1038234324 Chr.6: 136267329-136267329 on Build GRCh38
rs766974371 Chr.6: 136267093-136267093 on Build GRCh38
rs749105442 Chr.6: 136267218-136267219 on Build GRCh38
rs142314074 Chr.6: 136266957-136266957 on Build GRCh38
rs949208610 Chr.6: 136267371-136267371 on Build GRCh38
rs756221834 Chr.6: 136267141-136267141 on Build GRCh38
rs1001997271 Chr.6: 136267335-136267335 on Build GRCh38
rs752971536 Chr.6: 136267129-136267129 on Build GRCh38
rs112720821 Chr.6: 136266964-136266964 on Build GRCh38
rs1005401391 Chr.6: 136267300-136267300 on Build GRCh38
rs369630817 Chr.6: 136267193-136267193 on Build GRCh38
rs751867871 Chr.6: 136267100-136267100 on Build GRCh38
rs757109222 Chr.6: 136267195-136267195 on Build GRCh38
rs567660108 Chr.6: 136267268-136267268 on Build GRCh38
rs749188779 Chr.6: 136266980-136266980 on Build GRCh38

View Full Product Details