Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138812846-138813342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
UniGene
Hs.660603
Gene Chromosome Location
Chr.6: 138796107-138904070 on Build GRCh38
Transcripts
NM_001077706.2 NM_001195037.2 XM_006715472.3 XM_011535795.2 XM_011535797.2 XM_017010828.1 XM_017010829.1 XM_017010830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4909578 c.50A>T p.K17M Chr.6: 138813324-138813324 on Build GRCh38
1073858 c.8G>A p.S3N Chr.6: 138813282-138813282 on Build GRCh38
7111691 c.37C>G p.P13A Chr.6: 138813311-138813311 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs908207338 Chr.6: 138812967-138812967 on Build GRCh38
rs17067914 Chr.6: 138812973-138812973 on Build GRCh38
rs555772773 Chr.6: 138813039-138813039 on Build GRCh38
rs370601948 Chr.6: 138813241-138813241 on Build GRCh38
rs911967864 Chr.6: 138813092-138813092 on Build GRCh38
rs944914498 Chr.6: 138813094-138813094 on Build GRCh38
rs373205876 Chr.6: 138813015-138813015 on Build GRCh38
rs1024101412 Chr.6: 138813296-138813296 on Build GRCh38
rs1027747946 Chr.6: 138812949-138812949 on Build GRCh38
rs755807456 Chr.6: 138813076-138813076 on Build GRCh38
rs903160397 Chr.6: 138813112-138813112 on Build GRCh38
rs729920 Chr.6: 138812943-138812943 on Build GRCh38
rs1050579012 Chr.6: 138813036-138813036 on Build GRCh38
rs537835684 Chr.6: 138812978-138812978 on Build GRCh38
rs957501144 Chr.6: 138813002-138813002 on Build GRCh38
rs764990525 Chr.6: 138813233-138813233 on Build GRCh38
rs771630511 Chr.6: 138813311-138813311 on Build GRCh38
rs768172622 Chr.6: 138813297-138813297 on Build GRCh38
rs771860037 Chr.6: 138813277-138813277 on Build GRCh38
rs9403003 Chr.6: 138812904-138812904 on Build GRCh38
rs933462976 Chr.6: 138813026-138813026 on Build GRCh38
rs79219465 Chr.6: 138813286-138813286 on Build GRCh38
rs994541712 Chr.6: 138812933-138812933 on Build GRCh38
rs753786785 Chr.6: 138813246-138813246 on Build GRCh38
rs961044495 Chr.6: 138813315-138813315 on Build GRCh38
rs553515536 Chr.6: 138813165-138813165 on Build GRCh38
rs566741916 Chr.6: 138812925-138812925 on Build GRCh38
rs140053131 Chr.6: 138812897-138812897 on Build GRCh38
rs115206074 Chr.6: 138813121-138813121 on Build GRCh38
rs746866 Chr.6: 138812900-138812900 on Build GRCh38
rs778668888 Chr.6: 138813258-138813258 on Build GRCh38
rs945463598 Chr.6: 138812874-138812874 on Build GRCh38
rs865937784 Chr.6: 138813050-138813050 on Build GRCh38
rs371475573 Chr.6: 138813270-138813270 on Build GRCh38
rs199868239 Chr.6: 138812942-138812942 on Build GRCh38
rs900747438 Chr.6: 138812930-138812930 on Build GRCh38
rs765108112 Chr.6: 138813248-138813248 on Build GRCh38
rs990053265 Chr.6: 138813047-138813047 on Build GRCh38
rs971255799 Chr.6: 138812979-138812979 on Build GRCh38
rs763540724 Chr.6: 138813301-138813301 on Build GRCh38
rs776053139 Chr.6: 138813300-138813300 on Build GRCh38
rs1042426284 Chr.6: 138812896-138812896 on Build GRCh38
rs945356731 Chr.6: 138813054-138813054 on Build GRCh38
rs779480754 Chr.6: 138813240-138813240 on Build GRCh38
rs746865 Chr.6: 138812966-138812966 on Build GRCh38
rs1057182816 Chr.6: 138813128-138813128 on Build GRCh38
rs1036201857 Chr.6: 138812875-138812875 on Build GRCh38
rs950390421 Chr.6: 138812961-138812961 on Build GRCh38
rs927220577 Chr.6: 138812996-138812996 on Build GRCh38
rs746864 Chr.6: 138812975-138812975 on Build GRCh38
rs577295331 Chr.6: 138813075-138813075 on Build GRCh38
rs895892030 Chr.6: 138813185-138813185 on Build GRCh38
rs979931546 Chr.6: 138812982-138812982 on Build GRCh38

View Full Product Details