Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138862598-138863124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
UniGene
Hs.660603
Gene Chromosome Location
Chr.6: 138796107-138904070 on Build GRCh38
Transcripts
NM_001077706.2 NM_001195037.2 XM_006715472.3 XM_011535795.2 XM_011535797.2 XM_017010828.1 XM_017010829.1 XM_017010830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6547690 c.1255G>T p.A419S Chr.6: 138862683-138862683 on Build GRCh38
3715415 c.1216C>G p.L406V Chr.6: 138862644-138862644 on Build GRCh38
4429491 c.1279T>G p.S427A Chr.6: 138862707-138862707 on Build GRCh38
3410627 c.1254G>A p.T418T Chr.6: 138862682-138862682 on Build GRCh38
5530327 c.1285C>T p.Q429* Chr.6: 138862713-138862713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748077477 Chr.6: 138862929-138862929 on Build GRCh38
rs779747050 Chr.6: 138862659-138862660 on Build GRCh38
rs756925456 Chr.6: 138862636-138862636 on Build GRCh38
rs1022254234 Chr.6: 138862875-138862875 on Build GRCh38
rs1011457835 Chr.6: 138863063-138863063 on Build GRCh38
rs866113921 Chr.6: 138862638-138862638 on Build GRCh38
rs886191314 Chr.6: 138862798-138862798 on Build GRCh38
rs751318664 Chr.6: 138862813-138862813 on Build GRCh38
rs1049214609 Chr.6: 138862962-138862962 on Build GRCh38
rs748924495 Chr.6: 138862626-138862626 on Build GRCh38
rs778633299 Chr.6: 138862644-138862644 on Build GRCh38
rs757090517 Chr.6: 138862712-138862712 on Build GRCh38
rs941202425 Chr.6: 138863048-138863048 on Build GRCh38
rs922175566 Chr.6: 138862945-138862945 on Build GRCh38
rs777238666 Chr.6: 138862672-138862672 on Build GRCh38
rs538457828 Chr.6: 138862854-138862854 on Build GRCh38
rs375186496 Chr.6: 138862706-138862706 on Build GRCh38
rs1055752551 Chr.6: 138863015-138863015 on Build GRCh38
rs9389629 Chr.6: 138862855-138862855 on Build GRCh38
rs1022731385 Chr.6: 138863079-138863079 on Build GRCh38
rs909694439 Chr.6: 138863047-138863047 on Build GRCh38
rs567724787 Chr.6: 138862828-138862828 on Build GRCh38
rs745312252 Chr.6: 138862653-138862653 on Build GRCh38
rs12204850 Chr.6: 138862742-138862742 on Build GRCh38
rs759850807 Chr.6: 138862768-138862768 on Build GRCh38
rs753564705 Chr.6: 138862711-138862711 on Build GRCh38
rs752608962 Chr.6: 138862690-138862690 on Build GRCh38
rs778381235 Chr.6: 138862715-138862715 on Build GRCh38
rs774651967 Chr.6: 138862685-138862685 on Build GRCh38
rs147410610 Chr.6: 138863004-138863004 on Build GRCh38
rs966170828 Chr.6: 138862940-138862940 on Build GRCh38
rs912078414 Chr.6: 138862934-138862934 on Build GRCh38
rs778758896 Chr.6: 138862853-138862853 on Build GRCh38
rs557325985 Chr.6: 138863001-138863001 on Build GRCh38
rs980165427 Chr.6: 138863042-138863042 on Build GRCh38
rs186071696 Chr.6: 138862805-138862805 on Build GRCh38
rs529117983 Chr.6: 138862628-138862628 on Build GRCh38
rs759490329 Chr.6: 138862686-138862686 on Build GRCh38
rs907509115 Chr.6: 138862966-138862966 on Build GRCh38
rs533317912 Chr.6: 138862741-138862741 on Build GRCh38
rs528900774 Chr.6: 138862943-138862943 on Build GRCh38
rs774402036 Chr.6: 138862747-138862747 on Build GRCh38
rs940441927 Chr.6: 138862974-138862974 on Build GRCh38
rs544397434 Chr.6: 138863080-138863080 on Build GRCh38
rs999588528 Chr.6: 138863090-138863090 on Build GRCh38
rs1001476821 Chr.6: 138862843-138862843 on Build GRCh38
rs200520042 Chr.6: 138862673-138862673 on Build GRCh38
rs539934657 Chr.6: 138863021-138863021 on Build GRCh38
rs113425654 Chr.6: 138862946-138862946 on Build GRCh38
rs750706600 Chr.6: 138862682-138862682 on Build GRCh38
rs1045490686 Chr.6: 138862988-138862988 on Build GRCh38
rs374094056 Chr.6: 138862746-138862746 on Build GRCh38
rs549491271 Chr.6: 138862822-138862822 on Build GRCh38
rs1188827 Chr.6: 138862907-138862907 on Build GRCh38
rs970110324 Chr.6: 138863033-138863033 on Build GRCh38
rs757819997 Chr.6: 138862733-138862733 on Build GRCh38
rs745567701 Chr.6: 138862730-138862730 on Build GRCh38
rs555453039 Chr.6: 138863043-138863043 on Build GRCh38
rs77918311 Chr.6: 138862938-138862938 on Build GRCh38
rs779763665 Chr.6: 138862734-138862734 on Build GRCh38
rs893043776 Chr.6: 138863046-138863046 on Build GRCh38
rs767675274 Chr.6: 138862689-138862689 on Build GRCh38
rs573726090 Chr.6: 138863073-138863073 on Build GRCh38
rs181427570 Chr.6: 138862681-138862681 on Build GRCh38
rs771634204 Chr.6: 138862666-138862666 on Build GRCh38
rs748905070 Chr.6: 138862731-138862731 on Build GRCh38
rs773772289 Chr.6: 138862675-138862675 on Build GRCh38
rs756100667 Chr.6: 138862693-138862693 on Build GRCh38

View Full Product Details