Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143164960-143165502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
UniGene
Hs.567501
Gene Chromosome Location
Chr.6: 143059363-143343594 on Build GRCh38
Transcripts
NM_001286587.1 NM_001286588.1 NM_001286589.1 NM_016108.3 XM_011535881.2 XM_011535882.2 XM_011535883.2 XM_011535884.2 XM_011535885.2 XM_017010927.1 XM_017010928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6597195 c.298-2A>G p.? Chr.6: 143165080-143165080 on Build GRCh38
6597198 c.353C>T p.P118L Chr.6: 143165137-143165137 on Build GRCh38
1074039 c.399+1G>T p.? Chr.6: 143165184-143165184 on Build GRCh38
6597196 c.298-2A>G p.? Chr.6: 143165080-143165080 on Build GRCh38
7096618 c.391C>T p.H131Y Chr.6: 143165175-143165175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145897074 Chr.6: 143165133-143165133 on Build GRCh38
rs139594961 Chr.6: 143165247-143165247 on Build GRCh38
rs1019805804 Chr.6: 143165318-143165318 on Build GRCh38
rs755080669 Chr.6: 143165151-143165151 on Build GRCh38
rs149932444 Chr.6: 143165374-143165374 on Build GRCh38
rs143567319 Chr.6: 143165074-143165074 on Build GRCh38
rs927909020 Chr.6: 143165372-143165372 on Build GRCh38
rs749123575 Chr.6: 143165187-143165187 on Build GRCh38
rs558581971 Chr.6: 143165437-143165437 on Build GRCh38
rs957802811 Chr.6: 143165178-143165178 on Build GRCh38
rs72990403 Chr.6: 143165433-143165433 on Build GRCh38
rs982479630 Chr.6: 143165329-143165329 on Build GRCh38
rs112799152 Chr.6: 143165418-143165418 on Build GRCh38
rs778661847 Chr.6: 143165193-143165193 on Build GRCh38
rs374307455 Chr.6: 143164991-143164991 on Build GRCh38
rs528069400 Chr.6: 143165375-143165375 on Build GRCh38
rs142048628 Chr.6: 143165128-143165128 on Build GRCh38
rs1010595663 Chr.6: 143165185-143165185 on Build GRCh38
rs781006947 Chr.6: 143165163-143165163 on Build GRCh38
rs144980162 Chr.6: 143165426-143165426 on Build GRCh38
rs762454142 Chr.6: 143165121-143165121 on Build GRCh38
rs543514158 Chr.6: 143165063-143165063 on Build GRCh38
rs199553241 Chr.6: 143165192-143165192 on Build GRCh38
rs186986122 Chr.6: 143165233-143165233 on Build GRCh38
rs748037705 Chr.6: 143165044-143165044 on Build GRCh38
rs1026190795 Chr.6: 143165376-143165376 on Build GRCh38
rs565670294 Chr.6: 143165460-143165460 on Build GRCh38
rs751681589 Chr.6: 143165138-143165138 on Build GRCh38
rs972076718 Chr.6: 143165391-143165391 on Build GRCh38
rs531566302 Chr.6: 143165203-143165203 on Build GRCh38
rs777447431 Chr.6: 143165204-143165207 on Build GRCh38
rs770624126 Chr.6: 143165071-143165071 on Build GRCh38
rs767100274 Chr.6: 143165097-143165097 on Build GRCh38
rs930742629 Chr.6: 143165421-143165421 on Build GRCh38
rs775194611 Chr.6: 143165204-143165204 on Build GRCh38
rs3761998 Chr.6: 143165177-143165177 on Build GRCh38
rs746494835 Chr.6: 143165064-143165064 on Build GRCh38
rs775112959 Chr.6: 143165108-143165108 on Build GRCh38
rs903449777 Chr.6: 143165467-143165467 on Build GRCh38
rs373539132 Chr.6: 143165160-143165160 on Build GRCh38
rs182471677 Chr.6: 143165229-143165229 on Build GRCh38
rs532666321 Chr.6: 143165242-143165242 on Build GRCh38
rs565575443 Chr.6: 143165320-143165320 on Build GRCh38
rs751201238 Chr.6: 143165208-143165209 on Build GRCh38
rs138489752 Chr.6: 143165137-143165137 on Build GRCh38
rs745560148 Chr.6: 143165194-143165194 on Build GRCh38
rs4596500 Chr.6: 143165030-143165030 on Build GRCh38
rs549776028 Chr.6: 143165205-143165205 on Build GRCh38
rs566118746 Chr.6: 143165281-143165281 on Build GRCh38
rs777505657 Chr.6: 143165184-143165184 on Build GRCh38
rs571563995 Chr.6: 143165213-143165213 on Build GRCh38
rs757886213 Chr.6: 143165064-143165065 on Build GRCh38
rs944895611 Chr.6: 143165434-143165434 on Build GRCh38
rs773049628 Chr.6: 143165062-143165062 on Build GRCh38
rs756976087 Chr.6: 143165219-143165220 on Build GRCh38
rs375819862 Chr.6: 143165390-143165390 on Build GRCh38
rs1050605865 Chr.6: 143165126-143165126 on Build GRCh38
rs140037174 Chr.6: 143165001-143165001 on Build GRCh38
rs149392906 Chr.6: 143165165-143165165 on Build GRCh38
rs367657248 Chr.6: 143165045-143165045 on Build GRCh38
rs577211712 Chr.6: 143164997-143164997 on Build GRCh38
rs369953522 Chr.6: 143165076-143165076 on Build GRCh38
rs775248117 Chr.6: 143165409-143165409 on Build GRCh38
rs549057749 Chr.6: 143165006-143165006 on Build GRCh38

View Full Product Details