Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143284028-143284525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
UniGene
Hs.567501
Gene Chromosome Location
Chr.6: 143059363-143343594 on Build GRCh38
Transcripts
NM_001286587.1 NM_016108.3 XM_011535881.2 XM_011535882.2 XM_011535883.2 XM_011535884.2 XM_011535885.2 XM_017010927.1 XM_017010928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309011 c.481G>T p.A161S Chr.6: 143284191-143284191 on Build GRCh38
1074040 c.429C>T p.I143I Chr.6: 143284139-143284139 on Build GRCh38
4350381 c.447C>T p.H149H Chr.6: 143284157-143284157 on Build GRCh38
4350379 c.404C>T p.T135M Chr.6: 143284114-143284114 on Build GRCh38
1074041 c.430G>A p.E144K Chr.6: 143284140-143284140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556372072 Chr.6: 143284404-143284404 on Build GRCh38
rs199674284 Chr.6: 143284216-143284216 on Build GRCh38
rs921612414 Chr.6: 143284466-143284466 on Build GRCh38
rs879203115 Chr.6: 143284084-143284084 on Build GRCh38
rs115894390 Chr.6: 143284337-143284337 on Build GRCh38
rs11556576 Chr.6: 143284124-143284124 on Build GRCh38
rs750244261 Chr.6: 143284171-143284171 on Build GRCh38
rs760858719 Chr.6: 143284228-143284228 on Build GRCh38
rs912167520 Chr.6: 143284333-143284333 on Build GRCh38
rs916213825 Chr.6: 143284394-143284394 on Build GRCh38
rs753977781 Chr.6: 143284108-143284108 on Build GRCh38
rs754644942 Chr.6: 143284264-143284264 on Build GRCh38
rs758301941 Chr.6: 143284244-143284244 on Build GRCh38
rs761882561 Chr.6: 143284233-143284233 on Build GRCh38
rs780615123 Chr.6: 143284190-143284190 on Build GRCh38
rs573915727 Chr.6: 143284346-143284346 on Build GRCh38
rs775453921 Chr.6: 143284221-143284221 on Build GRCh38
rs372829634 Chr.6: 143284083-143284083 on Build GRCh38
rs572523485 Chr.6: 143284330-143284330 on Build GRCh38
rs1036173772 Chr.6: 143284478-143284479 on Build GRCh38
rs113787178 Chr.6: 143284261-143284261 on Build GRCh38
rs778626539 Chr.6: 143284069-143284069 on Build GRCh38
rs987699393 Chr.6: 143284332-143284332 on Build GRCh38
rs575602728 Chr.6: 143284407-143284407 on Build GRCh38
rs546329561 Chr.6: 143284444-143284444 on Build GRCh38
rs773799531 Chr.6: 143284112-143284112 on Build GRCh38
rs201448093 Chr.6: 143284140-143284140 on Build GRCh38
rs17591575 Chr.6: 143284270-143284270 on Build GRCh38
rs369614433 Chr.6: 143284082-143284082 on Build GRCh38
rs1053483083 Chr.6: 143284376-143284376 on Build GRCh38
rs746219895 Chr.6: 143284189-143284189 on Build GRCh38
rs779783866 Chr.6: 143284087-143284087 on Build GRCh38
rs771347382 Chr.6: 143284194-143284194 on Build GRCh38
rs765394666 Chr.6: 143284233-143284233 on Build GRCh38
rs932197135 Chr.6: 143284064-143284064 on Build GRCh38
rs775643320 Chr.6: 143284145-143284145 on Build GRCh38
rs112232118 Chr.6: 143284280-143284280 on Build GRCh38
rs1046495496 Chr.6: 143284054-143284054 on Build GRCh38
rs747689252 Chr.6: 143284191-143284191 on Build GRCh38
rs970540759 Chr.6: 143284356-143284356 on Build GRCh38
rs115165047 Chr.6: 143284182-143284182 on Build GRCh38
rs535650203 Chr.6: 143284229-143284229 on Build GRCh38
rs564476465 Chr.6: 143284151-143284151 on Build GRCh38
rs202049124 Chr.6: 143284170-143284170 on Build GRCh38
rs932911524 Chr.6: 143284209-143284209 on Build GRCh38
rs1004700584 Chr.6: 143284467-143284467 on Build GRCh38
rs946208697 Chr.6: 143284101-143284101 on Build GRCh38
rs768001203 Chr.6: 143284096-143284096 on Build GRCh38
rs746528294 Chr.6: 143284094-143284094 on Build GRCh38
rs758207832 Chr.6: 143284179-143284179 on Build GRCh38
rs750340072 Chr.6: 143284239-143284239 on Build GRCh38
rs779872124 Chr.6: 143284181-143284181 on Build GRCh38
rs758555841 Chr.6: 143284411-143284411 on Build GRCh38
rs763409147 Chr.6: 143284114-143284114 on Build GRCh38
rs551188449 Chr.6: 143284231-143284231 on Build GRCh38
rs1049961092 Chr.6: 143284210-143284210 on Build GRCh38
rs544069135 Chr.6: 143284399-143284399 on Build GRCh38
rs368308461 Chr.6: 143284272-143284272 on Build GRCh38
rs557281301 Chr.6: 143284325-143284325 on Build GRCh38
rs771446237 Chr.6: 143284115-143284115 on Build GRCh38
rs114392595 Chr.6: 143284200-143284200 on Build GRCh38
rs1053193 Chr.6: 143284161-143284161 on Build GRCh38
rs936346428 Chr.6: 143284153-143284153 on Build GRCh38
rs3748071 Chr.6: 143284292-143284292 on Build GRCh38
rs766317855 Chr.6: 143284255-143284255 on Build GRCh38
rs906470610 Chr.6: 143284437-143284437 on Build GRCh38
rs1024807647 Chr.6: 143284342-143284342 on Build GRCh38
rs781047767 Chr.6: 143284128-143284128 on Build GRCh38
rs746255348 Chr.6: 143284218-143284218 on Build GRCh38
rs765097294 Chr.6: 143284235-143284235 on Build GRCh38
rs116356431 Chr.6: 143284154-143284154 on Build GRCh38
rs772388736 Chr.6: 143284220-143284220 on Build GRCh38
rs923702596 Chr.6: 143284398-143284398 on Build GRCh38
rs977864785 Chr.6: 143284365-143284365 on Build GRCh38
rs753936019 Chr.6: 143284157-143284157 on Build GRCh38
rs148165797 Chr.6: 143284117-143284117 on Build GRCh38
rs115998778 Chr.6: 143284167-143284167 on Build GRCh38
rs9403480 Chr.6: 143284335-143284335 on Build GRCh38
rs922160175 Chr.6: 143284057-143284057 on Build GRCh38

View Full Product Details