Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149578743-149579251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINM1
Entrez Gene ID
116254
Gene Name
glycoprotein integral membrane 1
Gene Aliases
C6orf72, dJ12G14.2
UniGene
Hs.438872
Gene Chromosome Location
Chr.6: 149566392-149590931 on Build GRCh38
Transcripts
NM_138785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5011690 c.318T>G p.Y106* Chr.6: 149578862-149578862 on Build GRCh38
3018663 c.357G>T p.W119C Chr.6: 149578901-149578901 on Build GRCh38
6322803 c.352G>T p.E118* Chr.6: 149578896-149578896 on Build GRCh38
3018661 c.290A>G p.N97S Chr.6: 149578834-149578834 on Build GRCh38
740655 c.292C>G p.L98V Chr.6: 149578836-149578836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745500034 Chr.6: 149578808-149578808 on Build GRCh38
rs1052985732 Chr.6: 149578877-149578877 on Build GRCh38
rs947800767 Chr.6: 149578971-149578971 on Build GRCh38
rs147513433 Chr.6: 149578780-149578780 on Build GRCh38
rs762173427 Chr.6: 149578797-149578797 on Build GRCh38
rs762455345 Chr.6: 149578985-149578985 on Build GRCh38
rs772714368 Chr.6: 149578798-149578798 on Build GRCh38
rs779084458 Chr.6: 149578941-149578941 on Build GRCh38
rs943477905 Chr.6: 149578809-149578809 on Build GRCh38
rs749425002 Chr.6: 149578919-149578919 on Build GRCh38
rs1043496282 Chr.6: 149579051-149579054 on Build GRCh38
rs769473842 Chr.6: 149578808-149578811 on Build GRCh38
rs769387638 Chr.6: 149578967-149578967 on Build GRCh38
rs761649344 Chr.6: 149579003-149579003 on Build GRCh38
rs748594378 Chr.6: 149578948-149578948 on Build GRCh38
rs763745277 Chr.6: 149578891-149578891 on Build GRCh38
rs1001728159 Chr.6: 149579154-149579154 on Build GRCh38
rs763805220 Chr.6: 149578998-149578998 on Build GRCh38
rs1027143421 Chr.6: 149578829-149578837 on Build GRCh38
rs774009534 Chr.6: 149578999-149578999 on Build GRCh38
rs1024589826 Chr.6: 149579063-149579063 on Build GRCh38
rs1014069421 Chr.6: 149579004-149579004 on Build GRCh38
rs547613857 Chr.6: 149579170-149579171 on Build GRCh38
rs768939034 Chr.6: 149578929-149578929 on Build GRCh38
rs767417000 Chr.6: 149579006-149579006 on Build GRCh38
rs775085484 Chr.6: 149578984-149578984 on Build GRCh38
rs745341969 Chr.6: 149578966-149578966 on Build GRCh38
rs772693938 Chr.6: 149578954-149578954 on Build GRCh38
rs200356862 Chr.6: 149578901-149578901 on Build GRCh38
rs145071660 Chr.6: 149578869-149578869 on Build GRCh38
rs142895897 Chr.6: 149579095-149579095 on Build GRCh38
rs756233383 Chr.6: 149578914-149578914 on Build GRCh38
rs73781238 Chr.6: 149579222-149579222 on Build GRCh38
rs935140533 Chr.6: 149579118-149579118 on Build GRCh38
rs750396998 Chr.6: 149579016-149579016 on Build GRCh38
rs559042905 Chr.6: 149578769-149578769 on Build GRCh38
rs78838302 Chr.6: 149579211-149579211 on Build GRCh38
rs184346776 Chr.6: 149578845-149578845 on Build GRCh38
rs769995253 Chr.6: 149578771-149578771 on Build GRCh38
rs962656082 Chr.6: 149579199-149579199 on Build GRCh38
rs374493485 Chr.6: 149578807-149578808 on Build GRCh38
rs1052240124 Chr.6: 149579191-149579191 on Build GRCh38
rs139415037 Chr.6: 149578868-149578868 on Build GRCh38
rs567809378 Chr.6: 149578795-149578795 on Build GRCh38
rs557030904 Chr.6: 149578903-149578903 on Build GRCh38
rs774433249 Chr.6: 149579153-149579153 on Build GRCh38
rs911951211 Chr.6: 149578794-149578794 on Build GRCh38
rs112955637 Chr.6: 149578792-149578792 on Build GRCh38
rs148558124 Chr.6: 149578899-149578899 on Build GRCh38
rs938916757 Chr.6: 149578906-149578906 on Build GRCh38
rs775806678 Chr.6: 149578974-149578975 on Build GRCh38
rs759175308 Chr.6: 149578852-149578852 on Build GRCh38
rs1016589742 Chr.6: 149579181-149579181 on Build GRCh38
rs750484367 Chr.6: 149578905-149578905 on Build GRCh38
rs547926103 Chr.6: 149578783-149578783 on Build GRCh38
rs867547443 Chr.6: 149578918-149578918 on Build GRCh38
rs764963346 Chr.6: 149578857-149578857 on Build GRCh38

View Full Product Details