Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 156779346-156779853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID1B
Entrez Gene ID
57492
Gene Name
AT-rich interaction domain 1B
Gene Aliases
6A3-5, BAF250B, BRIGHT, CSS1, DAN15, ELD/OSA1, MRD12, OSA2, P250R
UniGene
Hs.291587
Gene Chromosome Location
Chr.6: 156777847-157210779 on Build GRCh38
Transcripts
NM_017519.2 NM_020732.3 XM_005267069.4 XM_011535984.2 XM_017011103.1 XM_017011104.1 XM_017011105.1 XM_017011106.1 XM_017011107.1 XM_017011108.1 NR_039676.1

Gene Symbol
MIR4466
Entrez Gene ID
100616154
Gene Name
microRNA 4466
Gene Aliases
mir-4466
UniGene
Gene Chromosome Location
Chr.6: 156779678-156779731 on Build GRCh38
Transcripts
NM_017519.2 NM_020732.3 XM_005267069.4 XM_011535984.2 XM_017011103.1 XM_017011104.1 XM_017011105.1 XM_017011106.1 XM_017011107.1 XM_017011108.1 NR_039676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6974698 c.1313G>T p.R438M Chr.6: 156779416-156779416 on Build GRCh38
6942462 c.1514delC p.G507fs*16 Chr.6: 156779443-156779443 on Build GRCh38
6955045 c.1368G>T p.Q456H Chr.6: 156779471-156779471 on Build GRCh38
6945875 c.1368_1425del58 p.A458fs*25 Chr.6: 156779297-156779354 on Build GRCh38
6944806 c.1295G>A p.W432* Chr.6: 156779398-156779398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374013482 Chr.6: 156779490-156779490 on Build GRCh38
rs952954478 Chr.6: 156779738-156779738 on Build GRCh38
rs201457919 Chr.6: 156779731-156779731 on Build GRCh38
rs757043988 Chr.6: 156779426-156779426 on Build GRCh38
rs1057445868 Chr.6: 156779741-156779741 on Build GRCh38
rs776060836 Chr.6: 156779385-156779385 on Build GRCh38
rs954294716 Chr.6: 156779593-156779593 on Build GRCh38
rs369180699 Chr.6: 156779740-156779740 on Build GRCh38
rs866333772 Chr.6: 156779531-156779531 on Build GRCh38
rs1048417678 Chr.6: 156779668-156779668 on Build GRCh38
rs113805612 Chr.6: 156779708-156779708 on Build GRCh38
rs1006981715 Chr.6: 156779694-156779694 on Build GRCh38
rs781091058 Chr.6: 156779370-156779370 on Build GRCh38
rs887659581 Chr.6: 156779524-156779524 on Build GRCh38
rs775006984 Chr.6: 156779489-156779489 on Build GRCh38
rs764750241 Chr.6: 156779487-156779487 on Build GRCh38
rs934280014 Chr.6: 156779824-156779824 on Build GRCh38
rs912771983 Chr.6: 156779484-156779484 on Build GRCh38
rs771204934 Chr.6: 156779368-156779368 on Build GRCh38
rs1013395627 Chr.6: 156779680-156779684 on Build GRCh38
rs985753980 Chr.6: 156779597-156779597 on Build GRCh38
rs780240139 Chr.6: 156779521-156779521 on Build GRCh38
rs889554130 Chr.6: 156779796-156779796 on Build GRCh38
rs1006520338 Chr.6: 156779806-156779806 on Build GRCh38
rs571744458 Chr.6: 156779676-156779676 on Build GRCh38
rs751389585 Chr.6: 156779425-156779425 on Build GRCh38
rs953766475 Chr.6: 156779693-156779693 on Build GRCh38
rs1013738446 Chr.6: 156779569-156779570 on Build GRCh38
rs902380524 Chr.6: 156779772-156779772 on Build GRCh38
rs763825843 Chr.6: 156779422-156779422 on Build GRCh38
rs775035352 Chr.6: 156779461-156779461 on Build GRCh38
rs781454908 Chr.6: 156779428-156779428 on Build GRCh38
rs563927911 Chr.6: 156779740-156779740 on Build GRCh38
rs1002160755 Chr.6: 156779558-156779558 on Build GRCh38
rs972762489 Chr.6: 156779735-156779735 on Build GRCh38
rs1032003769 Chr.6: 156779590-156779590 on Build GRCh38
rs918634432 Chr.6: 156779661-156779661 on Build GRCh38
rs759119564 Chr.6: 156779482-156779482 on Build GRCh38
rs753486869 Chr.6: 156779404-156779404 on Build GRCh38
rs757061915 Chr.6: 156779510-156779510 on Build GRCh38
rs1053439861 Chr.6: 156779795-156779795 on Build GRCh38
rs904689488 Chr.6: 156779679-156779688 on Build GRCh38
rs933434611 Chr.6: 156779621-156779621 on Build GRCh38
rs925501086 Chr.6: 156779397-156779397 on Build GRCh38
rs1000612645 Chr.6: 156779686-156779686 on Build GRCh38
rs751547411 Chr.6: 156779509-156779509 on Build GRCh38
rs371093846 Chr.6: 156779488-156779488 on Build GRCh38
rs1042211507 Chr.6: 156779682-156779682 on Build GRCh38
rs985602910 Chr.6: 156779771-156779771 on Build GRCh38
rs771434224 Chr.6: 156779415-156779417 on Build GRCh38
rs746135430 Chr.6: 156779438-156779438 on Build GRCh38
rs769813347 Chr.6: 156779379-156779379 on Build GRCh38
rs541066146 Chr.6: 156779731-156779732 on Build GRCh38
rs769085274 Chr.6: 156779392-156779392 on Build GRCh38
rs902265847 Chr.6: 156779684-156779684 on Build GRCh38
rs931280573 Chr.6: 156779516-156779516 on Build GRCh38
rs1020630288 Chr.6: 156779571-156779571 on Build GRCh38
rs534914874 Chr.6: 156779662-156779662 on Build GRCh38
rs917808909 Chr.6: 156779743-156779743 on Build GRCh38
rs774880058 Chr.6: 156779393-156779393 on Build GRCh38
rs559757088 Chr.6: 156779749-156779749 on Build GRCh38
rs1048427547 Chr.6: 156779519-156779519 on Build GRCh38
rs1031951814 Chr.6: 156779690-156779690 on Build GRCh38
rs965546597 Chr.6: 156779634-156779634 on Build GRCh38
rs1021018834 Chr.6: 156779375-156779375 on Build GRCh38
rs763559359 Chr.6: 156779386-156779386 on Build GRCh38
rs912709060 Chr.6: 156779613-156779613 on Build GRCh38
rs371019276 Chr.6: 156779515-156779515 on Build GRCh38
rs1000557295 Chr.6: 156779780-156779780 on Build GRCh38
rs776346531 Chr.6: 156779479-156779479 on Build GRCh38
rs764716697 Chr.6: 156779419-156779419 on Build GRCh38
rs1051844511 Chr.6: 156779709-156779709 on Build GRCh38
rs999059656 Chr.6: 156779679-156779712 on Build GRCh38
rs200682868 Chr.6: 156779451-156779451 on Build GRCh38
rs759078059 Chr.6: 156779417-156779417 on Build GRCh38
rs760366486 Chr.6: 156779394-156779394 on Build GRCh38
rs772375225 Chr.6: 156779476-156779476 on Build GRCh38
rs547211045 Chr.6: 156779667-156779667 on Build GRCh38
rs911416865 Chr.6: 156779785-156779785 on Build GRCh38
rs973330889 Chr.6: 156779640-156779640 on Build GRCh38
rs920138047 Chr.6: 156779737-156779737 on Build GRCh38
rs575325490 Chr.6: 156779758-156779758 on Build GRCh38
rs766027272 Chr.6: 156779396-156779396 on Build GRCh38
rs769316078 Chr.6: 156779460-156779460 on Build GRCh38
rs536209708 Chr.6: 156779818-156779818 on Build GRCh38
rs749761849 Chr.6: 156779459-156779459 on Build GRCh38
rs780293502 Chr.6: 156779457-156779457 on Build GRCh38
rs943085787 Chr.6: 156779803-156779803 on Build GRCh38
rs748498440 Chr.6: 156779462-156779462 on Build GRCh38
rs1033924784 Chr.6: 156779561-156779561 on Build GRCh38
rs994459233 Chr.6: 156779820-156779820 on Build GRCh38
rs927317304 Chr.6: 156779674-156779674 on Build GRCh38
rs966018057 Chr.6: 156779710-156779710 on Build GRCh38
rs538765400 Chr.6: 156779677-156779677 on Build GRCh38
rs745929521 Chr.6: 156779378-156779378 on Build GRCh38
rs896341761 Chr.6: 156779564-156779564 on Build GRCh38
rs758342480 Chr.6: 156779421-156779421 on Build GRCh38
rs1019705284 Chr.6: 156779810-156779810 on Build GRCh38
rs890986408 Chr.6: 156779739-156779739 on Build GRCh38
rs966436886 Chr.6: 156779581-156779581 on Build GRCh38
rs1040560993 Chr.6: 156779813-156779813 on Build GRCh38
rs376716533 Chr.6: 156779508-156779508 on Build GRCh38
rs756310926 Chr.6: 156779517-156779517 on Build GRCh38
rs961717130 Chr.6: 156779734-156779734 on Build GRCh38

View Full Product Details