Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158766442-158766941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EZR
Entrez Gene ID
7430
Gene Name
ezrin
Gene Aliases
CVIL, CVL, HEL-S-105, VIL2
UniGene
Hs.487027
Gene Chromosome Location
Chr.6: 158765741-158819424 on Build GRCh38
Transcripts
NM_001111077.1 NM_003379.4 XM_011536110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3022953 c.1749C>T p.F583F Chr.6: 158766926-158766926 on Build GRCh38
5869684 c.1736G>A p.R579H Chr.6: 158766939-158766939 on Build GRCh38
1075456 c.1756C>A p.L586M Chr.6: 158766919-158766919 on Build GRCh38
3022952 c.1750G>A p.E584K Chr.6: 158766925-158766925 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768336741 Chr.6: 158766911-158766911 on Build GRCh38
rs998804603 Chr.6: 158766555-158766555 on Build GRCh38
rs5881286 Chr.6: 158766846-158766847 on Build GRCh38
rs182678633 Chr.6: 158766594-158766594 on Build GRCh38
rs79494479 Chr.6: 158766817-158766817 on Build GRCh38
rs550560050 Chr.6: 158766712-158766712 on Build GRCh38
rs188249634 Chr.6: 158766651-158766651 on Build GRCh38
rs866744065 Chr.6: 158766759-158766759 on Build GRCh38
rs41267143 Chr.6: 158766859-158766859 on Build GRCh38
rs1033971613 Chr.6: 158766474-158766474 on Build GRCh38
rs768022562 Chr.6: 158766897-158766902 on Build GRCh38
rs544519538 Chr.6: 158766781-158766781 on Build GRCh38
rs56233811 Chr.6: 158766874-158766874 on Build GRCh38
rs992342676 Chr.6: 158766700-158766700 on Build GRCh38
rs971262467 Chr.6: 158766545-158766545 on Build GRCh38
rs1466582 Chr.6: 158766783-158766783 on Build GRCh38
rs774821624 Chr.6: 158766882-158766883 on Build GRCh38
rs879552434 Chr.6: 158766623-158766624 on Build GRCh38
rs764346678 Chr.6: 158766476-158766476 on Build GRCh38
rs780696470 Chr.6: 158766898-158766898 on Build GRCh38
rs187595394 Chr.6: 158766788-158766788 on Build GRCh38
rs1013688860 Chr.6: 158766855-158766855 on Build GRCh38
rs1017632676 Chr.6: 158766808-158766808 on Build GRCh38
rs750431779 Chr.6: 158766908-158766912 on Build GRCh38
rs534201251 Chr.6: 158766697-158766697 on Build GRCh38
rs77551523 Chr.6: 158766742-158766742 on Build GRCh38
rs775749130 Chr.6: 158766768-158766768 on Build GRCh38
rs41267141 Chr.6: 158766750-158766750 on Build GRCh38
rs1009890335 Chr.6: 158766601-158766601 on Build GRCh38
rs908197573 Chr.6: 158766714-158766715 on Build GRCh38
rs1030492982 Chr.6: 158766834-158766834 on Build GRCh38
rs765580487 Chr.6: 158766890-158766890 on Build GRCh38
rs950959151 Chr.6: 158766864-158766864 on Build GRCh38
rs370515638 Chr.6: 158766875-158766875 on Build GRCh38
rs933236058 Chr.6: 158766765-158766765 on Build GRCh38
rs1001049983 Chr.6: 158766664-158766664 on Build GRCh38
rs1051444324 Chr.6: 158766598-158766598 on Build GRCh38
rs762087641 Chr.6: 158766891-158766891 on Build GRCh38
rs1055311873 Chr.6: 158766852-158766852 on Build GRCh38
rs778855911 Chr.6: 158766554-158766555 on Build GRCh38
rs1004892293 Chr.6: 158766868-158766868 on Build GRCh38
rs1039271296 Chr.6: 158766564-158766564 on Build GRCh38
rs556142898 Chr.6: 158766525-158766525 on Build GRCh38
rs191909538 Chr.6: 158766741-158766741 on Build GRCh38
rs929406367 Chr.6: 158766521-158766521 on Build GRCh38
rs78423323 Chr.6: 158766607-158766607 on Build GRCh38
rs558753283 Chr.6: 158766751-158766751 on Build GRCh38
rs34894691 Chr.6: 158766851-158766851 on Build GRCh38
rs1001669322 Chr.6: 158766837-158766837 on Build GRCh38
rs754359350 Chr.6: 158766878-158766878 on Build GRCh38
rs372850570 Chr.6: 158766870-158766870 on Build GRCh38
rs746879555 Chr.6: 158766685-158766685 on Build GRCh38
rs532589131 Chr.6: 158766608-158766608 on Build GRCh38
rs567290131 Chr.6: 158766717-158766717 on Build GRCh38
rs370481018 Chr.6: 158766590-158766590 on Build GRCh38
rs987296583 Chr.6: 158766881-158766881 on Build GRCh38
rs945730331 Chr.6: 158766814-158766814 on Build GRCh38
rs3062709 Chr.6: 158766849-158766850 on Build GRCh38
rs376089037 Chr.6: 158766573-158766573 on Build GRCh38
rs757001899 Chr.6: 158766896-158766896 on Build GRCh38
rs371215514 Chr.6: 158766895-158766895 on Build GRCh38
rs941912571 Chr.6: 158766541-158766541 on Build GRCh38
rs886802494 Chr.6: 158766487-158766487 on Build GRCh38
rs532665522 Chr.6: 158766856-158766856 on Build GRCh38
rs200919889 Chr.6: 158766907-158766907 on Build GRCh38
rs1021415135 Chr.6: 158766611-158766611 on Build GRCh38
rs779987639 Chr.6: 158766905-158766905 on Build GRCh38
rs2230144 Chr.6: 158766909-158766909 on Build GRCh38
rs796443530 Chr.6: 158766829-158766829 on Build GRCh38
rs560211236 Chr.6: 158766854-158766854 on Build GRCh38
rs1024105752 Chr.6: 158766532-158766532 on Build GRCh38
rs540272246 Chr.6: 158766853-158766853 on Build GRCh38
rs1034225601 Chr.6: 158766687-158766687 on Build GRCh38
rs749936264 Chr.6: 158766833-158766833 on Build GRCh38
rs756341222 Chr.6: 158766471-158766471 on Build GRCh38
rs571928743 Chr.6: 158766524-158766524 on Build GRCh38
rs753553182 Chr.6: 158766852-158766852 on Build GRCh38
rs536303202 Chr.6: 158766734-158766734 on Build GRCh38
rs762025818 Chr.6: 158766863-158766863 on Build GRCh38
rs1025167940 Chr.6: 158766705-158766705 on Build GRCh38
rs891498802 Chr.6: 158766600-158766600 on Build GRCh38
rs2230145 Chr.6: 158766892-158766892 on Build GRCh38
rs201308333 Chr.6: 158766880-158766880 on Build GRCh38
rs756400625 Chr.6: 158766787-158766787 on Build GRCh38
rs530108801 Chr.6: 158766542-158766542 on Build GRCh38
rs749975221 Chr.6: 158766903-158766903 on Build GRCh38
rs755580961 Chr.6: 158766904-158766904 on Build GRCh38
rs376264050 Chr.6: 158766673-158766673 on Build GRCh38
rs964800606 Chr.6: 158766643-158766643 on Build GRCh38
rs544266885 Chr.6: 158766510-158766510 on Build GRCh38
rs558493857 Chr.6: 158766482-158766482 on Build GRCh38
rs904275027 Chr.6: 158766836-158766836 on Build GRCh38
rs762210286 Chr.6: 158766888-158766889 on Build GRCh38
rs763084832 Chr.6: 158766773-158766773 on Build GRCh38
rs999168713 Chr.6: 158766764-158766764 on Build GRCh38
rs377270609 Chr.6: 158766642-158766642 on Build GRCh38
rs575417627 Chr.6: 158766775-158766775 on Build GRCh38
rs917893449 Chr.6: 158766512-158766512 on Build GRCh38
rs1005701668 Chr.6: 158766496-158766496 on Build GRCh38
rs75522256 Chr.6: 158766699-158766699 on Build GRCh38
rs769395453 Chr.6: 158766858-158766858 on Build GRCh38
rs540332956 Chr.6: 158766565-158766565 on Build GRCh38
rs779523318 Chr.6: 158766849-158766849 on Build GRCh38
rs773479905 Chr.6: 158766871-158766871 on Build GRCh38
rs569427464 Chr.6: 158766698-158766698 on Build GRCh38
rs577592023 Chr.6: 158766826-158766826 on Build GRCh38

View Full Product Details