Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159169481-159169979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC1
Entrez Gene ID
84624
Gene Name
fibronectin type III domain containing 1
Gene Aliases
AGS8, FNDC2, MEL4B3, bA243O10.1, dJ322A24.1
UniGene
Hs.520525
Gene Chromosome Location
Chr.6: 159169397-159272108 on Build GRCh38
Transcripts
NM_032532.2 XM_011536190.2 XM_011536191.2 NR_121668.1

Gene Symbol
LOC101929122
Entrez Gene ID
101929122
Gene Name
uncharacterized LOC101929122
Gene Aliases
-
UniGene
Hs.134215
Gene Chromosome Location
Chr.6: 159165899-159170007 on Build GRCh38
Transcripts
NM_032532.2 XM_011536190.2 XM_011536191.2 NR_121668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6284603 c.10G>T p.E4* Chr.6: 159169606-159169606 on Build GRCh38
5480010 c.106T>C p.S36P Chr.6: 159169702-159169702 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs561026331 Chr.6: 159169854-159169854 on Build GRCh38
rs966310344 Chr.6: 159169916-159169916 on Build GRCh38
rs544425804 Chr.6: 159169769-159169769 on Build GRCh38
rs907153764 Chr.6: 159169826-159169826 on Build GRCh38
rs1491431 Chr.6: 159169933-159169933 on Build GRCh38
rs540565338 Chr.6: 159169888-159169888 on Build GRCh38
rs1024195861 Chr.6: 159169806-159169806 on Build GRCh38
rs545005095 Chr.6: 159169753-159169753 on Build GRCh38
rs912211167 Chr.6: 159169902-159169902 on Build GRCh38
rs558701793 Chr.6: 159169758-159169758 on Build GRCh38
rs975431005 Chr.6: 159169721-159169721 on Build GRCh38
rs568662089 Chr.6: 159169681-159169681 on Build GRCh38
rs940409585 Chr.6: 159169612-159169612 on Build GRCh38
rs953855907 Chr.6: 159169887-159169887 on Build GRCh38
rs295332 Chr.6: 159169702-159169702 on Build GRCh38
rs529842086 Chr.6: 159169856-159169856 on Build GRCh38
rs1004072935 Chr.6: 159169848-159169848 on Build GRCh38
rs189260029 Chr.6: 159169605-159169605 on Build GRCh38
rs947741515 Chr.6: 159169819-159169819 on Build GRCh38
rs1023592926 Chr.6: 159169651-159169651 on Build GRCh38
rs534463114 Chr.6: 159169697-159169697 on Build GRCh38
rs995666220 Chr.6: 159169882-159169882 on Build GRCh38
rs1047204980 Chr.6: 159169599-159169599 on Build GRCh38
rs976031875 Chr.6: 159169534-159169534 on Build GRCh38
rs1037381998 Chr.6: 159169620-159169620 on Build GRCh38
rs1037815212 Chr.6: 159169774-159169774 on Build GRCh38
rs868759075 Chr.6: 159169830-159169830 on Build GRCh38
rs989515398 Chr.6: 159169508-159169508 on Build GRCh38
rs942974862 Chr.6: 159169523-159169523 on Build GRCh38
rs1022298612 Chr.6: 159169717-159169717 on Build GRCh38
rs778293692 Chr.6: 159169833-159169833 on Build GRCh38
rs922853757 Chr.6: 159169578-159169578 on Build GRCh38
rs964431964 Chr.6: 159169720-159169720 on Build GRCh38
rs116831202 Chr.6: 159169945-159169945 on Build GRCh38
rs924778904 Chr.6: 159169931-159169931 on Build GRCh38
rs1028037587 Chr.6: 159169883-159169883 on Build GRCh38
rs986440426 Chr.6: 159169889-159169889 on Build GRCh38
rs193207381 Chr.6: 159169747-159169747 on Build GRCh38
rs915198514 Chr.6: 159169512-159169512 on Build GRCh38
rs1015968632 Chr.6: 159169505-159169505 on Build GRCh38
rs906436665 Chr.6: 159169652-159169652 on Build GRCh38
rs898791029 Chr.6: 159169624-159169624 on Build GRCh38
rs982930641 Chr.6: 159169766-159169766 on Build GRCh38
rs1003404090 Chr.6: 159169680-159169680 on Build GRCh38
rs1012166358 Chr.6: 159169628-159169628 on Build GRCh38
rs1003432191 Chr.6: 159169837-159169837 on Build GRCh38
rs1012892943 Chr.6: 159169772-159169772 on Build GRCh38
rs898909138 Chr.6: 159169800-159169800 on Build GRCh38
rs768929564 Chr.6: 159169851-159169851 on Build GRCh38
rs996826927 Chr.6: 159169910-159169910 on Build GRCh38
rs934193740 Chr.6: 159169582-159169582 on Build GRCh38
rs977669259 Chr.6: 159169925-159169925 on Build GRCh38
rs575385762 Chr.6: 159169767-159169767 on Build GRCh38
rs745445002 Chr.6: 159169633-159169633 on Build GRCh38

View Full Product Details