Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167004079-167004608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1OP
Entrez Gene ID
11116
Gene Name
FGFR1 oncogene partner
Gene Aliases
FOP
UniGene
Hs.487175
Gene Chromosome Location
Chr.6: 166999317-167042418 on Build GRCh38
Transcripts
NM_001278690.1 NM_007045.3 NM_194429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061633 c.434G>A p.G145E Chr.6: 167004397-167004397 on Build GRCh38
6105597 c.355G>A p.A119T Chr.6: 167004318-167004318 on Build GRCh38
6663692 c.381A>G p.L127L Chr.6: 167004344-167004344 on Build GRCh38
6368785 c.427A>T p.T143S Chr.6: 167004390-167004390 on Build GRCh38
5345102 c.411G>C p.Q137H Chr.6: 167004374-167004374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027303366 Chr.6: 167004426-167004426 on Build GRCh38
rs373877546 Chr.6: 167004440-167004440 on Build GRCh38
rs563320838 Chr.6: 167004235-167004235 on Build GRCh38
rs372181150 Chr.6: 167004438-167004438 on Build GRCh38
rs541491815 Chr.6: 167004187-167004187 on Build GRCh38
rs137945032 Chr.6: 167004200-167004200 on Build GRCh38
rs41269603 Chr.6: 167004237-167004237 on Build GRCh38
rs550989637 Chr.6: 167004482-167004482 on Build GRCh38
rs528148538 Chr.6: 167004363-167004363 on Build GRCh38
rs147838580 Chr.6: 167004336-167004336 on Build GRCh38
rs558085803 Chr.6: 167004513-167004513 on Build GRCh38
rs755408934 Chr.6: 167004414-167004414 on Build GRCh38
rs868230727 Chr.6: 167004282-167004282 on Build GRCh38
rs761928563 Chr.6: 167004328-167004328 on Build GRCh38
rs758281252 Chr.6: 167004280-167004280 on Build GRCh38
rs897254363 Chr.6: 167004465-167004465 on Build GRCh38
rs751553240 Chr.6: 167004385-167004385 on Build GRCh38
rs747089586 Chr.6: 167004441-167004441 on Build GRCh38
rs183464060 Chr.6: 167004231-167004231 on Build GRCh38
rs778650916 Chr.6: 167004423-167004424 on Build GRCh38
rs766532128 Chr.6: 167004344-167004344 on Build GRCh38
rs756768766 Chr.6: 167004288-167004288 on Build GRCh38
rs767097035 Chr.6: 167004390-167004390 on Build GRCh38
rs370159451 Chr.6: 167004227-167004227 on Build GRCh38
rs367732297 Chr.6: 167004446-167004446 on Build GRCh38
rs1016753202 Chr.6: 167004176-167004176 on Build GRCh38
rs569275694 Chr.6: 167004496-167004496 on Build GRCh38
rs775004829 Chr.6: 167004319-167004319 on Build GRCh38
rs750026770 Chr.6: 167004393-167004393 on Build GRCh38
rs192917663 Chr.6: 167004507-167004507 on Build GRCh38
rs779550639 Chr.6: 167004423-167004423 on Build GRCh38
rs577936102 Chr.6: 167004261-167004261 on Build GRCh38
rs768334999 Chr.6: 167004263-167004263 on Build GRCh38
rs571727258 Chr.6: 167004129-167004129 on Build GRCh38
rs993996116 Chr.6: 167004500-167004500 on Build GRCh38
rs925511261 Chr.6: 167004522-167004522 on Build GRCh38
rs201719790 Chr.6: 167004297-167004297 on Build GRCh38
rs149745085 Chr.6: 167004312-167004312 on Build GRCh38
rs749597430 Chr.6: 167004411-167004411 on Build GRCh38
rs980824653 Chr.6: 167004125-167004125 on Build GRCh38
rs566872828 Chr.6: 167004517-167004517 on Build GRCh38
rs534249601 Chr.6: 167004539-167004539 on Build GRCh38
rs749696198 Chr.6: 167004309-167004309 on Build GRCh38
rs748703185 Chr.6: 167004431-167004431 on Build GRCh38
rs759246301 Chr.6: 167004260-167004260 on Build GRCh38
rs761003904 Chr.6: 167004338-167004338 on Build GRCh38
rs747844706 Chr.6: 167004227-167004228 on Build GRCh38
rs1007953440 Chr.6: 167004307-167004307 on Build GRCh38
rs765120307 Chr.6: 167004269-167004269 on Build GRCh38
rs142365952 Chr.6: 167004276-167004276 on Build GRCh38
rs955257496 Chr.6: 167004568-167004568 on Build GRCh38
rs992162508 Chr.6: 167004481-167004481 on Build GRCh38
rs780519335 Chr.6: 167004300-167004300 on Build GRCh38
rs763557841 Chr.6: 167004219-167004219 on Build GRCh38
rs139592603 Chr.6: 167004298-167004298 on Build GRCh38
rs371234650 Chr.6: 167004248-167004248 on Build GRCh38
rs755746985 Chr.6: 167004399-167004399 on Build GRCh38
rs939413896 Chr.6: 167004190-167004190 on Build GRCh38
rs913001256 Chr.6: 167004334-167004334 on Build GRCh38
rs553173511 Chr.6: 167004197-167004197 on Build GRCh38
rs768837785 Chr.6: 167004226-167004226 on Build GRCh38
rs553236582 Chr.6: 167004549-167004549 on Build GRCh38
rs752493987 Chr.6: 167004348-167004348 on Build GRCh38
rs574660861 Chr.6: 167004554-167004554 on Build GRCh38
rs79205895 Chr.6: 167004447-167004447 on Build GRCh38
rs762737904 Chr.6: 167004349-167004349 on Build GRCh38
rs546806854 Chr.6: 167004386-167004386 on Build GRCh38
rs758781791 Chr.6: 167004228-167004228 on Build GRCh38
rs150980464 Chr.6: 167004287-167004287 on Build GRCh38
rs777715690 Chr.6: 167004286-167004286 on Build GRCh38
rs146935481 Chr.6: 167004148-167004148 on Build GRCh38
rs750672272 Chr.6: 167004249-167004249 on Build GRCh38
rs375338123 Chr.6: 167004275-167004275 on Build GRCh38
rs952133940 Chr.6: 167004114-167004114 on Build GRCh38
rs779604786 Chr.6: 167004401-167004401 on Build GRCh38
rs1050446324 Chr.6: 167004581-167004581 on Build GRCh38
rs1015360658 Chr.6: 167004364-167004364 on Build GRCh38
rs773273833 Chr.6: 167004337-167004337 on Build GRCh38

View Full Product Details