Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167024700-167025213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1OP
Entrez Gene ID
11116
Gene Name
FGFR1 oncogene partner
Gene Aliases
FOP
UniGene
Hs.487175
Gene Chromosome Location
Chr.6: 166999317-167042418 on Build GRCh38
Transcripts
NM_001278690.1 NM_007045.3 NM_194429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2854387 c.859delC p.L289fs*1 Chr.6: 167024834-167024834 on Build GRCh38
6663694 c.846G>A p.S282S Chr.6: 167024821-167024821 on Build GRCh38
4383361 c.901T>C p.S301P Chr.6: 167024876-167024876 on Build GRCh38
3622806 c.884C>T p.S295F Chr.6: 167024859-167024859 on Build GRCh38
4511159 c.859C>T p.P287S Chr.6: 167024834-167024834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572071822 Chr.6: 167025061-167025061 on Build GRCh38
rs374763053 Chr.6: 167024774-167024774 on Build GRCh38
rs372855523 Chr.6: 167024900-167024900 on Build GRCh38
rs754897235 Chr.6: 167024830-167024830 on Build GRCh38
rs142143894 Chr.6: 167024865-167024865 on Build GRCh38
rs367630074 Chr.6: 167024885-167024885 on Build GRCh38
rs548073256 Chr.6: 167025179-167025179 on Build GRCh38
rs201260130 Chr.6: 167024868-167024868 on Build GRCh38
rs16899801 Chr.6: 167024910-167024910 on Build GRCh38
rs576096362 Chr.6: 167024952-167024952 on Build GRCh38
rs980693981 Chr.6: 167024950-167024950 on Build GRCh38
rs970220555 Chr.6: 167024927-167024927 on Build GRCh38
rs921690958 Chr.6: 167025077-167025077 on Build GRCh38
rs1052040925 Chr.6: 167025096-167025096 on Build GRCh38
rs561476550 Chr.6: 167024906-167024906 on Build GRCh38
rs1050460937 Chr.6: 167024981-167024981 on Build GRCh38
rs907057299 Chr.6: 167025122-167025122 on Build GRCh38
rs753708860 Chr.6: 167024794-167024794 on Build GRCh38
rs139065630 Chr.6: 167024755-167024755 on Build GRCh38
rs970546961 Chr.6: 167025160-167025160 on Build GRCh38
rs542616378 Chr.6: 167025072-167025072 on Build GRCh38
rs571875042 Chr.6: 167024818-167024818 on Build GRCh38
rs368174160 Chr.6: 167024821-167024821 on Build GRCh38
rs780022240 Chr.6: 167025003-167025003 on Build GRCh38
rs527422006 Chr.6: 167024745-167024745 on Build GRCh38
rs1034720959 Chr.6: 167025124-167025124 on Build GRCh38
rs9366080 Chr.6: 167024954-167024954 on Build GRCh38
rs146053844 Chr.6: 167024799-167024799 on Build GRCh38
rs746247439 Chr.6: 167024882-167024882 on Build GRCh38
rs1010276800 Chr.6: 167025106-167025106 on Build GRCh38
rs760134894 Chr.6: 167024800-167024800 on Build GRCh38
rs750688106 Chr.6: 167024887-167024887 on Build GRCh38
rs759261178 Chr.6: 167024933-167024933 on Build GRCh38
rs539004868 Chr.6: 167025104-167025104 on Build GRCh38
rs373826808 Chr.6: 167024826-167024826 on Build GRCh38
rs34544438 Chr.6: 167024804-167024804 on Build GRCh38
rs556298683 Chr.6: 167024963-167024963 on Build GRCh38
rs774683787 Chr.6: 167024909-167024909 on Build GRCh38
rs777414178 Chr.6: 167024782-167024782 on Build GRCh38
rs776448285 Chr.6: 167024930-167024930 on Build GRCh38
rs1045515375 Chr.6: 167025109-167025109 on Build GRCh38
rs373374377 Chr.6: 167024969-167024970 on Build GRCh38
rs750178877 Chr.6: 167024733-167024733 on Build GRCh38
rs375006435 Chr.6: 167024971-167024971 on Build GRCh38
rs769917255 Chr.6: 167024932-167024933 on Build GRCh38
rs776157779 Chr.6: 167024890-167024890 on Build GRCh38
rs753697193 Chr.6: 167024828-167024828 on Build GRCh38
rs776968306 Chr.6: 167024793-167024793 on Build GRCh38
rs565468282 Chr.6: 167025132-167025132 on Build GRCh38
rs369413811 Chr.6: 167024932-167024932 on Build GRCh38
rs199785921 Chr.6: 167024783-167024783 on Build GRCh38
rs149375415 Chr.6: 167024827-167024827 on Build GRCh38
rs560981260 Chr.6: 167025119-167025119 on Build GRCh38
rs17856382 Chr.6: 167024788-167024788 on Build GRCh38
rs532558468 Chr.6: 167025148-167025148 on Build GRCh38
rs571645509 Chr.6: 167025003-167025003 on Build GRCh38
rs550155221 Chr.6: 167025130-167025130 on Build GRCh38
rs765144069 Chr.6: 167024942-167024942 on Build GRCh38
rs144758821 Chr.6: 167024838-167024838 on Build GRCh38
rs768927871 Chr.6: 167024901-167024901 on Build GRCh38
rs1004842171 Chr.6: 167025121-167025121 on Build GRCh38
rs771643421 Chr.6: 167024759-167024759 on Build GRCh38
rs754415909 Chr.6: 167024751-167024751 on Build GRCh38
rs78350210 Chr.6: 167025123-167025123 on Build GRCh38
rs1027045940 Chr.6: 167025138-167025138 on Build GRCh38
rs755954152 Chr.6: 167024737-167024737 on Build GRCh38
rs1049727673 Chr.6: 167024763-167024763 on Build GRCh38
rs778855080 Chr.6: 167024837-167024837 on Build GRCh38
rs148535456 Chr.6: 167024850-167024850 on Build GRCh38
rs73028285 Chr.6: 167025002-167025002 on Build GRCh38
rs971246302 Chr.6: 167024848-167024848 on Build GRCh38
rs192661051 Chr.6: 167024977-167024977 on Build GRCh38
rs747681897 Chr.6: 167024756-167024756 on Build GRCh38
rs1035856296 Chr.6: 167025187-167025187 on Build GRCh38
rs767657286 Chr.6: 167024926-167024926 on Build GRCh38
rs1027552744 Chr.6: 167024913-167024913 on Build GRCh38
rs753880798 Chr.6: 167024750-167024750 on Build GRCh38
rs995924433 Chr.6: 167025131-167025131 on Build GRCh38
rs34585172 Chr.6: 167024876-167024876 on Build GRCh38
rs931811136 Chr.6: 167025094-167025094 on Build GRCh38
rs890370031 Chr.6: 167025103-167025103 on Build GRCh38
rs147827735 Chr.6: 167024866-167024866 on Build GRCh38
rs371596730 Chr.6: 167024824-167024824 on Build GRCh38

View Full Product Details