Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167136088-167136631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR6
Entrez Gene ID
1235
Gene Name
C-C motif chemokine receptor 6
Gene Aliases
BN-1, C-C CKR-6, CC-CKR-6, CCR-6, CD196, CKR-L3, CKRL3, CMKBR6, DCR2, DRY6, GPR29, GPRCY4, STRL22
UniGene
Hs.46468
Gene Chromosome Location
Chr.6: 167111807-167139141 on Build GRCh38
Transcripts
NM_004367.5 NM_031409.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3622809 c.159C>T p.Y53Y Chr.6: 167136389-167136389 on Build GRCh38
6012578 c.228G>A p.K76K Chr.6: 167136458-167136458 on Build GRCh38
7285338 c.205A>G p.I69V Chr.6: 167136435-167136435 on Build GRCh38
3622808 c.111C>T p.S37S Chr.6: 167136341-167136341 on Build GRCh38
7283519 c.65T>C p.V22A Chr.6: 167136295-167136295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761667128 Chr.6: 167136130-167136130 on Build GRCh38
rs777851811 Chr.6: 167136283-167136283 on Build GRCh38
rs555016384 Chr.6: 167136139-167136139 on Build GRCh38
rs764255386 Chr.6: 167136249-167136249 on Build GRCh38
rs774555492 Chr.6: 167136485-167136485 on Build GRCh38
rs754544366 Chr.6: 167136142-167136142 on Build GRCh38
rs368226324 Chr.6: 167136187-167136187 on Build GRCh38
rs879208011 Chr.6: 167136468-167136468 on Build GRCh38
rs778156994 Chr.6: 167136165-167136165 on Build GRCh38
rs201556077 Chr.6: 167136559-167136559 on Build GRCh38
rs781647307 Chr.6: 167136462-167136462 on Build GRCh38
rs781755270 Chr.6: 167136308-167136308 on Build GRCh38
rs775673474 Chr.6: 167136335-167136335 on Build GRCh38
rs767284243 Chr.6: 167136574-167136574 on Build GRCh38
rs765162012 Chr.6: 167136366-167136366 on Build GRCh38
rs761331255 Chr.6: 167136569-167136569 on Build GRCh38
rs759425376 Chr.6: 167136365-167136365 on Build GRCh38
rs770445125 Chr.6: 167136331-167136331 on Build GRCh38
rs866553112 Chr.6: 167136553-167136554 on Build GRCh38
rs751102128 Chr.6: 167136568-167136568 on Build GRCh38
rs745938330 Chr.6: 167136195-167136195 on Build GRCh38
rs766330428 Chr.6: 167136262-167136262 on Build GRCh38
rs753843727 Chr.6: 167136263-167136263 on Build GRCh38
rs369165836 Chr.6: 167136260-167136260 on Build GRCh38
rs747383397 Chr.6: 167136296-167136296 on Build GRCh38
rs770674712 Chr.6: 167136358-167136358 on Build GRCh38
rs138813075 Chr.6: 167136495-167136495 on Build GRCh38
rs775830961 Chr.6: 167136206-167136206 on Build GRCh38
rs545588880 Chr.6: 167136274-167136274 on Build GRCh38
rs372714474 Chr.6: 167136353-167136353 on Build GRCh38
rs757628234 Chr.6: 167136304-167136304 on Build GRCh38
rs888046323 Chr.6: 167136266-167136266 on Build GRCh38
rs752140862 Chr.6: 167136378-167136378 on Build GRCh38
rs768865506 Chr.6: 167136348-167136348 on Build GRCh38
rs369997005 Chr.6: 167136560-167136560 on Build GRCh38
rs1013966200 Chr.6: 167136145-167136145 on Build GRCh38
rs145768514 Chr.6: 167136267-167136267 on Build GRCh38
rs748371509 Chr.6: 167136512-167136512 on Build GRCh38
rs760459591 Chr.6: 167136464-167136473 on Build GRCh38
rs751210852 Chr.6: 167136416-167136416 on Build GRCh38
rs143044968 Chr.6: 167136307-167136307 on Build GRCh38
rs193057414 Chr.6: 167136172-167136172 on Build GRCh38
rs9459886 Chr.6: 167136155-167136155 on Build GRCh38
rs776337420 Chr.6: 167136534-167136534 on Build GRCh38
rs145186145 Chr.6: 167136380-167136380 on Build GRCh38
rs761001428 Chr.6: 167136376-167136376 on Build GRCh38
rs1023318886 Chr.6: 167136253-167136253 on Build GRCh38
rs376737549 Chr.6: 167136221-167136221 on Build GRCh38
rs774953497 Chr.6: 167136257-167136257 on Build GRCh38
rs777299703 Chr.6: 167136292-167136292 on Build GRCh38
rs758000401 Chr.6: 167136290-167136290 on Build GRCh38
rs998980459 Chr.6: 167136170-167136170 on Build GRCh38
rs759335396 Chr.6: 167136551-167136551 on Build GRCh38
rs578256598 Chr.6: 167136255-167136255 on Build GRCh38
rs749384834 Chr.6: 167136338-167136338 on Build GRCh38
rs765542349 Chr.6: 167136119-167136119 on Build GRCh38
rs746424070 Chr.6: 167136323-167136323 on Build GRCh38
rs772307515 Chr.6: 167136527-167136527 on Build GRCh38
rs774495378 Chr.6: 167136350-167136350 on Build GRCh38
rs775469681 Chr.6: 167136555-167136555 on Build GRCh38
rs910055630 Chr.6: 167136464-167136464 on Build GRCh38
rs878877796 Chr.6: 167136194-167136194 on Build GRCh38
rs201769708 Chr.6: 167136129-167136129 on Build GRCh38
rs750849369 Chr.6: 167136473-167136473 on Build GRCh38
rs1014068342 Chr.6: 167136197-167136197 on Build GRCh38
rs185551935 Chr.6: 167136437-167136437 on Build GRCh38
rs3093007 Chr.6: 167136287-167136287 on Build GRCh38
rs200741825 Chr.6: 167136164-167136164 on Build GRCh38
rs112836572 Chr.6: 167136169-167136169 on Build GRCh38
rs747335022 Chr.6: 167136171-167136171 on Build GRCh38
rs754346206 Chr.6: 167136593-167136593 on Build GRCh38
rs2071171 Chr.6: 167136554-167136554 on Build GRCh38
rs976975121 Chr.6: 167136524-167136524 on Build GRCh38
rs778812775 Chr.6: 167136598-167136598 on Build GRCh38
rs373361643 Chr.6: 167136581-167136581 on Build GRCh38
rs372109980 Chr.6: 167136205-167136205 on Build GRCh38
rs755065875 Chr.6: 167136595-167136595 on Build GRCh38
rs747841027 Chr.6: 167136156-167136156 on Build GRCh38
rs766231476 Chr.6: 167136141-167136141 on Build GRCh38
rs763796957 Chr.6: 167136411-167136411 on Build GRCh38
rs950265536 Chr.6: 167136537-167136537 on Build GRCh38
rs370354168 Chr.6: 167136580-167136580 on Build GRCh38
rs762913226 Chr.6: 167136140-167136140 on Build GRCh38
rs76452893 Chr.6: 167136477-167136477 on Build GRCh38
rs998971859 Chr.6: 167136168-167136168 on Build GRCh38
rs41347847 Chr.6: 167136476-167136476 on Build GRCh38

View Full Product Details