Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167138448-167138982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR6
Entrez Gene ID
1235
Gene Name
C-C motif chemokine receptor 6
Gene Aliases
BN-1, C-C CKR-6, CC-CKR-6, CCR-6, CD196, CKR-L3, CKRL3, CMKBR6, DCR2, DRY6, GPR29, GPRCY4, STRL22
UniGene
Hs.46468
Gene Chromosome Location
Chr.6: 167111807-167139141 on Build GRCh38
Transcripts
NM_004367.5 NM_031409.3

SNP Detail

SNP ID Chromosome Location
rs745720012 Chr.6: 167138751-167138751 on Build GRCh38
rs35977074 Chr.6: 167138949-167138950 on Build GRCh38
rs10946217 Chr.6: 167138539-167138539 on Build GRCh38
rs768799983 Chr.6: 167138843-167138843 on Build GRCh38
rs973110352 Chr.6: 167138662-167138662 on Build GRCh38
rs952063798 Chr.6: 167138580-167138580 on Build GRCh38
rs374914584 Chr.6: 167138800-167138800 on Build GRCh38
rs888056770 Chr.6: 167138842-167138842 on Build GRCh38
rs949191837 Chr.6: 167138773-167138773 on Build GRCh38
rs116855733 Chr.6: 167138633-167138633 on Build GRCh38
rs1055983314 Chr.6: 167138816-167138816 on Build GRCh38
rs763472427 Chr.6: 167138598-167138598 on Build GRCh38
rs1004593094 Chr.6: 167138605-167138605 on Build GRCh38
rs11575084 Chr.6: 167138704-167138704 on Build GRCh38
rs998019977 Chr.6: 167138942-167138942 on Build GRCh38
rs986048423 Chr.6: 167138672-167138672 on Build GRCh38
rs898802553 Chr.6: 167138844-167138844 on Build GRCh38
rs11575083 Chr.6: 167138625-167138625 on Build GRCh38
rs925168696 Chr.6: 167138698-167138698 on Build GRCh38
rs757987475 Chr.6: 167138571-167138571 on Build GRCh38
rs947464261 Chr.6: 167138523-167138523 on Build GRCh38
rs932176604 Chr.6: 167138789-167138789 on Build GRCh38
rs1053528438 Chr.6: 167138490-167138490 on Build GRCh38
rs185625723 Chr.6: 167138846-167138846 on Build GRCh38
rs969181838 Chr.6: 167138670-167138670 on Build GRCh38
rs947494952 Chr.6: 167138769-167138769 on Build GRCh38
rs962065709 Chr.6: 167138929-167138929 on Build GRCh38
rs550036935 Chr.6: 167138700-167138700 on Build GRCh38
rs11575086 Chr.6: 167138915-167138915 on Build GRCh38
rs1030571222 Chr.6: 167138590-167138590 on Build GRCh38
rs531527941 Chr.6: 167138665-167138665 on Build GRCh38
rs891917210 Chr.6: 167138522-167138522 on Build GRCh38
rs1043211717 Chr.6: 167138534-167138534 on Build GRCh38
rs755844205 Chr.6: 167138772-167138772 on Build GRCh38
rs191584399 Chr.6: 167138717-167138717 on Build GRCh38
rs1005511890 Chr.6: 167138836-167138836 on Build GRCh38
rs41472245 Chr.6: 167138797-167138797 on Build GRCh38
rs1015379845 Chr.6: 167138929-167138930 on Build GRCh38
rs181542353 Chr.6: 167138750-167138750 on Build GRCh38
rs11575085 Chr.6: 167138798-167138798 on Build GRCh38
rs768821373 Chr.6: 167138790-167138790 on Build GRCh38
rs780256313 Chr.6: 167138784-167138784 on Build GRCh38
rs994601684 Chr.6: 167138878-167138878 on Build GRCh38
rs9366094 Chr.6: 167138912-167138912 on Build GRCh38
rs1014914817 Chr.6: 167138625-167138629 on Build GRCh38

View Full Product Details