Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168307182-168307682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT2
Entrez Gene ID
168002
Gene Name
dishevelled binding antagonist of beta catenin 2
Gene Aliases
C6orf116, DAPPER2, DPR2, bA503C24.7
UniGene
Hs.673059
Gene Chromosome Location
Chr.6: 168292830-168319754 on Build GRCh38
Transcripts
NM_001286350.1 NM_001286351.1 NM_214462.4 NR_104425.1 XM_011535507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5404807 c.2290C>T p.Q764* Chr.6: 168307467-168307467 on Build GRCh38
4477322 c.2142C>T p.D714D Chr.6: 168307615-168307615 on Build GRCh38
2855024 c.1058G>A p.R353H Chr.6: 168307475-168307475 on Build GRCh38
7317274 c.1100G>A p.*367* Chr.6: 168307433-168307433 on Build GRCh38
4478280 c.2226C>T p.S742S Chr.6: 168307531-168307531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777452202 Chr.6: 168307388-168307388 on Build GRCh38
rs939875880 Chr.6: 168307418-168307418 on Build GRCh38
rs755191135 Chr.6: 168307358-168307358 on Build GRCh38
rs749556652 Chr.6: 168307279-168307279 on Build GRCh38
rs141181495 Chr.6: 168307594-168307594 on Build GRCh38
rs386708739 Chr.6: 168307268-168307269 on Build GRCh38
rs1022596797 Chr.6: 168307508-168307508 on Build GRCh38
rs1013001400 Chr.6: 168307366-168307366 on Build GRCh38
rs538595072 Chr.6: 168307531-168307531 on Build GRCh38
rs1026219940 Chr.6: 168307437-168307437 on Build GRCh38
rs3734904 Chr.6: 168307269-168307269 on Build GRCh38
rs528258862 Chr.6: 168307312-168307312 on Build GRCh38
rs753920748 Chr.6: 168307584-168307584 on Build GRCh38
rs779514990 Chr.6: 168307512-168307512 on Build GRCh38
rs191740169 Chr.6: 168307590-168307590 on Build GRCh38
rs376391573 Chr.6: 168307433-168307433 on Build GRCh38
rs933896853 Chr.6: 168307347-168307347 on Build GRCh38
rs909312744 Chr.6: 168307527-168307527 on Build GRCh38
rs975444541 Chr.6: 168307322-168307322 on Build GRCh38
rs150741979 Chr.6: 168307619-168307619 on Build GRCh38
rs76687634 Chr.6: 168307313-168307313 on Build GRCh38
rs145931031 Chr.6: 168307555-168307555 on Build GRCh38
rs773925540 Chr.6: 168307582-168307582 on Build GRCh38
rs143076446 Chr.6: 168307372-168307372 on Build GRCh38
rs906894440 Chr.6: 168307649-168307649 on Build GRCh38
rs748726350 Chr.6: 168307648-168307648 on Build GRCh38
rs542687859 Chr.6: 168307233-168307233 on Build GRCh38
rs919131785 Chr.6: 168307265-168307265 on Build GRCh38
rs757141349 Chr.6: 168307598-168307598 on Build GRCh38
rs546503764 Chr.6: 168307334-168307334 on Build GRCh38
rs761233827 Chr.6: 168307583-168307583 on Build GRCh38
rs539288457 Chr.6: 168307539-168307539 on Build GRCh38
rs568823591 Chr.6: 168307638-168307638 on Build GRCh38
rs1039010120 Chr.6: 168307301-168307301 on Build GRCh38
rs950560804 Chr.6: 168307460-168307460 on Build GRCh38
rs529604381 Chr.6: 168307518-168307518 on Build GRCh38
rs376125439 Chr.6: 168307647-168307647 on Build GRCh38
rs991156059 Chr.6: 168307247-168307247 on Build GRCh38
rs778690232 Chr.6: 168307651-168307651 on Build GRCh38
rs971463179 Chr.6: 168307417-168307417 on Build GRCh38
rs921282121 Chr.6: 168307591-168307591 on Build GRCh38
rs559080276 Chr.6: 168307650-168307650 on Build GRCh38
rs1044536046 Chr.6: 168307637-168307637 on Build GRCh38
rs397840733 Chr.6: 168307268-168307268 on Build GRCh38
rs975154313 Chr.6: 168307576-168307576 on Build GRCh38
rs3832456 Chr.6: 168307267-168307267 on Build GRCh38
rs781247710 Chr.6: 168307475-168307475 on Build GRCh38
rs1013024191 Chr.6: 168307234-168307234 on Build GRCh38
rs1032719849 Chr.6: 168307232-168307232 on Build GRCh38
rs575958765 Chr.6: 168307595-168307595 on Build GRCh38
rs909284784 Chr.6: 168307296-168307296 on Build GRCh38
rs1022585565 Chr.6: 168307251-168307251 on Build GRCh38
rs765595101 Chr.6: 168307363-168307363 on Build GRCh38
rs1038190101 Chr.6: 168307428-168307428 on Build GRCh38
rs982296605 Chr.6: 168307255-168307255 on Build GRCh38
rs550211485 Chr.6: 168307503-168307503 on Build GRCh38
rs774580804 Chr.6: 168307547-168307547 on Build GRCh38
rs780932715 Chr.6: 168307599-168307599 on Build GRCh38
rs754404453 Chr.6: 168307268-168307268 on Build GRCh38
rs370941715 Chr.6: 168307635-168307635 on Build GRCh38
rs769332995 Chr.6: 168307511-168307511 on Build GRCh38
rs201216441 Chr.6: 168307269-168307269 on Build GRCh38
rs1035154687 Chr.6: 168307264-168307264 on Build GRCh38
rs772312400 Chr.6: 168307556-168307556 on Build GRCh38
rs1035206749 Chr.6: 168307549-168307549 on Build GRCh38
rs933551685 Chr.6: 168307321-168307321 on Build GRCh38
rs956964757 Chr.6: 168307554-168307554 on Build GRCh38
rs75202118 Chr.6: 168307297-168307297 on Build GRCh38
rs78695494 Chr.6: 168307636-168307636 on Build GRCh38
rs138529256 Chr.6: 168307566-168307566 on Build GRCh38
rs981691137 Chr.6: 168307256-168307256 on Build GRCh38
rs370013092 Chr.6: 168307459-168307459 on Build GRCh38
rs12530360 Chr.6: 168307365-168307365 on Build GRCh38
rs746780026 Chr.6: 168307404-168307404 on Build GRCh38
rs559267718 Chr.6: 168307346-168307346 on Build GRCh38
rs969648604 Chr.6: 168307254-168307254 on Build GRCh38
rs768009869 Chr.6: 168307479-168307481 on Build GRCh38
rs1022596226 Chr.6: 168307393-168307394 on Build GRCh38
rs1043022135 Chr.6: 168307328-168307328 on Build GRCh38
rs1012661748 Chr.6: 168307463-168307463 on Build GRCh38

View Full Product Details