Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168308285-168308776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT2
Entrez Gene ID
168002
Gene Name
dishevelled binding antagonist of beta catenin 2
Gene Aliases
C6orf116, DAPPER2, DPR2, bA503C24.7
UniGene
Hs.673059
Gene Chromosome Location
Chr.6: 168292830-168319754 on Build GRCh38
Transcripts
NM_001286350.1 NM_001286351.1 NM_214462.4 NR_104425.1 XM_011535507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5404812 c.143G>A p.G48E Chr.6: 168308390-168308390 on Build GRCh38
1076092 c.1329C>A p.S443R Chr.6: 168308428-168308428 on Build GRCh38
1076094 c.1269G>T p.E423D Chr.6: 168308488-168308488 on Build GRCh38
5404814 c.7C>T p.L3F Chr.6: 168308526-168308526 on Build GRCh38
7248364 c.149G>A p.G50D Chr.6: 168308384-168308384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760355743 Chr.6: 168308730-168308730 on Build GRCh38
rs973585969 Chr.6: 168308625-168308625 on Build GRCh38
rs377662346 Chr.6: 168308413-168308413 on Build GRCh38
rs781216476 Chr.6: 168308686-168308686 on Build GRCh38
rs1057055877 Chr.6: 168308618-168308618 on Build GRCh38
rs766517269 Chr.6: 168308722-168308722 on Build GRCh38
rs756656016 Chr.6: 168308357-168308357 on Build GRCh38
rs780439375 Chr.6: 168308586-168308586 on Build GRCh38
rs568170901 Chr.6: 168308339-168308339 on Build GRCh38
rs750683098 Chr.6: 168308354-168308354 on Build GRCh38
rs748395420 Chr.6: 168308452-168308452 on Build GRCh38
rs570366910 Chr.6: 168308421-168308421 on Build GRCh38
rs891314047 Chr.6: 168308340-168308340 on Build GRCh38
rs1035597574 Chr.6: 168308453-168308453 on Build GRCh38
rs978880302 Chr.6: 168308477-168308477 on Build GRCh38
rs867445231 Chr.6: 168308678-168308678 on Build GRCh38
rs772353783 Chr.6: 168308634-168308634 on Build GRCh38
rs866825669 Chr.6: 168308495-168308495 on Build GRCh38
rs776701452 Chr.6: 168308626-168308626 on Build GRCh38
rs757463036 Chr.6: 168308674-168308674 on Build GRCh38
rs115241161 Chr.6: 168308593-168308593 on Build GRCh38
rs769524155 Chr.6: 168308675-168308675 on Build GRCh38
rs537304272 Chr.6: 168308438-168308438 on Build GRCh38
rs1020527741 Chr.6: 168308464-168308464 on Build GRCh38
rs912842538 Chr.6: 168308545-168308545 on Build GRCh38
rs552681708 Chr.6: 168308521-168308521 on Build GRCh38
rs778946827 Chr.6: 168308444-168308444 on Build GRCh38
rs542050052 Chr.6: 168308554-168308554 on Build GRCh38
rs1024025428 Chr.6: 168308318-168308318 on Build GRCh38
rs142218608 Chr.6: 168308554-168308555 on Build GRCh38
rs370520754 Chr.6: 168308458-168308458 on Build GRCh38
rs763015294 Chr.6: 168308696-168308696 on Build GRCh38
rs766679635 Chr.6: 168308754-168308754 on Build GRCh38
rs953176594 Chr.6: 168308635-168308635 on Build GRCh38
rs763720009 Chr.6: 168308533-168308533 on Build GRCh38
rs1001511743 Chr.6: 168308611-168308611 on Build GRCh38
rs6925614 Chr.6: 168308705-168308705 on Build GRCh38
rs565295919 Chr.6: 168308729-168308729 on Build GRCh38
rs764133000 Chr.6: 168308538-168308538 on Build GRCh38
rs570083918 Chr.6: 168308459-168308459 on Build GRCh38
rs1037589769 Chr.6: 168308371-168308371 on Build GRCh38
rs1020120816 Chr.6: 168308664-168308664 on Build GRCh38
rs188181495 Chr.6: 168308393-168308393 on Build GRCh38
rs192935251 Chr.6: 168308529-168308529 on Build GRCh38
rs893902173 Chr.6: 168308519-168308519 on Build GRCh38
rs569343262 Chr.6: 168308610-168308610 on Build GRCh38
rs867130647 Chr.6: 168308502-168308502 on Build GRCh38
rs530345058 Chr.6: 168308377-168308377 on Build GRCh38
rs541840837 Chr.6: 168308387-168308387 on Build GRCh38
rs982919768 Chr.6: 168308400-168308400 on Build GRCh38
rs911540065 Chr.6: 168308750-168308750 on Build GRCh38
rs775375348 Chr.6: 168308684-168308684 on Build GRCh38
rs200186315 Chr.6: 168308624-168308624 on Build GRCh38
rs532586396 Chr.6: 168308755-168308755 on Build GRCh38
rs767015702 Chr.6: 168308562-168308562 on Build GRCh38
rs548494411 Chr.6: 168308388-168308388 on Build GRCh38
rs1052183130 Chr.6: 168308500-168308500 on Build GRCh38
rs953090262 Chr.6: 168308725-168308725 on Build GRCh38
rs367909442 Chr.6: 168308651-168308651 on Build GRCh38
rs541416019 Chr.6: 168308756-168308756 on Build GRCh38
rs944248642 Chr.6: 168308368-168308368 on Build GRCh38
rs371278832 Chr.6: 168308316-168308316 on Build GRCh38
rs201301805 Chr.6: 168308338-168308338 on Build GRCh38
rs140583763 Chr.6: 168308595-168308595 on Build GRCh38
rs1003530912 Chr.6: 168308574-168308574 on Build GRCh38
rs376868583 Chr.6: 168308563-168308563 on Build GRCh38
rs374278981 Chr.6: 168308705-168308705 on Build GRCh38
rs370353122 Chr.6: 168308592-168308592 on Build GRCh38
rs1001420026 Chr.6: 168308446-168308446 on Build GRCh38
rs1037236107 Chr.6: 168308557-168308557 on Build GRCh38
rs767045985 Chr.6: 168308713-168308713 on Build GRCh38
rs1045150642 Chr.6: 168308537-168308537 on Build GRCh38
rs769619726 Chr.6: 168308516-168308516 on Build GRCh38
rs756498977 Chr.6: 168308583-168308583 on Build GRCh38
rs41266321 Chr.6: 168308467-168308467 on Build GRCh38
rs201449983 Chr.6: 168308608-168308608 on Build GRCh38
rs759195961 Chr.6: 168308470-168308470 on Build GRCh38
rs774006853 Chr.6: 168308706-168308706 on Build GRCh38
rs907644071 Chr.6: 168308427-168308427 on Build GRCh38
rs921618080 Chr.6: 168308486-168308486 on Build GRCh38

View Full Product Details