Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 193493-193988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20C
Entrez Gene ID
56975
Gene Name
family with sequence similarity 20 member C
Gene Aliases
DMP-4, DMP4, GEF-CK, RNS
UniGene
Hs.134742
Gene Chromosome Location
Chr.7: 192939-260774 on Build GRCh38
Transcripts
NM_020223.3 XM_017012450.1 XM_017012451.1 XM_017012452.1 XM_017012453.1 XM_017012454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6380021 c.371G>A p.R124Q Chr.7: 193570-193570 on Build GRCh38
6048634 c.451_452insC p.G153fs*56 Chr.7: 193650-193651 on Build GRCh38
4908233 c.362T>A p.L121* Chr.7: 193561-193561 on Build GRCh38
6378146 c.401C>A p.P134H Chr.7: 193600-193600 on Build GRCh38
4908259 c.342C>T p.A114A Chr.7: 193541-193541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764540903 Chr.7: 193826-193826 on Build GRCh38
rs961993783 Chr.7: 193920-193920 on Build GRCh38
rs913504706 Chr.7: 193823-193823 on Build GRCh38
rs962245579 Chr.7: 193679-193679 on Build GRCh38
rs935756354 Chr.7: 193953-193953 on Build GRCh38
rs1043070492 Chr.7: 193849-193849 on Build GRCh38
rs980553723 Chr.7: 193733-193733 on Build GRCh38
rs765249975 Chr.7: 193811-193811 on Build GRCh38
rs750593988 Chr.7: 193930-193930 on Build GRCh38
rs973094493 Chr.7: 193711-193711 on Build GRCh38
rs754720957 Chr.7: 193542-193542 on Build GRCh38
rs761779092 Chr.7: 193763-193763 on Build GRCh38
rs1027567275 Chr.7: 193924-193924 on Build GRCh38
rs946254491 Chr.7: 193847-193847 on Build GRCh38
rs543250603 Chr.7: 193887-193887 on Build GRCh38
rs773210635 Chr.7: 193518-193518 on Build GRCh38
rs868137814 Chr.7: 193599-193599 on Build GRCh38
rs747581446 Chr.7: 193636-193636 on Build GRCh38
rs771901909 Chr.7: 193734-193734 on Build GRCh38
rs996631258 Chr.7: 193633-193633 on Build GRCh38
rs918666016 Chr.7: 193563-193563 on Build GRCh38
rs745916292 Chr.7: 193687-193687 on Build GRCh38
rs970907110 Chr.7: 193727-193727 on Build GRCh38
rs1043428625 Chr.7: 193608-193608 on Build GRCh38
rs1029026754 Chr.7: 193639-193639 on Build GRCh38
rs1057293876 Chr.7: 193819-193819 on Build GRCh38
rs781379485 Chr.7: 193680-193680 on Build GRCh38
rs942650241 Chr.7: 193612-193612 on Build GRCh38
rs890511590 Chr.7: 193873-193873 on Build GRCh38
rs904557178 Chr.7: 193855-193855 on Build GRCh38
rs770065356 Chr.7: 193691-193691 on Build GRCh38
rs762471930 Chr.7: 193807-193807 on Build GRCh38
rs777933402 Chr.7: 193625-193625 on Build GRCh38
rs996131660 Chr.7: 193861-193861 on Build GRCh38
rs995195254 Chr.7: 193923-193923 on Build GRCh38
rs374213961 Chr.7: 193951-193952 on Build GRCh38
rs753171820 Chr.7: 193554-193554 on Build GRCh38
rs113842436 Chr.7: 193619-193619 on Build GRCh38
rs759105808 Chr.7: 193781-193781 on Build GRCh38
rs575912804 Chr.7: 193837-193837 on Build GRCh38
rs925749811 Chr.7: 193943-193943 on Build GRCh38
rs537411703 Chr.7: 193578-193578 on Build GRCh38
rs977260732 Chr.7: 193935-193935 on Build GRCh38
rs1025732619 Chr.7: 193752-193752 on Build GRCh38
rs539116395 Chr.7: 193584-193584 on Build GRCh38
rs572411891 Chr.7: 193718-193718 on Build GRCh38
rs764977929 Chr.7: 193546-193546 on Build GRCh38
rs542580647 Chr.7: 193746-193746 on Build GRCh38
rs999915013 Chr.7: 193667-193667 on Build GRCh38
rs945908652 Chr.7: 193928-193928 on Build GRCh38
rs775673987 Chr.7: 193716-193716 on Build GRCh38
rs751845284 Chr.7: 193833-193833 on Build GRCh38
rs201631664 Chr.7: 193799-193799 on Build GRCh38
rs980422861 Chr.7: 193939-193939 on Build GRCh38
rs751592577 Chr.7: 193657-193657 on Build GRCh38
rs770746273 Chr.7: 193774-193774 on Build GRCh38
rs794726946 Chr.7: 193803-193803 on Build GRCh38
rs970854489 Chr.7: 193517-193517 on Build GRCh38
rs969071982 Chr.7: 193719-193719 on Build GRCh38
rs777240362 Chr.7: 193753-193753 on Build GRCh38
rs532155751 Chr.7: 193961-193961 on Build GRCh38
rs772689263 Chr.7: 193842-193843 on Build GRCh38
rs911183872 Chr.7: 193604-193604 on Build GRCh38
rs772838347 Chr.7: 193742-193742 on Build GRCh38
rs996109101 Chr.7: 193824-193824 on Build GRCh38
rs753268141 Chr.7: 193522-193522 on Build GRCh38
rs531384573 Chr.7: 193626-193626 on Build GRCh38
rs976731241 Chr.7: 193905-193905 on Build GRCh38
rs757656546 Chr.7: 193660-193660 on Build GRCh38
rs866515009 Chr.7: 193770-193770 on Build GRCh38
rs1041528504 Chr.7: 193655-193655 on Build GRCh38
rs181853684 Chr.7: 193916-193916 on Build GRCh38
rs945854813 Chr.7: 193642-193642 on Build GRCh38
rs760493570 Chr.7: 193521-193521 on Build GRCh38
rs866592884 Chr.7: 193802-193802 on Build GRCh38
rs776349784 Chr.7: 193780-193780 on Build GRCh38
rs756369436 Chr.7: 193955-193955 on Build GRCh38
rs373625849 Chr.7: 193937-193937 on Build GRCh38
rs749450491 Chr.7: 193717-193717 on Build GRCh38
rs972824224 Chr.7: 193550-193550 on Build GRCh38
rs1050494601 Chr.7: 193868-193868 on Build GRCh38
rs553928524 Chr.7: 193673-193673 on Build GRCh38
rs781543689 Chr.7: 193854-193854 on Build GRCh38

View Full Product Details