Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 256568-257077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20C
Entrez Gene ID
56975
Gene Name
family with sequence similarity 20 member C
Gene Aliases
DMP-4, DMP4, GEF-CK, RNS
UniGene
Hs.134742
Gene Chromosome Location
Chr.7: 192939-260774 on Build GRCh38
Transcripts
NM_020223.3 XM_017012450.1 XM_017012451.1 XM_017012455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7421071 c.1321A>T p.I441F Chr.7: 256721-256721 on Build GRCh38
6351320 c.1416G>A p.T472T Chr.7: 257057-257057 on Build GRCh38
7224893 c.1363+10G>A p.? Chr.7: 256773-256773 on Build GRCh38
1088997 c.1386C>T p.Y462Y Chr.7: 257027-257027 on Build GRCh38
5475038 c.1348T>C p.F450L Chr.7: 256748-256748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941287934 Chr.7: 257002-257002 on Build GRCh38
rs941248139 Chr.7: 256858-256858 on Build GRCh38
rs532421749 Chr.7: 256601-256601 on Build GRCh38
rs144664066 Chr.7: 256750-256750 on Build GRCh38
rs955238974 Chr.7: 256991-256991 on Build GRCh38
rs530956875 Chr.7: 256701-256701 on Build GRCh38
rs1023779289 Chr.7: 256592-256592 on Build GRCh38
rs922401002 Chr.7: 256678-256678 on Build GRCh38
rs756159495 Chr.7: 256698-256698 on Build GRCh38
rs762150603 Chr.7: 256962-256962 on Build GRCh38
rs894237158 Chr.7: 256769-256769 on Build GRCh38
rs376166238 Chr.7: 257000-257000 on Build GRCh38
rs1012673164 Chr.7: 256890-256890 on Build GRCh38
rs796051853 Chr.7: 257003-257003 on Build GRCh38
rs990010145 Chr.7: 256644-256644 on Build GRCh38
rs1009942788 Chr.7: 256617-256617 on Build GRCh38
rs768319683 Chr.7: 256811-256811 on Build GRCh38
rs974360011 Chr.7: 257011-257011 on Build GRCh38
rs899287612 Chr.7: 256597-256597 on Build GRCh38
rs757421711 Chr.7: 256973-256973 on Build GRCh38
rs986193909 Chr.7: 256881-256881 on Build GRCh38
rs1011537176 Chr.7: 256926-256926 on Build GRCh38
rs989482728 Chr.7: 256813-256813 on Build GRCh38
rs894092140 Chr.7: 256864-256864 on Build GRCh38
rs948387893 Chr.7: 257051-257051 on Build GRCh38
rs748939278 Chr.7: 256798-256798 on Build GRCh38
rs868457988 Chr.7: 256719-256719 on Build GRCh38
rs536782006 Chr.7: 256847-256847 on Build GRCh38
rs778956322 Chr.7: 256650-256650 on Build GRCh38
rs141910291 Chr.7: 256814-256814 on Build GRCh38
rs964164814 Chr.7: 256818-256818 on Build GRCh38
rs376430486 Chr.7: 257027-257027 on Build GRCh38
rs541885459 Chr.7: 256925-256925 on Build GRCh38
rs548664628 Chr.7: 256631-256631 on Build GRCh38
rs865798747 Chr.7: 256743-256743 on Build GRCh38
rs1001180164 Chr.7: 256957-256957 on Build GRCh38
rs1013056971 Chr.7: 256772-256772 on Build GRCh38
rs766557527 Chr.7: 256801-256801 on Build GRCh38
rs570988860 Chr.7: 256744-256744 on Build GRCh38
rs1014764557 Chr.7: 257000-257001 on Build GRCh38
rs546072656 Chr.7: 256878-256878 on Build GRCh38
rs138087711 Chr.7: 256614-256614 on Build GRCh38
rs546135270 Chr.7: 256602-256602 on Build GRCh38
rs1050563008 Chr.7: 256762-256762 on Build GRCh38
rs955877838 Chr.7: 256613-256613 on Build GRCh38
rs373380352 Chr.7: 256604-256604 on Build GRCh38
rs754021228 Chr.7: 256904-256904 on Build GRCh38
rs773741100 Chr.7: 256972-256972 on Build GRCh38
rs906739471 Chr.7: 256907-256907 on Build GRCh38
rs920643572 Chr.7: 256729-256729 on Build GRCh38
rs1021783703 Chr.7: 256625-256626 on Build GRCh38
rs187349552 Chr.7: 256596-256596 on Build GRCh38
rs36156504 Chr.7: 256773-256773 on Build GRCh38
rs554308252 Chr.7: 256808-256808 on Build GRCh38
rs141959001 Chr.7: 256705-256705 on Build GRCh38
rs975435451 Chr.7: 256829-256829 on Build GRCh38
rs1003694217 Chr.7: 256922-256922 on Build GRCh38
rs890675096 Chr.7: 256607-256607 on Build GRCh38
rs568534039 Chr.7: 256699-256699 on Build GRCh38
rs762346873 Chr.7: 256638-256638 on Build GRCh38
rs922474475 Chr.7: 256846-256846 on Build GRCh38
rs912016644 Chr.7: 256901-256901 on Build GRCh38
rs895604196 Chr.7: 256964-256964 on Build GRCh38
rs371869318 Chr.7: 256956-256956 on Build GRCh38
rs774671985 Chr.7: 256632-256632 on Build GRCh38
rs755206779 Chr.7: 256706-256706 on Build GRCh38
rs775013594 Chr.7: 256783-256783 on Build GRCh38
rs777751608 Chr.7: 256718-256718 on Build GRCh38
rs908455382 Chr.7: 256853-256853 on Build GRCh38
rs959847727 Chr.7: 256822-256822 on Build GRCh38
rs996742928 Chr.7: 256599-256599 on Build GRCh38
rs757324719 Chr.7: 256753-256753 on Build GRCh38
rs1039558550 Chr.7: 256664-256664 on Build GRCh38
rs377726179 Chr.7: 256649-256649 on Build GRCh38
rs541956072 Chr.7: 256965-256965 on Build GRCh38
rs1045497356 Chr.7: 256906-256906 on Build GRCh38
rs1043086159 Chr.7: 256946-256946 on Build GRCh38
rs1010282171 Chr.7: 256776-256776 on Build GRCh38
rs750557992 Chr.7: 256679-256679 on Build GRCh38
rs36164325 Chr.7: 256591-256591 on Build GRCh38
rs534654982 Chr.7: 256915-256915 on Build GRCh38
rs534326893 Chr.7: 256782-256782 on Build GRCh38
rs764385814 Chr.7: 256624-256624 on Build GRCh38
rs553270499 Chr.7: 256767-256767 on Build GRCh38
rs1001252051 Chr.7: 256805-256805 on Build GRCh38
rs992661306 Chr.7: 256823-256823 on Build GRCh38
rs1009173989 Chr.7: 256611-256611 on Build GRCh38
rs944877644 Chr.7: 256768-256768 on Build GRCh38
rs758891729 Chr.7: 256963-256963 on Build GRCh38
rs150635519 Chr.7: 256850-256850 on Build GRCh38
rs959919676 Chr.7: 256642-256642 on Build GRCh38
rs973464584 Chr.7: 256723-256723 on Build GRCh38
rs756216667 Chr.7: 256791-256791 on Build GRCh38
rs376826867 Chr.7: 256859-256859 on Build GRCh38
rs768006208 Chr.7: 256654-256654 on Build GRCh38
rs1029686859 Chr.7: 256982-256982 on Build GRCh38
rs559698121 Chr.7: 256923-256923 on Build GRCh38
rs778317529 Chr.7: 257001-257001 on Build GRCh38
rs956683757 Chr.7: 256639-256639 on Build GRCh38
rs779217257 Chr.7: 256931-256931 on Build GRCh38

View Full Product Details