Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16792501-16793003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGR2
Entrez Gene ID
10551
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
UniGene
Hs.530009
Gene Chromosome Location
Chr.7: 16791811-16805114 on Build GRCh38
Transcripts
NM_006408.3 XM_005249581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467092 c.497A>C p.K166T Chr.7: 16792939-16792939 on Build GRCh38
236964 c.479-5A>G p.? Chr.7: 16792962-16792962 on Build GRCh38
3636919 c.492G>A p.M164I Chr.7: 16792944-16792944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556565008 Chr.7: 16792672-16792672 on Build GRCh38
rs931859872 Chr.7: 16792812-16792815 on Build GRCh38
rs1051905 Chr.7: 16792684-16792684 on Build GRCh38
rs902896099 Chr.7: 16792930-16792930 on Build GRCh38
rs762996070 Chr.7: 16792868-16792868 on Build GRCh38
rs753830981 Chr.7: 16792709-16792709 on Build GRCh38
rs541219367 Chr.7: 16792906-16792906 on Build GRCh38
rs558712090 Chr.7: 16792538-16792538 on Build GRCh38
rs907735449 Chr.7: 16792717-16792717 on Build GRCh38
rs1051869 Chr.7: 16792851-16792851 on Build GRCh38
rs375328865 Chr.7: 16792807-16792807 on Build GRCh38
rs3173624 Chr.7: 16792811-16792811 on Build GRCh38
rs1051880 Chr.7: 16792809-16792809 on Build GRCh38
rs777605755 Chr.7: 16792961-16792961 on Build GRCh38
rs1051894 Chr.7: 16792766-16792766 on Build GRCh38
rs759351939 Chr.7: 16792743-16792743 on Build GRCh38
rs574640705 Chr.7: 16792564-16792564 on Build GRCh38
rs143032356 Chr.7: 16792910-16792910 on Build GRCh38
rs747674876 Chr.7: 16792956-16792956 on Build GRCh38
rs937650671 Chr.7: 16792539-16792539 on Build GRCh38
rs573471377 Chr.7: 16792857-16792857 on Build GRCh38
rs970507860 Chr.7: 16792776-16792776 on Build GRCh38
rs775748089 Chr.7: 16792967-16792967 on Build GRCh38
rs148410236 Chr.7: 16792796-16792796 on Build GRCh38
rs543437188 Chr.7: 16792813-16792813 on Build GRCh38
rs775599522 Chr.7: 16792866-16792866 on Build GRCh38
rs903315819 Chr.7: 16792651-16792651 on Build GRCh38
rs374504036 Chr.7: 16792963-16792963 on Build GRCh38
rs754532797 Chr.7: 16792920-16792920 on Build GRCh38
rs893578595 Chr.7: 16792569-16792569 on Build GRCh38
rs1050066994 Chr.7: 16792530-16792530 on Build GRCh38
rs560550442 Chr.7: 16792882-16792882 on Build GRCh38
rs892787716 Chr.7: 16792844-16792844 on Build GRCh38
rs767921752 Chr.7: 16792876-16792879 on Build GRCh38
rs538817030 Chr.7: 16792671-16792671 on Build GRCh38
rs554048643 Chr.7: 16792675-16792675 on Build GRCh38
rs540529208 Chr.7: 16792846-16792846 on Build GRCh38
rs753474844 Chr.7: 16792911-16792911 on Build GRCh38
rs572327332 Chr.7: 16792745-16792745 on Build GRCh38
rs772072049 Chr.7: 16792960-16792960 on Build GRCh38
rs756465833 Chr.7: 16792900-16792900 on Build GRCh38
rs914912032 Chr.7: 16792863-16792863 on Build GRCh38
rs750731771 Chr.7: 16792898-16792898 on Build GRCh38
rs554521706 Chr.7: 16792793-16792793 on Build GRCh38
rs1802157 Chr.7: 16792873-16792873 on Build GRCh38
rs570993722 Chr.7: 16792666-16792666 on Build GRCh38
rs763224709 Chr.7: 16792980-16792980 on Build GRCh38
rs548525357 Chr.7: 16792553-16792553 on Build GRCh38
rs902674660 Chr.7: 16792574-16792574 on Build GRCh38
rs892891750 Chr.7: 16792537-16792537 on Build GRCh38
rs1008496545 Chr.7: 16792657-16792657 on Build GRCh38
rs1009312256 Chr.7: 16792596-16792596 on Build GRCh38
rs997283286 Chr.7: 16792695-16792695 on Build GRCh38
rs368448343 Chr.7: 16792966-16792966 on Build GRCh38
rs564805090 Chr.7: 16792819-16792819 on Build GRCh38
rs770021368 Chr.7: 16792862-16792862 on Build GRCh38
rs760395687 Chr.7: 16792858-16792858 on Build GRCh38
rs778625347 Chr.7: 16792936-16792936 on Build GRCh38
rs1026979023 Chr.7: 16792697-16792697 on Build GRCh38
rs148204661 Chr.7: 16792950-16792950 on Build GRCh38
rs369355847 Chr.7: 16792854-16792854 on Build GRCh38
rs77364030 Chr.7: 16792618-16792618 on Build GRCh38
rs767923511 Chr.7: 16792885-16792885 on Build GRCh38
rs762361024 Chr.7: 16792881-16792881 on Build GRCh38
rs999650664 Chr.7: 16792945-16792945 on Build GRCh38
rs113723926 Chr.7: 16792658-16792658 on Build GRCh38
rs999763937 Chr.7: 16792575-16792575 on Build GRCh38
rs8071 Chr.7: 16792758-16792758 on Build GRCh38
rs1050118716 Chr.7: 16792812-16792812 on Build GRCh38
rs750452130 Chr.7: 16792898-16792898 on Build GRCh38
rs1041457820 Chr.7: 16792848-16792850 on Build GRCh38
rs752053196 Chr.7: 16792878-16792878 on Build GRCh38
rs866903433 Chr.7: 16792619-16792619 on Build GRCh38
rs940544512 Chr.7: 16792737-16792737 on Build GRCh38
rs28945080 Chr.7: 16792565-16792565 on Build GRCh38

View Full Product Details