Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20643093-20643609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB5
Entrez Gene ID
340273
Gene Name
ATP binding cassette subfamily B member 5
Gene Aliases
ABCB5alpha, ABCB5beta, EST422562
UniGene
Hs.404102
Gene Chromosome Location
Chr.7: 20615622-20757014 on Build GRCh38
Transcripts
NM_001163941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5773583 c.644C>A p.P215H Chr.7: 20643598-20643598 on Build GRCh38
5062370 c.426_427insT p.H145fs*12 Chr.7: 20643295-20643296 on Build GRCh38
7353574 c.370C>G p.Q124E Chr.7: 20643239-20643239 on Build GRCh38
5757024 c.638C>T p.T213M Chr.7: 20643592-20643592 on Build GRCh38
485174 c.412A>C p.R138R Chr.7: 20643281-20643281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371630547 Chr.7: 20643373-20643373 on Build GRCh38
rs750666662 Chr.7: 20643371-20643371 on Build GRCh38
rs772958187 Chr.7: 20643336-20643336 on Build GRCh38
rs763832811 Chr.7: 20643160-20643160 on Build GRCh38
rs753193962 Chr.7: 20643486-20643486 on Build GRCh38
rs753647911 Chr.7: 20643287-20643287 on Build GRCh38
rs757514065 Chr.7: 20643302-20643302 on Build GRCh38
rs369918100 Chr.7: 20643423-20643423 on Build GRCh38
rs765461658 Chr.7: 20643538-20643538 on Build GRCh38
rs372132207 Chr.7: 20643315-20643315 on Build GRCh38
rs766397154 Chr.7: 20643184-20643184 on Build GRCh38
rs762746021 Chr.7: 20643445-20643445 on Build GRCh38
rs7349983 Chr.7: 20643581-20643581 on Build GRCh38
rs776042795 Chr.7: 20643415-20643415 on Build GRCh38
rs548365815 Chr.7: 20643249-20643249 on Build GRCh38
rs751894548 Chr.7: 20643559-20643559 on Build GRCh38
rs117477399 Chr.7: 20643366-20643366 on Build GRCh38
rs374004335 Chr.7: 20643337-20643337 on Build GRCh38
rs778345637 Chr.7: 20643494-20643494 on Build GRCh38
rs946181751 Chr.7: 20643281-20643281 on Build GRCh38
rs951368928 Chr.7: 20643582-20643582 on Build GRCh38
rs181716362 Chr.7: 20643326-20643326 on Build GRCh38
rs767702759 Chr.7: 20643356-20643357 on Build GRCh38
rs756158660 Chr.7: 20643179-20643179 on Build GRCh38
rs557169551 Chr.7: 20643413-20643413 on Build GRCh38
rs190460902 Chr.7: 20643270-20643270 on Build GRCh38
rs776189414 Chr.7: 20643340-20643340 on Build GRCh38
rs754794923 Chr.7: 20643376-20643377 on Build GRCh38
rs891481152 Chr.7: 20643484-20643484 on Build GRCh38
rs200714278 Chr.7: 20643533-20643533 on Build GRCh38
rs372654621 Chr.7: 20643451-20643451 on Build GRCh38
rs780852333 Chr.7: 20643500-20643500 on Build GRCh38
rs759547568 Chr.7: 20643535-20643535 on Build GRCh38
rs751342832 Chr.7: 20643454-20643454 on Build GRCh38
rs772896092 Chr.7: 20643273-20643273 on Build GRCh38
rs186728686 Chr.7: 20643327-20643327 on Build GRCh38
rs539506410 Chr.7: 20643519-20643519 on Build GRCh38
rs950647261 Chr.7: 20643216-20643216 on Build GRCh38
rs777577873 Chr.7: 20643404-20643404 on Build GRCh38
rs753950706 Chr.7: 20643390-20643390 on Build GRCh38
rs138449195 Chr.7: 20643574-20643574 on Build GRCh38
rs770284056 Chr.7: 20643145-20643145 on Build GRCh38
rs753883618 Chr.7: 20643478-20643478 on Build GRCh38
rs768578610 Chr.7: 20643532-20643532 on Build GRCh38
rs755043625 Chr.7: 20643479-20643479 on Build GRCh38
rs770321564 Chr.7: 20643524-20643524 on Build GRCh38
rs199687406 Chr.7: 20643253-20643253 on Build GRCh38
rs755750745 Chr.7: 20643576-20643576 on Build GRCh38
rs904548656 Chr.7: 20643124-20643124 on Build GRCh38
rs905771934 Chr.7: 20643420-20643420 on Build GRCh38
rs886296579 Chr.7: 20643394-20643394 on Build GRCh38
rs186136997 Chr.7: 20643141-20643141 on Build GRCh38
rs368226947 Chr.7: 20643288-20643288 on Build GRCh38
rs767761999 Chr.7: 20643365-20643365 on Build GRCh38
rs771871738 Chr.7: 20643335-20643335 on Build GRCh38
rs757604214 Chr.7: 20643560-20643560 on Build GRCh38
rs267601449 Chr.7: 20643513-20643513 on Build GRCh38
rs77409024 Chr.7: 20643332-20643332 on Build GRCh38
rs777940445 Chr.7: 20643125-20643125 on Build GRCh38
rs148357630 Chr.7: 20643148-20643148 on Build GRCh38
rs745404517 Chr.7: 20643501-20643501 on Build GRCh38
rs926227488 Chr.7: 20643551-20643551 on Build GRCh38
rs766371098 Chr.7: 20643282-20643282 on Build GRCh38
rs752549151 Chr.7: 20643408-20643408 on Build GRCh38
rs372749249 Chr.7: 20643238-20643238 on Build GRCh38
rs759066207 Chr.7: 20643419-20643419 on Build GRCh38
rs887634404 Chr.7: 20643211-20643211 on Build GRCh38
rs868015590 Chr.7: 20643462-20643462 on Build GRCh38
rs777133464 Chr.7: 20643134-20643134 on Build GRCh38
rs191118671 Chr.7: 20643399-20643399 on Build GRCh38
rs770339087 Chr.7: 20643251-20643251 on Build GRCh38
rs757454697 Chr.7: 20643245-20643245 on Build GRCh38
rs141511940 Chr.7: 20643269-20643269 on Build GRCh38
rs764870010 Chr.7: 20643390-20643390 on Build GRCh38
rs752539619 Chr.7: 20643352-20643352 on Build GRCh38
rs577483576 Chr.7: 20643126-20643126 on Build GRCh38
rs763209984 Chr.7: 20643156-20643156 on Build GRCh38
rs767080146 Chr.7: 20643172-20643172 on Build GRCh38
rs761412272 Chr.7: 20643169-20643169 on Build GRCh38
rs374303845 Chr.7: 20643277-20643277 on Build GRCh38
rs745974541 Chr.7: 20643323-20643323 on Build GRCh38
rs1047512159 Chr.7: 20643199-20643199 on Build GRCh38
rs775420502 Chr.7: 20643522-20643522 on Build GRCh38
rs780716908 Chr.7: 20643570-20643570 on Build GRCh38
rs369274708 Chr.7: 20643180-20643180 on Build GRCh38
rs754685763 Chr.7: 20643212-20643212 on Build GRCh38
rs17143212 Chr.7: 20643261-20643261 on Build GRCh38
rs755485786 Chr.7: 20643395-20643395 on Build GRCh38
rs763281218 Chr.7: 20643349-20643349 on Build GRCh38
rs374933535 Chr.7: 20643534-20643534 on Build GRCh38
rs202100779 Chr.7: 20643234-20643234 on Build GRCh38
rs941855336 Chr.7: 20643403-20643403 on Build GRCh38
rs758956710 Chr.7: 20643487-20643487 on Build GRCh38
rs565994506 Chr.7: 20643279-20643279 on Build GRCh38
rs747180120 Chr.7: 20643217-20643217 on Build GRCh38
rs746713945 Chr.7: 20643410-20643410 on Build GRCh38
rs150545949 Chr.7: 20643547-20643547 on Build GRCh38
rs572751870 Chr.7: 20643546-20643546 on Build GRCh38
rs1021357784 Chr.7: 20643509-20643509 on Build GRCh38
rs752816084 Chr.7: 20643544-20643544 on Build GRCh38
rs146831815 Chr.7: 20643583-20643583 on Build GRCh38
rs769194692 Chr.7: 20643331-20643331 on Build GRCh38
rs779566714 Chr.7: 20643480-20643480 on Build GRCh38
rs548933985 Chr.7: 20643325-20643325 on Build GRCh38
rs779426149 Chr.7: 20643396-20643396 on Build GRCh38
rs775369630 Chr.7: 20643441-20643441 on Build GRCh38
rs866062576 Chr.7: 20643129-20643129 on Build GRCh38
rs570793675 Chr.7: 20643324-20643324 on Build GRCh38
rs375602893 Chr.7: 20643477-20643477 on Build GRCh38
rs918665949 Chr.7: 20643159-20643159 on Build GRCh38
rs748553729 Chr.7: 20643398-20643398 on Build GRCh38
rs1056795194 Chr.7: 20643466-20643466 on Build GRCh38
rs869245984 Chr.7: 20643392-20643392 on Build GRCh38
rs369460474 Chr.7: 20643496-20643496 on Build GRCh38
rs765193743 Chr.7: 20643350-20643350 on Build GRCh38
rs546157805 Chr.7: 20643146-20643146 on Build GRCh38
rs62456140 Chr.7: 20643409-20643409 on Build GRCh38
rs377268116 Chr.7: 20643162-20643162 on Build GRCh38
rs895883976 Chr.7: 20643154-20643154 on Build GRCh38
rs569006123 Chr.7: 20643499-20643499 on Build GRCh38
rs375764523 Chr.7: 20643250-20643250 on Build GRCh38
rs559696180 Chr.7: 20643144-20643144 on Build GRCh38
rs1046233553 Chr.7: 20643370-20643370 on Build GRCh38
rs774068423 Chr.7: 20643262-20643262 on Build GRCh38
rs1035362873 Chr.7: 20643252-20643252 on Build GRCh38
rs758361200 Chr.7: 20643570-20643571 on Build GRCh38

View Full Product Details