Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 21543113-21543608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH11
Entrez Gene ID
8701
Gene Name
dynein axonemal heavy chain 11
Gene Aliases
CILD7, DNAHBL, DNAHC11, DNHBL, DPL11
UniGene
Hs.520245
Gene Chromosome Location
Chr.7: 21543215-21901568 on Build GRCh38
Transcripts
NM_001277115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3394665 c.296C>T p.P99L Chr.7: 21543541-21543541 on Build GRCh38
5795046 c.7G>T p.A3S Chr.7: 21543252-21543252 on Build GRCh38
6222267 c.134C>A p.A45E Chr.7: 21543379-21543379 on Build GRCh38
6646152 c.330G>A p.G110G Chr.7: 21543575-21543575 on Build GRCh38
7364754 c.146G>T p.R49I Chr.7: 21543391-21543391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978373825 Chr.7: 21543413-21543413 on Build GRCh38
rs72655967 Chr.7: 21543303-21543303 on Build GRCh38
rs1016139987 Chr.7: 21543518-21543518 on Build GRCh38
rs1042644843 Chr.7: 21543229-21543229 on Build GRCh38
rs969842140 Chr.7: 21543430-21543430 on Build GRCh38
rs918116615 Chr.7: 21543381-21543381 on Build GRCh38
rs930834798 Chr.7: 21543340-21543340 on Build GRCh38
rs2285944 Chr.7: 21543346-21543346 on Build GRCh38
rs201804029 Chr.7: 21543318-21543318 on Build GRCh38
rs919406417 Chr.7: 21543314-21543314 on Build GRCh38
rs1000525671 Chr.7: 21543292-21543292 on Build GRCh38
rs754495872 Chr.7: 21543183-21543184 on Build GRCh38
rs775754675 Chr.7: 21543336-21543336 on Build GRCh38
rs1004777420 Chr.7: 21543333-21543333 on Build GRCh38
rs779462489 Chr.7: 21543561-21543561 on Build GRCh38
rs750522660 Chr.7: 21543515-21543515 on Build GRCh38
rs372762132 Chr.7: 21543508-21543508 on Build GRCh38
rs771623831 Chr.7: 21543457-21543457 on Build GRCh38
rs755966311 Chr.7: 21543264-21543264 on Build GRCh38
rs952333583 Chr.7: 21543575-21543575 on Build GRCh38
rs6966525 Chr.7: 21543433-21543433 on Build GRCh38
rs1017341981 Chr.7: 21543143-21543143 on Build GRCh38
rs573879149 Chr.7: 21543447-21543447 on Build GRCh38
rs774172154 Chr.7: 21543438-21543438 on Build GRCh38
rs992756191 Chr.7: 21543379-21543379 on Build GRCh38
rs1022774244 Chr.7: 21543214-21543214 on Build GRCh38
rs767753234 Chr.7: 21543509-21543509 on Build GRCh38
rs900261273 Chr.7: 21543445-21543445 on Build GRCh38
rs1056603995 Chr.7: 21543189-21543189 on Build GRCh38
rs1051931486 Chr.7: 21543464-21543464 on Build GRCh38
rs576486289 Chr.7: 21543533-21543533 on Build GRCh38
rs772648210 Chr.7: 21543453-21543453 on Build GRCh38
rs959617971 Chr.7: 21543536-21543536 on Build GRCh38
rs898030150 Chr.7: 21543159-21543159 on Build GRCh38
rs1007614931 Chr.7: 21543184-21543184 on Build GRCh38
rs950600455 Chr.7: 21543174-21543174 on Build GRCh38
rs748502650 Chr.7: 21543570-21543570 on Build GRCh38
rs756156734 Chr.7: 21543521-21543521 on Build GRCh38
rs997355592 Chr.7: 21543163-21543163 on Build GRCh38
rs945587987 Chr.7: 21543227-21543227 on Build GRCh38
rs943142146 Chr.7: 21543351-21543351 on Build GRCh38
rs560422553 Chr.7: 21543504-21543504 on Build GRCh38
rs1030665124 Chr.7: 21543172-21543172 on Build GRCh38
rs777975395 Chr.7: 21543406-21543406 on Build GRCh38
rs899054221 Chr.7: 21543520-21543520 on Build GRCh38
rs771726068 Chr.7: 21543452-21543452 on Build GRCh38
rs749678146 Chr.7: 21543434-21543434 on Build GRCh38
rs751721373 Chr.7: 21543422-21543422 on Build GRCh38
rs398123604 Chr.7: 21543345-21543346 on Build GRCh38
rs745842220 Chr.7: 21543313-21543313 on Build GRCh38
rs542513475 Chr.7: 21543458-21543458 on Build GRCh38
rs886309783 Chr.7: 21543467-21543467 on Build GRCh38
rs1010979498 Chr.7: 21543202-21543202 on Build GRCh38
rs938181972 Chr.7: 21543188-21543188 on Build GRCh38
rs908524160 Chr.7: 21543181-21543181 on Build GRCh38
rs757349371 Chr.7: 21543424-21543424 on Build GRCh38
rs945514775 Chr.7: 21543383-21543383 on Build GRCh38
rs143954803 Chr.7: 21543186-21543186 on Build GRCh38
rs995927629 Chr.7: 21543531-21543531 on Build GRCh38
rs767572611 Chr.7: 21543215-21543215 on Build GRCh38
rs771680598 Chr.7: 21543578-21543578 on Build GRCh38
rs537596387 Chr.7: 21543352-21543352 on Build GRCh38
rs939584907 Chr.7: 21543134-21543134 on Build GRCh38
rs964935835 Chr.7: 21543145-21543145 on Build GRCh38
rs773363827 Chr.7: 21543579-21543579 on Build GRCh38
rs754826899 Chr.7: 21543351-21543353 on Build GRCh38
rs758658916 Chr.7: 21543276-21543276 on Build GRCh38
rs749766034 Chr.7: 21543543-21543543 on Build GRCh38
rs72655966 Chr.7: 21543219-21543219 on Build GRCh38
rs755427308 Chr.7: 21543559-21543559 on Build GRCh38
rs531840083 Chr.7: 21543180-21543180 on Build GRCh38
rs746309042 Chr.7: 21543323-21543323 on Build GRCh38
rs770221913 Chr.7: 21543324-21543324 on Build GRCh38
rs746528555 Chr.7: 21543187-21543187 on Build GRCh38
rs981140256 Chr.7: 21543228-21543228 on Build GRCh38
rs763383078 Chr.7: 21543339-21543339 on Build GRCh38
rs2285942 Chr.7: 21543299-21543299 on Build GRCh38
rs759339238 Chr.7: 21543478-21543478 on Build GRCh38
rs772913999 Chr.7: 21543583-21543583 on Build GRCh38
rs2285943 Chr.7: 21543345-21543345 on Build GRCh38
rs992285091 Chr.7: 21543540-21543540 on Build GRCh38
rs369285943 Chr.7: 21543440-21543440 on Build GRCh38
rs571142873 Chr.7: 21543428-21543428 on Build GRCh38
rs557408462 Chr.7: 21543382-21543382 on Build GRCh38
rs753741123 Chr.7: 21543465-21543465 on Build GRCh38
rs1052997441 Chr.7: 21543410-21543410 on Build GRCh38
rs753645186 Chr.7: 21543270-21543270 on Build GRCh38
rs553424997 Chr.7: 21543185-21543185 on Build GRCh38
rs886038455 Chr.7: 21543232-21543232 on Build GRCh38
rs756810530 Chr.7: 21543196-21543196 on Build GRCh38
rs750407994 Chr.7: 21543376-21543376 on Build GRCh38
rs957303519 Chr.7: 21543152-21543152 on Build GRCh38
rs781089233 Chr.7: 21543425-21543425 on Build GRCh38
rs978215170 Chr.7: 21543567-21543567 on Build GRCh38
rs772333047 Chr.7: 21543378-21543378 on Build GRCh38
rs779664174 Chr.7: 21543266-21543266 on Build GRCh38
rs769087304 Chr.7: 21543436-21543436 on Build GRCh38
rs966907044 Chr.7: 21543547-21543547 on Build GRCh38
rs753634502 Chr.7: 21543397-21543397 on Build GRCh38
rs748250611 Chr.7: 21543371-21543371 on Build GRCh38
rs556570369 Chr.7: 21543506-21543506 on Build GRCh38
rs1039233944 Chr.7: 21543298-21543298 on Build GRCh38
rs886038461 Chr.7: 21543542-21543542 on Build GRCh38

View Full Product Details