Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 21559430-21559949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH11
Entrez Gene ID
8701
Gene Name
dynein axonemal heavy chain 11
Gene Aliases
CILD7, DNAHBL, DNAHC11, DNHBL, DPL11
UniGene
Hs.520245
Gene Chromosome Location
Chr.7: 21543215-21901568 on Build GRCh38
Transcripts
NM_001277115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661484 c.801G>T p.L267F Chr.7: 21559711-21559711 on Build GRCh38
6371204 c.703A>G p.N235D Chr.7: 21559613-21559613 on Build GRCh38
4448403 c.734C>T p.S245F Chr.7: 21559644-21559644 on Build GRCh38
3394667 c.699G>A p.P233P Chr.7: 21559609-21559609 on Build GRCh38
3025308 c.793C>G p.R265G Chr.7: 21559703-21559703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376917588 Chr.7: 21559649-21559649 on Build GRCh38
rs750214113 Chr.7: 21559638-21559638 on Build GRCh38
rs375392320 Chr.7: 21559903-21559903 on Build GRCh38
rs752866813 Chr.7: 21559743-21559743 on Build GRCh38
rs878854447 Chr.7: 21559774-21559774 on Build GRCh38
rs192581558 Chr.7: 21559563-21559563 on Build GRCh38
rs185273259 Chr.7: 21559768-21559768 on Build GRCh38
rs762496532 Chr.7: 21559838-21559838 on Build GRCh38
rs761709401 Chr.7: 21559724-21559724 on Build GRCh38
rs72655974 Chr.7: 21559832-21559832 on Build GRCh38
rs574585676 Chr.7: 21559589-21559589 on Build GRCh38
rs768404895 Chr.7: 21559785-21559785 on Build GRCh38
rs766063976 Chr.7: 21559626-21559628 on Build GRCh38
rs758504634 Chr.7: 21559744-21559744 on Build GRCh38
rs752311298 Chr.7: 21559616-21559616 on Build GRCh38
rs767364968 Chr.7: 21559801-21559801 on Build GRCh38
rs371923630 Chr.7: 21559735-21559735 on Build GRCh38
rs751479427 Chr.7: 21559634-21559634 on Build GRCh38
rs776909112 Chr.7: 21559688-21559688 on Build GRCh38
rs533700730 Chr.7: 21559799-21559799 on Build GRCh38
rs751575627 Chr.7: 21559721-21559721 on Build GRCh38
rs769397639 Chr.7: 21559703-21559703 on Build GRCh38
rs747087320 Chr.7: 21559755-21559755 on Build GRCh38
rs746865163 Chr.7: 21559564-21559564 on Build GRCh38
rs189000268 Chr.7: 21559637-21559637 on Build GRCh38
rs750255266 Chr.7: 21559733-21559733 on Build GRCh38
rs748215520 Chr.7: 21559580-21559580 on Build GRCh38
rs905592352 Chr.7: 21559544-21559544 on Build GRCh38
rs764129443 Chr.7: 21559622-21559622 on Build GRCh38
rs1047426704 Chr.7: 21559481-21559481 on Build GRCh38
rs1040888811 Chr.7: 21559776-21559776 on Build GRCh38
rs919600272 Chr.7: 21559471-21559471 on Build GRCh38
rs908898940 Chr.7: 21559700-21559700 on Build GRCh38
rs929568749 Chr.7: 21559473-21559473 on Build GRCh38
rs1003135993 Chr.7: 21559920-21559920 on Build GRCh38
rs200454446 Chr.7: 21559605-21559605 on Build GRCh38
rs1044099430 Chr.7: 21559536-21559536 on Build GRCh38
rs779122799 Chr.7: 21559741-21559741 on Build GRCh38
rs374505775 Chr.7: 21559716-21559716 on Build GRCh38
rs142908581 Chr.7: 21559546-21559546 on Build GRCh38
rs770222065 Chr.7: 21559762-21559762 on Build GRCh38
rs746574838 Chr.7: 21559566-21559567 on Build GRCh38
rs768896027 Chr.7: 21559576-21559576 on Build GRCh38
rs756933485 Chr.7: 21559567-21559567 on Build GRCh38
rs536865462 Chr.7: 21559552-21559552 on Build GRCh38
rs770628646 Chr.7: 21559703-21559704 on Build GRCh38
rs887408100 Chr.7: 21559482-21559482 on Build GRCh38
rs777870588 Chr.7: 21559746-21559746 on Build GRCh38
rs759930423 Chr.7: 21559694-21559694 on Build GRCh38
rs74792549 Chr.7: 21559891-21559891 on Build GRCh38
rs192989152 Chr.7: 21559704-21559704 on Build GRCh38
rs10950854 Chr.7: 21559615-21559615 on Build GRCh38
rs189585783 Chr.7: 21559855-21559855 on Build GRCh38
rs931183931 Chr.7: 21559474-21559474 on Build GRCh38
rs191519341 Chr.7: 21559925-21559925 on Build GRCh38
rs540942795 Chr.7: 21559819-21559819 on Build GRCh38
rs778274749 Chr.7: 21559679-21559679 on Build GRCh38
rs772084560 Chr.7: 21559667-21559667 on Build GRCh38
rs757094396 Chr.7: 21559635-21559635 on Build GRCh38
rs375029778 Chr.7: 21559822-21559822 on Build GRCh38
rs781372148 Chr.7: 21559766-21559767 on Build GRCh38
rs376910187 Chr.7: 21559569-21559569 on Build GRCh38
rs774347191 Chr.7: 21559579-21559579 on Build GRCh38
rs923782426 Chr.7: 21559781-21559781 on Build GRCh38
rs72655973 Chr.7: 21559594-21559594 on Build GRCh38
rs1011605149 Chr.7: 21559849-21559849 on Build GRCh38
rs771437656 Chr.7: 21559683-21559683 on Build GRCh38
rs1049684674 Chr.7: 21559545-21559545 on Build GRCh38
rs748907119 Chr.7: 21559771-21559771 on Build GRCh38
rs893097751 Chr.7: 21559830-21559830 on Build GRCh38
rs1005936158 Chr.7: 21559495-21559495 on Build GRCh38
rs773145026 Chr.7: 21559802-21559802 on Build GRCh38
rs755215984 Chr.7: 21559648-21559648 on Build GRCh38
rs368102189 Chr.7: 21559757-21559757 on Build GRCh38
rs981629773 Chr.7: 21559625-21559625 on Build GRCh38
rs375023124 Chr.7: 21559756-21559756 on Build GRCh38
rs777561596 Chr.7: 21559557-21559558 on Build GRCh38
rs760409867 Chr.7: 21559813-21559813 on Build GRCh38
rs373971291 Chr.7: 21559609-21559609 on Build GRCh38
rs533662540 Chr.7: 21559879-21559884 on Build GRCh38
rs564646990 Chr.7: 21559795-21559795 on Build GRCh38
rs747435111 Chr.7: 21559682-21559682 on Build GRCh38
rs746285171 Chr.7: 21559817-21559821 on Build GRCh38
rs763644686 Chr.7: 21559718-21559718 on Build GRCh38
rs776376992 Chr.7: 21559608-21559608 on Build GRCh38
rs368459116 Chr.7: 21559727-21559727 on Build GRCh38
rs560735811 Chr.7: 21559888-21559888 on Build GRCh38

View Full Product Details